Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 74379325-74379840 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EREG
Entrez Gene ID
2069
Gene Name
epiregulin
Gene Aliases
EPR, ER, Ep
UniGene
Hs.115263
Gene Chromosome Location
Chr.4: 74365143-74388760 on Build GRCh38
Transcripts
NM_001432.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1619064 c.68-1G>A p.? Chr.4: 74379447-74379447 on Build GRCh38
6376756 c.80T>C p.L27P Chr.4: 74379460-74379460 on Build GRCh38
6168073 c.77T>C p.L26P Chr.4: 74379457-74379457 on Build GRCh38
109166 c.136G>A p.D46N Chr.4: 74379516-74379516 on Build GRCh38
32225 c.125G>C p.G42A Chr.4: 74379505-74379505 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs868644900 Chr.4: 74379531-74379531 on Build GRCh38
rs745452926 Chr.4: 74379413-74379413 on Build GRCh38
rs965966890 Chr.4: 74379719-74379719 on Build GRCh38
rs746275124 Chr.4: 74379542-74379542 on Build GRCh38
rs966623110 Chr.4: 74379452-74379452 on Build GRCh38
rs573972075 Chr.4: 74379717-74379717 on Build GRCh38
rs563838917 Chr.4: 74379813-74379813 on Build GRCh38
rs758601900 Chr.4: 74379496-74379496 on Build GRCh38
rs774261552 Chr.4: 74379574-74379574 on Build GRCh38
rs748763071 Chr.4: 74379545-74379545 on Build GRCh38
rs10016047 Chr.4: 74379625-74379625 on Build GRCh38
rs750427842 Chr.4: 74379618-74379618 on Build GRCh38
rs769491007 Chr.4: 74379424-74379424 on Build GRCh38
rs184692801 Chr.4: 74379785-74379785 on Build GRCh38
rs7660209 Chr.4: 74379626-74379626 on Build GRCh38
rs975975594 Chr.4: 74379725-74379725 on Build GRCh38
rs982768387 Chr.4: 74379707-74379707 on Build GRCh38
rs752776909 Chr.4: 74379487-74379487 on Build GRCh38
rs191813322 Chr.4: 74379592-74379592 on Build GRCh38
rs779563514 Chr.4: 74379372-74379372 on Build GRCh38
rs200556662 Chr.4: 74379449-74379449 on Build GRCh38
rs768871543 Chr.4: 74379601-74379601 on Build GRCh38
rs141527002 Chr.4: 74379461-74379461 on Build GRCh38
rs538432993 Chr.4: 74379520-74379520 on Build GRCh38
rs1043503548 Chr.4: 74379382-74379382 on Build GRCh38
rs139770823 Chr.4: 74379816-74379816 on Build GRCh38
rs912559510 Chr.4: 74379718-74379718 on Build GRCh38
rs773064799 Chr.4: 74379482-74379483 on Build GRCh38
rs896781258 Chr.4: 74379518-74379518 on Build GRCh38
rs762982346 Chr.4: 74379499-74379499 on Build GRCh38
rs762469076 Chr.4: 74379429-74379429 on Build GRCh38
rs1020857176 Chr.4: 74379421-74379421 on Build GRCh38
rs747448119 Chr.4: 74379481-74379481 on Build GRCh38
rs771242923 Chr.4: 74379403-74379403 on Build GRCh38
rs781343689 Chr.4: 74379535-74379535 on Build GRCh38
rs1043939168 Chr.4: 74379763-74379763 on Build GRCh38
rs537510492 Chr.4: 74379603-74379603 on Build GRCh38
rs571711516 Chr.4: 74379365-74379365 on Build GRCh38
rs887904426 Chr.4: 74379726-74379726 on Build GRCh38
rs757794241 Chr.4: 74379528-74379528 on Build GRCh38
rs921822584 Chr.4: 74379727-74379727 on Build GRCh38
rs187400567 Chr.4: 74379475-74379475 on Build GRCh38
rs777295975 Chr.4: 74379664-74379664 on Build GRCh38
rs1045357944 Chr.4: 74379815-74379815 on Build GRCh38
rs567213096 Chr.4: 74379430-74379430 on Build GRCh38
rs1052135111 Chr.4: 74379802-74379802 on Build GRCh38
rs79727217 Chr.4: 74379803-74379803 on Build GRCh38
rs141587819 Chr.4: 74379516-74379516 on Build GRCh38
rs768220381 Chr.4: 74379558-74379558 on Build GRCh38
rs1052084487 Chr.4: 74379514-74379514 on Build GRCh38
rs919512677 Chr.4: 74379681-74379681 on Build GRCh38
rs68107539 Chr.4: 74379621-74379621 on Build GRCh38
rs778394820 Chr.4: 74379515-74379515 on Build GRCh38
rs557859793 Chr.4: 74379576-74379576 on Build GRCh38
rs747357835 Chr.4: 74379401-74379401 on Build GRCh38
rs925152892 Chr.4: 74379488-74379488 on Build GRCh38
rs1037126911 Chr.4: 74379627-74379627 on Build GRCh38
rs775168602 Chr.4: 74379544-74379544 on Build GRCh38
rs548719774 Chr.4: 74379428-74379428 on Build GRCh38
rs999231158 Chr.4: 74379467-74379467 on Build GRCh38
rs541401326 Chr.4: 74379755-74379755 on Build GRCh38
rs776332514 Chr.4: 74379412-74379412 on Build GRCh38
rs769348496 Chr.4: 74379462-74379462 on Build GRCh38
rs951531427 Chr.4: 74379694-74379694 on Build GRCh38
rs769384011 Chr.4: 74379543-74379543 on Build GRCh38
rs56197135 Chr.4: 74379779-74379779 on Build GRCh38
rs1023859368 Chr.4: 74379562-74379562 on Build GRCh38
rs761666107 Chr.4: 74379459-74379459 on Build GRCh38
rs931850396 Chr.4: 74379734-74379734 on Build GRCh38
rs929576267 Chr.4: 74379706-74379706 on Build GRCh38
rs369290734 Chr.4: 74379468-74379468 on Build GRCh38

View Full Product Details