Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 74380642-74381140 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EREG
Entrez Gene ID
2069
Gene Name
epiregulin
Gene Aliases
EPR, ER, Ep
UniGene
Hs.115263
Gene Chromosome Location
Chr.4: 74365143-74388760 on Build GRCh38
Transcripts
NM_001432.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2831473 c.238C>G p.Q80E Chr.4: 74381097-74381097 on Build GRCh38
6168072 c.201G>A p.K67K Chr.4: 74381060-74381060 on Build GRCh38
4386775 c.237A>G p.G79G Chr.4: 74381096-74381096 on Build GRCh38
6100979 c.189G>A p.V63V Chr.4: 74381048-74381048 on Build GRCh38
4125931 c.276C>T p.C92C Chr.4: 74381135-74381135 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs557627524 Chr.4: 74380686-74380686 on Build GRCh38
rs1033877220 Chr.4: 74380672-74380672 on Build GRCh38
rs371135213 Chr.4: 74381069-74381069 on Build GRCh38
rs773562293 Chr.4: 74380744-74380744 on Build GRCh38
rs543387942 Chr.4: 74380787-74380787 on Build GRCh38
rs113254970 Chr.4: 74381072-74381072 on Build GRCh38
rs765534458 Chr.4: 74381015-74381015 on Build GRCh38
rs201201335 Chr.4: 74380991-74380991 on Build GRCh38
rs149770501 Chr.4: 74380792-74380792 on Build GRCh38
rs767497744 Chr.4: 74380981-74380981 on Build GRCh38
rs751662634 Chr.4: 74381031-74381031 on Build GRCh38
rs942678402 Chr.4: 74380707-74380707 on Build GRCh38
rs758549905 Chr.4: 74381018-74381018 on Build GRCh38
rs984234950 Chr.4: 74380684-74380684 on Build GRCh38
rs891808399 Chr.4: 74380881-74380881 on Build GRCh38
rs922834209 Chr.4: 74380798-74380798 on Build GRCh38
rs908212315 Chr.4: 74380697-74380697 on Build GRCh38
rs763197664 Chr.4: 74381107-74381107 on Build GRCh38
rs1031227852 Chr.4: 74380968-74380968 on Build GRCh38
rs766180343 Chr.4: 74380964-74380964 on Build GRCh38
rs370963182 Chr.4: 74381008-74381008 on Build GRCh38
rs929916549 Chr.4: 74380763-74380763 on Build GRCh38
rs778220054 Chr.4: 74381077-74381077 on Build GRCh38
rs4694187 Chr.4: 74380900-74380900 on Build GRCh38
rs145761841 Chr.4: 74380949-74380949 on Build GRCh38
rs770383310 Chr.4: 74380683-74380683 on Build GRCh38
rs932826547 Chr.4: 74380799-74380799 on Build GRCh38
rs1053141951 Chr.4: 74380877-74380877 on Build GRCh38
rs776578500 Chr.4: 74381104-74381104 on Build GRCh38
rs922502977 Chr.4: 74380758-74380758 on Build GRCh38
rs773255236 Chr.4: 74381092-74381092 on Build GRCh38
rs764438350 Chr.4: 74381113-74381113 on Build GRCh38
rs575785901 Chr.4: 74380784-74380784 on Build GRCh38
rs756509590 Chr.4: 74381046-74381046 on Build GRCh38
rs888246306 Chr.4: 74380795-74380795 on Build GRCh38
rs999701385 Chr.4: 74380960-74380960 on Build GRCh38
rs926053361 Chr.4: 74380822-74380822 on Build GRCh38
rs367705319 Chr.4: 74381070-74381070 on Build GRCh38
rs55702318 Chr.4: 74381011-74381011 on Build GRCh38
rs145916659 Chr.4: 74381045-74381045 on Build GRCh38
rs564583215 Chr.4: 74380963-74380963 on Build GRCh38
rs1024004957 Chr.4: 74380903-74380903 on Build GRCh38
rs572902314 Chr.4: 74380928-74380928 on Build GRCh38
rs952478111 Chr.4: 74380679-74380679 on Build GRCh38
rs529424022 Chr.4: 74381035-74381035 on Build GRCh38
rs375592554 Chr.4: 74381034-74381034 on Build GRCh38
rs1041080933 Chr.4: 74380747-74380747 on Build GRCh38
rs772846778 Chr.4: 74380994-74380994 on Build GRCh38
rs189413524 Chr.4: 74381093-74381093 on Build GRCh38
rs936097240 Chr.4: 74380860-74380860 on Build GRCh38
rs764211023 Chr.4: 74381024-74381024 on Build GRCh38
rs771860305 Chr.4: 74381086-74381086 on Build GRCh38
rs376675128 Chr.4: 74381066-74381066 on Build GRCh38
rs1049908611 Chr.4: 74380817-74380817 on Build GRCh38
rs56794876 Chr.4: 74380943-74380943 on Build GRCh38
rs747725770 Chr.4: 74381080-74381080 on Build GRCh38

View Full Product Details