Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 78551548-78552055 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANXA3
Entrez Gene ID
306
Gene Name
annexin A3
Gene Aliases
ANX3
UniGene
Hs.480042
Gene Chromosome Location
Chr.4: 78551588-78610451 on Build GRCh38
Transcripts
NM_005139.2

SNP Detail

SNP ID Chromosome Location
rs145708718 Chr.4: 78551982-78551982 on Build GRCh38
rs997259852 Chr.4: 78551639-78551639 on Build GRCh38
rs769564088 Chr.4: 78551676-78551676 on Build GRCh38
rs1007439750 Chr.4: 78551663-78551663 on Build GRCh38
rs533578153 Chr.4: 78551934-78551934 on Build GRCh38
rs998550101 Chr.4: 78551864-78551864 on Build GRCh38
rs1036234034 Chr.4: 78551709-78551709 on Build GRCh38
rs919144256 Chr.4: 78551939-78551939 on Build GRCh38
rs34115316 Chr.4: 78551875-78551876 on Build GRCh38
rs1011723674 Chr.4: 78551989-78551989 on Build GRCh38
rs909138296 Chr.4: 78551703-78551703 on Build GRCh38
rs928675857 Chr.4: 78551692-78551692 on Build GRCh38
rs910213160 Chr.4: 78551954-78551954 on Build GRCh38
rs770131202 Chr.4: 78551690-78551690 on Build GRCh38
rs568964515 Chr.4: 78551831-78551831 on Build GRCh38
rs897487589 Chr.4: 78551583-78551583 on Build GRCh38
rs903397615 Chr.4: 78552005-78552005 on Build GRCh38
rs1002852708 Chr.4: 78551860-78551860 on Build GRCh38
rs1040312307 Chr.4: 78551799-78551799 on Build GRCh38
rs1030016320 Chr.4: 78551644-78551644 on Build GRCh38
rs1042010709 Chr.4: 78552002-78552002 on Build GRCh38
rs937209943 Chr.4: 78551697-78551697 on Build GRCh38
rs562323258 Chr.4: 78551631-78551631 on Build GRCh38
rs888837647 Chr.4: 78551782-78551782 on Build GRCh38
rs1036133610 Chr.4: 78552029-78552029 on Build GRCh38
rs971982236 Chr.4: 78551924-78551924 on Build GRCh38
rs548099349 Chr.4: 78551850-78551850 on Build GRCh38
rs1035952428 Chr.4: 78551574-78551574 on Build GRCh38
rs28541479 Chr.4: 78552035-78552035 on Build GRCh38
rs533263557 Chr.4: 78551780-78551780 on Build GRCh38
rs1020339151 Chr.4: 78551684-78551684 on Build GRCh38
rs527709378 Chr.4: 78551653-78551653 on Build GRCh38
rs1005430851 Chr.4: 78551586-78551586 on Build GRCh38
rs933003666 Chr.4: 78551948-78551948 on Build GRCh38
rs982111554 Chr.4: 78551987-78551987 on Build GRCh38
rs991580532 Chr.4: 78551908-78551908 on Build GRCh38
rs957822950 Chr.4: 78551677-78551677 on Build GRCh38
rs183879975 Chr.4: 78551685-78551685 on Build GRCh38
rs759343064 Chr.4: 78551956-78551956 on Build GRCh38
rs1051472186 Chr.4: 78551775-78551775 on Build GRCh38
rs34816574 Chr.4: 78551598-78551599 on Build GRCh38
rs1029321844 Chr.4: 78551874-78551874 on Build GRCh38
rs564631796 Chr.4: 78551750-78551750 on Build GRCh38
rs34317700 Chr.4: 78551857-78551858 on Build GRCh38
rs950374316 Chr.4: 78551736-78551736 on Build GRCh38
rs773054387 Chr.4: 78551764-78551764 on Build GRCh38
rs963225206 Chr.4: 78551922-78551922 on Build GRCh38
rs1390740 Chr.4: 78551902-78551902 on Build GRCh38
rs991691220 Chr.4: 78551698-78551698 on Build GRCh38
rs762307010 Chr.4: 78551619-78551619 on Build GRCh38
rs888256069 Chr.4: 78551816-78551816 on Build GRCh38
rs909077914 Chr.4: 78551911-78551911 on Build GRCh38
rs967385625 Chr.4: 78551859-78551859 on Build GRCh38
rs1049143710 Chr.4: 78551999-78551999 on Build GRCh38
rs767496436 Chr.4: 78551969-78551969 on Build GRCh38
rs941706276 Chr.4: 78551573-78551573 on Build GRCh38
rs1015870722 Chr.4: 78551842-78551843 on Build GRCh38
rs893323464 Chr.4: 78551988-78551988 on Build GRCh38
rs1035783424 Chr.4: 78551879-78551879 on Build GRCh38
rs773779356 Chr.4: 78551708-78551708 on Build GRCh38
rs961303043 Chr.4: 78551633-78551633 on Build GRCh38
rs143738600 Chr.4: 78551791-78551791 on Build GRCh38
rs893304604 Chr.4: 78551800-78551800 on Build GRCh38
rs1002882169 Chr.4: 78552013-78552013 on Build GRCh38
rs990411157 Chr.4: 78551683-78551683 on Build GRCh38
rs888286962 Chr.4: 78551572-78551572 on Build GRCh38
rs937340716 Chr.4: 78551997-78551997 on Build GRCh38
rs187998228 Chr.4: 78551949-78551949 on Build GRCh38
rs988977722 Chr.4: 78551952-78551952 on Build GRCh38
rs531354735 Chr.4: 78551840-78551840 on Build GRCh38

View Full Product Details