Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 78595304-78595824 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANXA3
Entrez Gene ID
306
Gene Name
annexin A3
Gene Aliases
ANX3
UniGene
Hs.480042
Gene Chromosome Location
Chr.4: 78551588-78610451 on Build GRCh38
Transcripts
NM_005139.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3661212 c.536C>T p.A179V Chr.4: 78595433-78595433 on Build GRCh38
3993800 c.498A>G p.E166E Chr.4: 78595395-78595395 on Build GRCh38
3365712 c.522G>A p.L174L Chr.4: 78595419-78595419 on Build GRCh38
3826295 c.505A>G p.K169E Chr.4: 78595402-78595402 on Build GRCh38
6167979 c.514G>A p.E172K Chr.4: 78595411-78595411 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs778744080 Chr.4: 78595792-78595792 on Build GRCh38
rs751358210 Chr.4: 78595383-78595383 on Build GRCh38
rs150363744 Chr.4: 78595341-78595341 on Build GRCh38
rs1017664788 Chr.4: 78595596-78595596 on Build GRCh38
rs1030329518 Chr.4: 78595581-78595581 on Build GRCh38
rs369599553 Chr.4: 78595334-78595334 on Build GRCh38
rs553422236 Chr.4: 78595798-78595798 on Build GRCh38
rs966771583 Chr.4: 78595588-78595588 on Build GRCh38
rs183884121 Chr.4: 78595730-78595730 on Build GRCh38
rs774185206 Chr.4: 78595797-78595797 on Build GRCh38
rs938851579 Chr.4: 78595721-78595721 on Build GRCh38
rs368207621 Chr.4: 78595459-78595459 on Build GRCh38
rs752915875 Chr.4: 78595371-78595371 on Build GRCh38
rs201929153 Chr.4: 78595536-78595537 on Build GRCh38
rs202085239 Chr.4: 78595339-78595339 on Build GRCh38
rs1008661071 Chr.4: 78595490-78595490 on Build GRCh38
rs781021866 Chr.4: 78595388-78595388 on Build GRCh38
rs369969635 Chr.4: 78595546-78595547 on Build GRCh38
rs745335373 Chr.4: 78595391-78595391 on Build GRCh38
rs773692289 Chr.4: 78595438-78595438 on Build GRCh38
rs774639298 Chr.4: 78595393-78595393 on Build GRCh38
rs777527551 Chr.4: 78595378-78595378 on Build GRCh38
rs759721320 Chr.4: 78595593-78595593 on Build GRCh38
rs750332994 Chr.4: 78595480-78595480 on Build GRCh38
rs770648116 Chr.4: 78595796-78595799 on Build GRCh38
rs191355673 Chr.4: 78595727-78595727 on Build GRCh38
rs890296103 Chr.4: 78595464-78595464 on Build GRCh38
rs977340655 Chr.4: 78595702-78595702 on Build GRCh38
rs558889270 Chr.4: 78595452-78595452 on Build GRCh38
rs146641986 Chr.4: 78595424-78595424 on Build GRCh38
rs924839074 Chr.4: 78595326-78595326 on Build GRCh38
rs756189245 Chr.4: 78595481-78595481 on Build GRCh38
rs769201266 Chr.4: 78595392-78595392 on Build GRCh38
rs779699967 Chr.4: 78595765-78595765 on Build GRCh38
rs771431917 Chr.4: 78595446-78595446 on Build GRCh38
rs543741912 Chr.4: 78595350-78595350 on Build GRCh38
rs899157015 Chr.4: 78595375-78595375 on Build GRCh38
rs773780380 Chr.4: 78595357-78595357 on Build GRCh38
rs778159747 Chr.4: 78595421-78595424 on Build GRCh38
rs201313982 Chr.4: 78595355-78595355 on Build GRCh38
rs748860166 Chr.4: 78595770-78595770 on Build GRCh38
rs762248953 Chr.4: 78595544-78595544 on Build GRCh38
rs770933204 Chr.4: 78595362-78595362 on Build GRCh38
rs201205822 Chr.4: 78595432-78595432 on Build GRCh38
rs551045479 Chr.4: 78595524-78595524 on Build GRCh38
rs529601264 Chr.4: 78595543-78595544 on Build GRCh38
rs757023674 Chr.4: 78595476-78595476 on Build GRCh38
rs1038348532 Chr.4: 78595749-78595749 on Build GRCh38
rs778784985 Chr.4: 78595478-78595478 on Build GRCh38
rs748946062 Chr.4: 78595419-78595419 on Build GRCh38
rs374764867 Chr.4: 78595360-78595360 on Build GRCh38
rs112477936 Chr.4: 78595541-78595541 on Build GRCh38
rs765381202 Chr.4: 78595370-78595370 on Build GRCh38
rs199722757 Chr.4: 78595785-78595785 on Build GRCh38
rs927665917 Chr.4: 78595713-78595713 on Build GRCh38
rs958375426 Chr.4: 78595607-78595607 on Build GRCh38
rs989338651 Chr.4: 78595545-78595545 on Build GRCh38
rs374602716 Chr.4: 78595367-78595367 on Build GRCh38
rs757186348 Chr.4: 78595387-78595387 on Build GRCh38
rs760989856 Chr.4: 78595445-78595445 on Build GRCh38
rs548713553 Chr.4: 78595434-78595434 on Build GRCh38
rs536914599 Chr.4: 78595736-78595736 on Build GRCh38
rs751328923 Chr.4: 78595473-78595473 on Build GRCh38
rs571547326 Chr.4: 78595631-78595631 on Build GRCh38
rs749478664 Chr.4: 78595775-78595777 on Build GRCh38
rs1022000654 Chr.4: 78595587-78595587 on Build GRCh38
rs989908048 Chr.4: 78595638-78595638 on Build GRCh38
rs914409361 Chr.4: 78595642-78595642 on Build GRCh38
rs199680808 Chr.4: 78595463-78595463 on Build GRCh38
rs537243794 Chr.4: 78595687-78595687 on Build GRCh38
rs767616187 Chr.4: 78595726-78595726 on Build GRCh38
rs200735427 Chr.4: 78595366-78595366 on Build GRCh38
rs954763539 Chr.4: 78595582-78595582 on Build GRCh38
rs759589738 Chr.4: 78595451-78595451 on Build GRCh38
rs977877308 Chr.4: 78595597-78595597 on Build GRCh38
rs765039716 Chr.4: 78595520-78595520 on Build GRCh38
rs758683446 Chr.4: 78595374-78595374 on Build GRCh38
rs141392132 Chr.4: 78595800-78595800 on Build GRCh38
rs777116557 Chr.4: 78595801-78595801 on Build GRCh38

View Full Product Details