Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 78776225-78776725 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BMP2K
Entrez Gene ID
55589
Gene Name
BMP2 inducible kinase
Gene Aliases
BIKE, HRIHFB2017
UniGene
Hs.146551
Gene Chromosome Location
Chr.4: 78776378-78916365 on Build GRCh38
Transcripts
NM_017593.3 NM_198892.1 XM_005263117.1 XM_011532102.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3696739 c.173C>A p.A58D Chr.4: 78776716-78776716 on Build GRCh38
5423565 c.85delG p.A30fs*49 Chr.4: 78776628-78776628 on Build GRCh38
4767002 c.172G>A p.A58T Chr.4: 78776715-78776715 on Build GRCh38
5423567 c.85delG p.A30fs*49 Chr.4: 78776628-78776628 on Build GRCh38
4767000 c.172G>A p.A58T Chr.4: 78776715-78776715 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs945802301 Chr.4: 78776406-78776406 on Build GRCh38
rs979273044 Chr.4: 78776697-78776697 on Build GRCh38
rs755778782 Chr.4: 78776529-78776529 on Build GRCh38
rs890506252 Chr.4: 78776645-78776645 on Build GRCh38
rs914288160 Chr.4: 78776310-78776310 on Build GRCh38
rs989900305 Chr.4: 78776288-78776288 on Build GRCh38
rs960249287 Chr.4: 78776614-78776614 on Build GRCh38
rs897890837 Chr.4: 78776380-78776380 on Build GRCh38
rs914340410 Chr.4: 78776405-78776405 on Build GRCh38
rs890966712 Chr.4: 78776522-78776522 on Build GRCh38
rs919301565 Chr.4: 78776635-78776635 on Build GRCh38
rs1037912195 Chr.4: 78776628-78776628 on Build GRCh38
rs1025507744 Chr.4: 78776385-78776385 on Build GRCh38
rs775704807 Chr.4: 78776700-78776700 on Build GRCh38
rs1037690608 Chr.4: 78776470-78776470 on Build GRCh38
rs867452798 Chr.4: 78776267-78776267 on Build GRCh38
rs1056060022 Chr.4: 78776428-78776428 on Build GRCh38
rs916061516 Chr.4: 78776620-78776620 on Build GRCh38
rs563478630 Chr.4: 78776393-78776393 on Build GRCh38
rs1006145351 Chr.4: 78776350-78776350 on Build GRCh38
rs542064293 Chr.4: 78776657-78776657 on Build GRCh38
rs989784087 Chr.4: 78776397-78776397 on Build GRCh38
rs941204543 Chr.4: 78776603-78776603 on Build GRCh38
rs765379124 Chr.4: 78776694-78776694 on Build GRCh38
rs987111058 Chr.4: 78776273-78776273 on Build GRCh38
rs986776313 Chr.4: 78776389-78776389 on Build GRCh38
rs1056441698 Chr.4: 78776341-78776341 on Build GRCh38
rs1041882895 Chr.4: 78776322-78776322 on Build GRCh38
rs946208106 Chr.4: 78776313-78776313 on Build GRCh38
rs1018202372 Chr.4: 78776526-78776526 on Build GRCh38
rs776646655 Chr.4: 78776691-78776691 on Build GRCh38
rs894692282 Chr.4: 78776445-78776445 on Build GRCh38
rs772334759 Chr.4: 78776641-78776641 on Build GRCh38
rs910925975 Chr.4: 78776278-78776278 on Build GRCh38
rs1038225845 Chr.4: 78776606-78776606 on Build GRCh38
rs759647822 Chr.4: 78776692-78776692 on Build GRCh38
rs928852772 Chr.4: 78776421-78776421 on Build GRCh38
rs559454547 Chr.4: 78776534-78776534 on Build GRCh38
rs139143500 Chr.4: 78776386-78776386 on Build GRCh38
rs993524614 Chr.4: 78776382-78776382 on Build GRCh38
rs942315705 Chr.4: 78776627-78776627 on Build GRCh38
rs1021329427 Chr.4: 78776538-78776538 on Build GRCh38
rs938975534 Chr.4: 78776338-78776338 on Build GRCh38
rs977237103 Chr.4: 78776570-78776570 on Build GRCh38
rs992044398 Chr.4: 78776615-78776620 on Build GRCh38
rs558513234 Chr.4: 78776258-78776258 on Build GRCh38
rs747463613 Chr.4: 78776678-78776678 on Build GRCh38
rs1016316758 Chr.4: 78776378-78776378 on Build GRCh38
rs1008441799 Chr.4: 78776524-78776524 on Build GRCh38
rs928333815 Chr.4: 78776572-78776572 on Build GRCh38
rs866376617 Chr.4: 78776581-78776581 on Build GRCh38
rs894515228 Chr.4: 78776345-78776345 on Build GRCh38
rs1052280454 Chr.4: 78776510-78776510 on Build GRCh38
rs899773007 Chr.4: 78776626-78776626 on Build GRCh38
rs542863685 Chr.4: 78776519-78776519 on Build GRCh38
rs575578180 Chr.4: 78776296-78776296 on Build GRCh38
rs548621984 Chr.4: 78776487-78776487 on Build GRCh38
rs938845558 Chr.4: 78776422-78776424 on Build GRCh38
rs962527040 Chr.4: 78776253-78776253 on Build GRCh38
rs942342242 Chr.4: 78776285-78776285 on Build GRCh38
rs974616062 Chr.4: 78776670-78776670 on Build GRCh38
rs929260466 Chr.4: 78776637-78776637 on Build GRCh38
rs907345786 Chr.4: 78776337-78776338 on Build GRCh38
rs955344092 Chr.4: 78776268-78776268 on Build GRCh38
rs528176559 Chr.4: 78776542-78776542 on Build GRCh38
rs745419954 Chr.4: 78776501-78776501 on Build GRCh38
rs748455653 Chr.4: 78776559-78776559 on Build GRCh38
rs977560873 Chr.4: 78776420-78776420 on Build GRCh38
rs771505168 Chr.4: 78776685-78776685 on Build GRCh38
rs1011479549 Chr.4: 78776535-78776535 on Build GRCh38

View Full Product Details