Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 78850812-78851313 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BMP2K
Entrez Gene ID
55589
Gene Name
BMP2 inducible kinase
Gene Aliases
BIKE, HRIHFB2017
UniGene
Hs.146551
Gene Chromosome Location
Chr.4: 78776378-78916365 on Build GRCh38
Transcripts
NM_017593.3 NM_198892.1 XM_005263117.1 XM_011532102.1 XM_017008381.1 XM_017008382.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1619122 c.769T>G p.Y257D Chr.4: 78850942-78850942 on Build GRCh38
5709360 c.793_794CC>TT p.P265F Chr.4: 78850966-78850967 on Build GRCh38
1057964 c.772A>G p.K258E Chr.4: 78850945-78850945 on Build GRCh38
5527376 c.794C>T p.P265L Chr.4: 78850967-78850967 on Build GRCh38
5527378 c.794C>T p.P265L Chr.4: 78850967-78850967 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750000743 Chr.4: 78850983-78850983 on Build GRCh38
rs548963773 Chr.4: 78850843-78850843 on Build GRCh38
rs755673845 Chr.4: 78850989-78850989 on Build GRCh38
rs938291716 Chr.4: 78851047-78851047 on Build GRCh38
rs1028227440 Chr.4: 78851239-78851239 on Build GRCh38
rs748568395 Chr.4: 78850873-78850873 on Build GRCh38
rs577299598 Chr.4: 78851199-78851199 on Build GRCh38
rs925821098 Chr.4: 78850902-78850902 on Build GRCh38
rs140182381 Chr.4: 78851027-78851027 on Build GRCh38
rs759735850 Chr.4: 78850916-78850916 on Build GRCh38
rs775112302 Chr.4: 78850927-78850927 on Build GRCh38
rs779786569 Chr.4: 78850997-78850997 on Build GRCh38
rs777696060 Chr.4: 78850884-78850884 on Build GRCh38
rs937312179 Chr.4: 78851215-78851215 on Build GRCh38
rs775795262 Chr.4: 78851052-78851052 on Build GRCh38
rs761910235 Chr.4: 78850973-78850973 on Build GRCh38
rs926898798 Chr.4: 78851045-78851045 on Build GRCh38
rs200371464 Chr.4: 78850898-78850898 on Build GRCh38
rs770080415 Chr.4: 78850920-78850920 on Build GRCh38
rs751532101 Chr.4: 78850958-78850958 on Build GRCh38
rs953884720 Chr.4: 78850887-78850887 on Build GRCh38
rs762239902 Chr.4: 78850908-78850908 on Build GRCh38
rs55782848 Chr.4: 78851036-78851036 on Build GRCh38
rs955764113 Chr.4: 78850910-78850910 on Build GRCh38
rs941228653 Chr.4: 78851290-78851290 on Build GRCh38
rs762908263 Chr.4: 78851003-78851003 on Build GRCh38
rs563279015 Chr.4: 78851216-78851216 on Build GRCh38
rs374919012 Chr.4: 78850896-78850896 on Build GRCh38
rs757867339 Chr.4: 78851189-78851189 on Build GRCh38
rs916879044 Chr.4: 78851002-78851002 on Build GRCh38
rs763142901 Chr.4: 78851065-78851065 on Build GRCh38
rs752934891 Chr.4: 78850998-78850998 on Build GRCh38
rs556653753 Chr.4: 78851233-78851233 on Build GRCh38
rs774329943 Chr.4: 78851078-78851078 on Build GRCh38
rs778344910 Chr.4: 78850882-78850882 on Build GRCh38
rs113647501 Chr.4: 78851035-78851035 on Build GRCh38
rs770904934 Chr.4: 78851162-78851162 on Build GRCh38
rs888620513 Chr.4: 78851153-78851153 on Build GRCh38
rs1045919987 Chr.4: 78851188-78851188 on Build GRCh38
rs768495567 Chr.4: 78851069-78851069 on Build GRCh38
rs780720210 Chr.4: 78851054-78851057 on Build GRCh38
rs758798935 Chr.4: 78851000-78851000 on Build GRCh38
rs763880615 Chr.4: 78850953-78850953 on Build GRCh38
rs373314842 Chr.4: 78851088-78851088 on Build GRCh38
rs754088937 Chr.4: 78850837-78850837 on Build GRCh38
rs767582901 Chr.4: 78851092-78851092 on Build GRCh38
rs1004266815 Chr.4: 78851154-78851154 on Build GRCh38
rs901171516 Chr.4: 78851227-78851227 on Build GRCh38
rs765886021 Chr.4: 78851101-78851101 on Build GRCh38
rs760236990 Chr.4: 78851098-78851098 on Build GRCh38
rs376471857 Chr.4: 78851095-78851095 on Build GRCh38
rs767102767 Chr.4: 78850979-78850979 on Build GRCh38
rs537898784 Chr.4: 78850912-78850912 on Build GRCh38
rs146652536 Chr.4: 78851026-78851026 on Build GRCh38
rs368941274 Chr.4: 78850932-78850932 on Build GRCh38
rs772417958 Chr.4: 78850880-78850880 on Build GRCh38
rs367946700 Chr.4: 78851173-78851173 on Build GRCh38
rs536721142 Chr.4: 78851205-78851205 on Build GRCh38

View Full Product Details