Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 78861199-78861721 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BMP2K
Entrez Gene ID
55589
Gene Name
BMP2 inducible kinase
Gene Aliases
BIKE, HRIHFB2017
UniGene
Hs.146551
Gene Chromosome Location
Chr.4: 78776378-78916365 on Build GRCh38
Transcripts
NM_017593.3 NM_198892.1 XM_005263117.1 XM_011532102.1 XM_017008381.1 XM_017008382.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4854055 c.1009C>G p.L337V Chr.4: 78861410-78861410 on Build GRCh38
6611926 c.992C>T p.S331F Chr.4: 78861393-78861393 on Build GRCh38
7324963 c.1045C>A p.L349I Chr.4: 78861402-78861402 on Build GRCh38
7324964 c.1001C>A p.P334H Chr.4: 78861402-78861402 on Build GRCh38
448182 c.995C>G p.S332C Chr.4: 78861396-78861396 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765374115 Chr.4: 78861358-78861358 on Build GRCh38
rs562375575 Chr.4: 78861372-78861373 on Build GRCh38
rs758603573 Chr.4: 78861372-78861372 on Build GRCh38
rs964827861 Chr.4: 78861594-78861594 on Build GRCh38
rs34280018 Chr.4: 78861685-78861686 on Build GRCh38
rs372950196 Chr.4: 78861518-78861518 on Build GRCh38
rs1022383260 Chr.4: 78861605-78861605 on Build GRCh38
rs185072037 Chr.4: 78861558-78861558 on Build GRCh38
rs978110145 Chr.4: 78861659-78861659 on Build GRCh38
rs756992927 Chr.4: 78861381-78861381 on Build GRCh38
rs550540993 Chr.4: 78861269-78861269 on Build GRCh38
rs566440493 Chr.4: 78861445-78861445 on Build GRCh38
rs759625875 Chr.4: 78861696-78861696 on Build GRCh38
rs531304177 Chr.4: 78861685-78861685 on Build GRCh38
rs751730530 Chr.4: 78861505-78861505 on Build GRCh38
rs781112272 Chr.4: 78861384-78861384 on Build GRCh38
rs1022379138 Chr.4: 78861623-78861623 on Build GRCh38
rs768286874 Chr.4: 78861420-78861420 on Build GRCh38
rs748826365 Chr.4: 78861401-78861401 on Build GRCh38
rs917118290 Chr.4: 78861328-78861328 on Build GRCh38
rs781210946 Chr.4: 78861502-78861503 on Build GRCh38
rs765865666 Chr.4: 78861448-78861448 on Build GRCh38
rs915895834 Chr.4: 78861289-78861289 on Build GRCh38
rs751320448 Chr.4: 78861381-78861381 on Build GRCh38
rs765849778 Chr.4: 78861250-78861254 on Build GRCh38
rs764295707 Chr.4: 78861500-78861500 on Build GRCh38
rs373386639 Chr.4: 78861476-78861476 on Build GRCh38
rs569492468 Chr.4: 78861602-78861602 on Build GRCh38
rs752870932 Chr.4: 78861366-78861366 on Build GRCh38
rs369993657 Chr.4: 78861230-78861230 on Build GRCh38
rs370350603 Chr.4: 78861498-78861498 on Build GRCh38
rs112204213 Chr.4: 78861307-78861307 on Build GRCh38
rs758642777 Chr.4: 78861367-78861367 on Build GRCh38
rs1022498703 Chr.4: 78861229-78861229 on Build GRCh38
rs777171473 Chr.4: 78861443-78861443 on Build GRCh38
rs777865164 Chr.4: 78861372-78861372 on Build GRCh38
rs751314475 Chr.4: 78861379-78861379 on Build GRCh38
rs930544200 Chr.4: 78861396-78861396 on Build GRCh38
rs148493975 Chr.4: 78861677-78861677 on Build GRCh38
rs189946655 Chr.4: 78861597-78861597 on Build GRCh38
rs1017339129 Chr.4: 78861537-78861537 on Build GRCh38
rs761584223 Chr.4: 78861428-78861428 on Build GRCh38
rs776456250 Chr.4: 78861474-78861474 on Build GRCh38
rs924779446 Chr.4: 78861258-78861258 on Build GRCh38
rs779492635 Chr.4: 78861398-78861398 on Build GRCh38
rs766575691 Chr.4: 78861598-78861598 on Build GRCh38
rs747995119 Chr.4: 78861503-78861503 on Build GRCh38
rs745775212 Chr.4: 78861385-78861385 on Build GRCh38
rs554892404 Chr.4: 78861679-78861679 on Build GRCh38
rs904113878 Chr.4: 78861614-78861614 on Build GRCh38
rs1057275487 Chr.4: 78861287-78861287 on Build GRCh38
rs1031434978 Chr.4: 78861650-78861650 on Build GRCh38
rs570030189 Chr.4: 78861415-78861415 on Build GRCh38
rs17857084 Chr.4: 78861425-78861425 on Build GRCh38
rs771191822 Chr.4: 78861440-78861440 on Build GRCh38
rs567439469 Chr.4: 78861480-78861481 on Build GRCh38
rs775647037 Chr.4: 78861451-78861451 on Build GRCh38
rs769773013 Chr.4: 78861397-78861397 on Build GRCh38
rs753792637 Chr.4: 78861421-78861421 on Build GRCh38
rs768402020 Chr.4: 78861302-78861302 on Build GRCh38
rs968962186 Chr.4: 78861644-78861644 on Build GRCh38
rs992229376 Chr.4: 78861313-78861313 on Build GRCh38
rs996290920 Chr.4: 78861589-78861589 on Build GRCh38
rs546739831 Chr.4: 78861330-78861330 on Build GRCh38
rs1038125518 Chr.4: 78861294-78861294 on Build GRCh38
rs116196541 Chr.4: 78861293-78861293 on Build GRCh38

View Full Product Details