Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 78870640-78871159 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BMP2K
Entrez Gene ID
55589
Gene Name
BMP2 inducible kinase
Gene Aliases
BIKE, HRIHFB2017
UniGene
Hs.146551
Gene Chromosome Location
Chr.4: 78776378-78916365 on Build GRCh38
Transcripts
NM_017593.3 NM_198892.1 XM_005263117.1 XM_011532102.1 XM_017008381.1 XM_017008382.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6356747 c.1493A>T p.D498V Chr.4: 78871044-78871044 on Build GRCh38
1594616 c.1459_1461delCAC p.H494delH Chr.4: 78871010-78871012 on Build GRCh38
5989655 c.1431delA p.Q477fs*43 Chr.4: 78870982-78870982 on Build GRCh38
4747544 c.1432_1434delCAG p.Q486delQ Chr.4: 78870983-78870985 on Build GRCh38
6293693 c.1373C>A p.P458H Chr.4: 78870924-78870924 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs534035187 Chr.4: 78870949-78870949 on Build GRCh38
rs766997083 Chr.4: 78870929-78870929 on Build GRCh38
rs1051992970 Chr.4: 78870851-78870851 on Build GRCh38
rs933604970 Chr.4: 78871124-78871124 on Build GRCh38
rs562112527 Chr.4: 78870997-78870997 on Build GRCh38
rs759844671 Chr.4: 78871065-78871066 on Build GRCh38
rs748786542 Chr.4: 78870955-78870956 on Build GRCh38
rs202184856 Chr.4: 78871007-78871015 on Build GRCh38
rs748657636 Chr.4: 78870748-78870748 on Build GRCh38
rs749102729 Chr.4: 78871018-78871018 on Build GRCh38
rs746145828 Chr.4: 78870860-78870860 on Build GRCh38
rs753787696 Chr.4: 78870983-78870991 on Build GRCh38
rs775319075 Chr.4: 78871015-78871015 on Build GRCh38
rs765418240 Chr.4: 78870935-78870955 on Build GRCh38
rs772642517 Chr.4: 78871057-78871057 on Build GRCh38
rs757356061 Chr.4: 78871003-78871003 on Build GRCh38
rs750083207 Chr.4: 78870945-78870945 on Build GRCh38
rs372837691 Chr.4: 78871070-78871070 on Build GRCh38
rs375179282 Chr.4: 78870946-78870946 on Build GRCh38
rs772163101 Chr.4: 78871072-78871072 on Build GRCh38
rs749728323 Chr.4: 78871010-78871018 on Build GRCh38
rs774419973 Chr.4: 78871019-78871019 on Build GRCh38
rs575644040 Chr.4: 78871006-78871006 on Build GRCh38
rs756933262 Chr.4: 78870806-78870806 on Build GRCh38
rs764162326 Chr.4: 78870908-78870908 on Build GRCh38
rs752405558 Chr.4: 78870982-78870983 on Build GRCh38
rs997863601 Chr.4: 78870849-78870849 on Build GRCh38
rs779489845 Chr.4: 78870746-78870746 on Build GRCh38
rs17855588 Chr.4: 78870984-78870984 on Build GRCh38
rs751731669 Chr.4: 78870946-78870947 on Build GRCh38
rs922362030 Chr.4: 78871096-78871096 on Build GRCh38
rs941649302 Chr.4: 78870879-78870879 on Build GRCh38
rs199897907 Chr.4: 78870940-78870940 on Build GRCh38
rs778744082 Chr.4: 78870962-78870964 on Build GRCh38
rs752991478 Chr.4: 78871091-78871091 on Build GRCh38
rs747031407 Chr.4: 78870986-78870986 on Build GRCh38
rs368848734 Chr.4: 78870836-78870836 on Build GRCh38
rs763960588 Chr.4: 78870983-78870988 on Build GRCh38
rs755759610 Chr.4: 78870947-78870947 on Build GRCh38
rs570962845 Chr.4: 78870774-78870774 on Build GRCh38
rs201266597 Chr.4: 78871047-78871047 on Build GRCh38
rs958360371 Chr.4: 78870923-78870923 on Build GRCh38
rs777202582 Chr.4: 78870929-78870955 on Build GRCh38
rs749697430 Chr.4: 78870744-78870744 on Build GRCh38
rs747123204 Chr.4: 78870846-78870846 on Build GRCh38
rs957972800 Chr.4: 78870689-78870692 on Build GRCh38
rs770166159 Chr.4: 78870971-78870976 on Build GRCh38
rs767401459 Chr.4: 78870983-78870985 on Build GRCh38
rs776761309 Chr.4: 78870718-78870718 on Build GRCh38
rs769082087 Chr.4: 78870962-78870991 on Build GRCh38
rs766406376 Chr.4: 78870976-78870977 on Build GRCh38
rs753083008 Chr.4: 78870967-78870967 on Build GRCh38
rs989390331 Chr.4: 78870691-78870691 on Build GRCh38
rs774559559 Chr.4: 78870977-78870988 on Build GRCh38
rs780905484 Chr.4: 78870812-78870812 on Build GRCh38
rs763038959 Chr.4: 78870974-78870982 on Build GRCh38
rs778255977 Chr.4: 78870983-78870983 on Build GRCh38
rs761906204 Chr.4: 78871031-78871031 on Build GRCh38
rs768980559 Chr.4: 78870745-78870745 on Build GRCh38
rs777087113 Chr.4: 78870925-78870926 on Build GRCh38
rs200441916 Chr.4: 78871012-78871012 on Build GRCh38
rs538102569 Chr.4: 78870852-78870852 on Build GRCh38
rs62310843 Chr.4: 78870686-78870686 on Build GRCh38
rs1007749374 Chr.4: 78870685-78870685 on Build GRCh38
rs201291536 Chr.4: 78870785-78870785 on Build GRCh38
rs35071794 Chr.4: 78870929-78870930 on Build GRCh38
rs927600749 Chr.4: 78871100-78871100 on Build GRCh38
rs777781041 Chr.4: 78870843-78870843 on Build GRCh38
rs148040824 Chr.4: 78870830-78870830 on Build GRCh38
rs767150309 Chr.4: 78870950-78870976 on Build GRCh38
rs374800739 Chr.4: 78870918-78870918 on Build GRCh38
rs762936092 Chr.4: 78870905-78870905 on Build GRCh38
rs967000814 Chr.4: 78871059-78871059 on Build GRCh38
rs958157640 Chr.4: 78870715-78870715 on Build GRCh38
rs758861263 Chr.4: 78870977-78870977 on Build GRCh38
rs756003466 Chr.4: 78870950-78870951 on Build GRCh38
rs553571896 Chr.4: 78870955-78870955 on Build GRCh38
rs368667994 Chr.4: 78870780-78870780 on Build GRCh38
rs756779697 Chr.4: 78870958-78870959 on Build GRCh38
rs747479746 Chr.4: 78870931-78870932 on Build GRCh38
rs752677759 Chr.4: 78870789-78870789 on Build GRCh38
rs773315901 Chr.4: 78870761-78870761 on Build GRCh38
rs1007374591 Chr.4: 78870980-78870982 on Build GRCh38
rs745450554 Chr.4: 78870962-78870967 on Build GRCh38
rs772131634 Chr.4: 78870753-78870753 on Build GRCh38
rs776849681 Chr.4: 78870962-78870988 on Build GRCh38
rs778282890 Chr.4: 78871009-78871010 on Build GRCh38
rs374975025 Chr.4: 78870822-78870822 on Build GRCh38
rs770143790 Chr.4: 78871010-78871012 on Build GRCh38
rs757985580 Chr.4: 78871000-78871000 on Build GRCh38
rs3063772 Chr.4: 78870954-78870955 on Build GRCh38
rs748754486 Chr.4: 78870970-78870971 on Build GRCh38
rs762094478 Chr.4: 78870777-78870777 on Build GRCh38
rs570086432 Chr.4: 78870805-78870805 on Build GRCh38
rs781648913 Chr.4: 78871002-78871009 on Build GRCh38
rs745576770 Chr.4: 78870998-78871021 on Build GRCh38
rs747400785 Chr.4: 78870928-78870929 on Build GRCh38
rs576900606 Chr.4: 78870970-78870970 on Build GRCh38
rs982152861 Chr.4: 78870794-78870794 on Build GRCh38
rs1020892285 Chr.4: 78871036-78871036 on Build GRCh38
rs777383965 Chr.4: 78870953-78870955 on Build GRCh38
rs971035100 Chr.4: 78870764-78870764 on Build GRCh38
rs765979105 Chr.4: 78871082-78871082 on Build GRCh38
rs758775044 Chr.4: 78871112-78871112 on Build GRCh38
rs769349891 Chr.4: 78870836-78870847 on Build GRCh38
rs748436557 Chr.4: 78871006-78871007 on Build GRCh38
rs758949057 Chr.4: 78870842-78870842 on Build GRCh38
rs763025663 Chr.4: 78870793-78870793 on Build GRCh38
rs754797676 Chr.4: 78870949-78870950 on Build GRCh38
rs761424407 Chr.4: 78870921-78870921 on Build GRCh38
rs371396282 Chr.4: 78870886-78870886 on Build GRCh38
rs970676013 Chr.4: 78870961-78870961 on Build GRCh38
rs371557810 Chr.4: 78870818-78870818 on Build GRCh38
rs778511354 Chr.4: 78870998-78871012 on Build GRCh38
rs142887199 Chr.4: 78870804-78870804 on Build GRCh38
rs763199077 Chr.4: 78870941-78870961 on Build GRCh38
rs769601417 Chr.4: 78870876-78870876 on Build GRCh38
rs985993396 Chr.4: 78870674-78870674 on Build GRCh38
rs545604264 Chr.4: 78870982-78870982 on Build GRCh38
rs1033519382 Chr.4: 78870976-78870976 on Build GRCh38
rs766562226 Chr.4: 78870947-78870961 on Build GRCh38
rs776924252 Chr.4: 78870854-78870854 on Build GRCh38
rs939753036 Chr.4: 78871029-78871029 on Build GRCh38
rs771163147 Chr.4: 78870775-78870775 on Build GRCh38
rs376418550 Chr.4: 78870929-78870931 on Build GRCh38
rs776047407 Chr.4: 78870929-78870943 on Build GRCh38
rs2114202 Chr.4: 78871009-78871009 on Build GRCh38
rs868411155 Chr.4: 78871120-78871120 on Build GRCh38
rs1007698816 Chr.4: 78870902-78870902 on Build GRCh38
rs762171549 Chr.4: 78870932-78870933 on Build GRCh38
rs773158762 Chr.4: 78870938-78870961 on Build GRCh38
rs752692166 Chr.4: 78870950-78870955 on Build GRCh38
rs757579061 Chr.4: 78870983-78870994 on Build GRCh38
rs755543999 Chr.4: 78870992-78870994 on Build GRCh38
rs937804102 Chr.4: 78871113-78871113 on Build GRCh38
rs770677902 Chr.4: 78870929-78870934 on Build GRCh38
rs972480983 Chr.4: 78871032-78871032 on Build GRCh38

View Full Product Details