Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 83429524-83430053 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HELQ
Entrez Gene ID
113510
Gene Name
helicase, POLQ-like
Gene Aliases
HEL308
UniGene
Hs.480101
Gene Chromosome Location
Chr.4: 83407343-83455883 on Build GRCh38
Transcripts
NM_001297755.1 NM_001297756.1 NM_001297757.1 NM_133636.3 XM_005262711.1 XM_005262713.2 XM_006714076.2 XM_017007679.1 XM_017007680.1 XM_017007682.1 XM_017007683.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6545573 c.2467G>A p.E823K Chr.4: 83429575-83429575 on Build GRCh38
1737611 c.2350G>A p.V784I Chr.4: 83429692-83429692 on Build GRCh38
5043006 c.2388G>A p.W796* Chr.4: 83429654-83429654 on Build GRCh38
4612176 c.2346delT p.F782fs*9 Chr.4: 83429696-83429696 on Build GRCh38
4854197 c.2401G>A p.E801K Chr.4: 83429641-83429641 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1002257382 Chr.4: 83429903-83429903 on Build GRCh38
rs1000302713 Chr.4: 83429703-83429703 on Build GRCh38
rs59255439 Chr.4: 83429717-83429717 on Build GRCh38
rs773260888 Chr.4: 83429702-83429702 on Build GRCh38
rs754627153 Chr.4: 83429721-83429721 on Build GRCh38
rs940224353 Chr.4: 83429880-83429880 on Build GRCh38
rs984585920 Chr.4: 83429839-83429839 on Build GRCh38
rs920272166 Chr.4: 83429689-83429689 on Build GRCh38
rs369861870 Chr.4: 83429745-83429745 on Build GRCh38
rs59175583 Chr.4: 83429763-83429763 on Build GRCh38
rs115519275 Chr.4: 83429934-83429934 on Build GRCh38
rs75238520 Chr.4: 83429586-83429586 on Build GRCh38
rs940318633 Chr.4: 83429685-83429685 on Build GRCh38
rs1049037418 Chr.4: 83429555-83429555 on Build GRCh38
rs749809175 Chr.4: 83429662-83429662 on Build GRCh38
rs751837060 Chr.4: 83429740-83429740 on Build GRCh38
rs771300521 Chr.4: 83429633-83429633 on Build GRCh38
rs775735529 Chr.4: 83429548-83429548 on Build GRCh38
rs755735927 Chr.4: 83429589-83429589 on Build GRCh38
rs777759233 Chr.4: 83429719-83429719 on Build GRCh38
rs764972377 Chr.4: 83429734-83429734 on Build GRCh38
rs751369936 Chr.4: 83429573-83429573 on Build GRCh38
rs763940386 Chr.4: 83429569-83429569 on Build GRCh38
rs375130293 Chr.4: 83429641-83429641 on Build GRCh38
rs771971379 Chr.4: 83429768-83429768 on Build GRCh38
rs892745900 Chr.4: 83429875-83429875 on Build GRCh38
rs183085174 Chr.4: 83429893-83429893 on Build GRCh38
rs781742056 Chr.4: 83429634-83429634 on Build GRCh38
rs747112217 Chr.4: 83429762-83429762 on Build GRCh38
rs756613750 Chr.4: 83429747-83429747 on Build GRCh38
rs769917951 Chr.4: 83429654-83429654 on Build GRCh38
rs779841368 Chr.4: 83429597-83429597 on Build GRCh38
rs767524211 Chr.4: 83429585-83429585 on Build GRCh38
rs4235062 Chr.4: 83430000-83430000 on Build GRCh38
rs572207994 Chr.4: 83429969-83429969 on Build GRCh38
rs372487292 Chr.4: 83429625-83429625 on Build GRCh38
rs768891763 Chr.4: 83429671-83429671 on Build GRCh38
rs199826536 Chr.4: 83429660-83429660 on Build GRCh38
rs757687904 Chr.4: 83429742-83429742 on Build GRCh38
rs754683972 Chr.4: 83429605-83429605 on Build GRCh38
rs927002940 Chr.4: 83429805-83429805 on Build GRCh38
rs577649332 Chr.4: 83429883-83429883 on Build GRCh38
rs1033244772 Chr.4: 83429714-83429714 on Build GRCh38
rs778224985 Chr.4: 83429612-83429612 on Build GRCh38
rs753612557 Chr.4: 83429598-83429598 on Build GRCh38
rs747884204 Chr.4: 83429766-83429766 on Build GRCh38
rs200219888 Chr.4: 83429556-83429556 on Build GRCh38
rs889817351 Chr.4: 83429975-83429975 on Build GRCh38
rs369448607 Chr.4: 83429662-83429662 on Build GRCh38
rs781533843 Chr.4: 83429743-83429743 on Build GRCh38
rs372600577 Chr.4: 83429680-83429680 on Build GRCh38
rs545088806 Chr.4: 83429900-83429900 on Build GRCh38
rs115975794 Chr.4: 83429687-83429687 on Build GRCh38
rs188530231 Chr.4: 83429709-83429709 on Build GRCh38
rs746046642 Chr.4: 83429892-83429892 on Build GRCh38
rs959897193 Chr.4: 83429946-83429946 on Build GRCh38
rs993092812 Chr.4: 83429950-83429950 on Build GRCh38
rs780047237 Chr.4: 83429748-83429748 on Build GRCh38
rs561544969 Chr.4: 83430013-83430013 on Build GRCh38
rs191626794 Chr.4: 83429832-83429832 on Build GRCh38
rs761611311 Chr.4: 83429581-83429581 on Build GRCh38
rs762719876 Chr.4: 83429550-83429550 on Build GRCh38
rs910013788 Chr.4: 83429866-83429866 on Build GRCh38
rs998191467 Chr.4: 83429629-83429629 on Build GRCh38
rs376682646 Chr.4: 83429684-83429684 on Build GRCh38
rs772887651 Chr.4: 83429798-83429798 on Build GRCh38

View Full Product Details