Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 29605480-29605986 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABBR1
Entrez Gene ID
2550
Gene Name
gamma-aminobutyric acid type B receptor subunit 1
Gene Aliases
GABABR1, GABBR1-3, GB1, GPRC3A
UniGene
Hs.167017
Gene Chromosome Location
Chr.6: 29602228-29633135 on Build GRCh38
Transcripts
NM_001319053.1 NM_001470.3 NM_021903.2 NM_021904.3 XM_005248982.2 XM_006715047.3 XM_011514453.2 XM_017010676.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5996847 c.2383T>C p.S795P Chr.6: 29605625-29605625 on Build GRCh38
7289087 c.2072C>T p.A691V Chr.6: 29605585-29605585 on Build GRCh38
1721971 c.2347_2349delCTG p.L783delL Chr.6: 29605659-29605661 on Build GRCh38
1734167 c.2434G>A p.V812M Chr.6: 29605574-29605574 on Build GRCh38
4776271 c.2349G>T p.L783L Chr.6: 29605659-29605659 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs140026528 Chr.6: 29605821-29605821 on Build GRCh38
rs1032859481 Chr.6: 29605706-29605706 on Build GRCh38
rs199842945 Chr.6: 29605687-29605687 on Build GRCh38
rs368201041 Chr.6: 29605659-29605661 on Build GRCh38
rs113441182 Chr.6: 29605753-29605753 on Build GRCh38
rs759821122 Chr.6: 29605566-29605566 on Build GRCh38
rs771402374 Chr.6: 29605736-29605736 on Build GRCh38
rs575051335 Chr.6: 29605844-29605844 on Build GRCh38
rs370516847 Chr.6: 29605742-29605742 on Build GRCh38
rs993403332 Chr.6: 29605818-29605818 on Build GRCh38
rs955239688 Chr.6: 29605855-29605855 on Build GRCh38
rs779610040 Chr.6: 29605638-29605638 on Build GRCh38
rs773453964 Chr.6: 29605728-29605728 on Build GRCh38
rs748804622 Chr.6: 29605646-29605646 on Build GRCh38
rs911871878 Chr.6: 29605937-29605937 on Build GRCh38
rs756005728 Chr.6: 29605609-29605609 on Build GRCh38
rs765607527 Chr.6: 29605569-29605569 on Build GRCh38
rs778575005 Chr.6: 29605664-29605664 on Build GRCh38
rs747828620 Chr.6: 29605673-29605673 on Build GRCh38
rs766416073 Chr.6: 29605543-29605543 on Build GRCh38
rs776752387 Chr.6: 29605738-29605738 on Build GRCh38
rs562761841 Chr.6: 29605889-29605889 on Build GRCh38
rs761100416 Chr.6: 29605542-29605542 on Build GRCh38
rs1023351738 Chr.6: 29605922-29605922 on Build GRCh38
rs540949469 Chr.6: 29605911-29605911 on Build GRCh38
rs768273622 Chr.6: 29605656-29605656 on Build GRCh38
rs989050425 Chr.6: 29605601-29605601 on Build GRCh38
rs751370961 Chr.6: 29605572-29605572 on Build GRCh38
rs987471642 Chr.6: 29605858-29605858 on Build GRCh38
rs1037581767 Chr.6: 29605775-29605775 on Build GRCh38
rs913593639 Chr.6: 29605674-29605674 on Build GRCh38
rs943075439 Chr.6: 29605763-29605763 on Build GRCh38
rs1028234677 Chr.6: 29605852-29605852 on Build GRCh38
rs895712981 Chr.6: 29605908-29605908 on Build GRCh38
rs970169657 Chr.6: 29605924-29605924 on Build GRCh38
rs898993684 Chr.6: 29605806-29605806 on Build GRCh38
rs773728186 Chr.6: 29605541-29605541 on Build GRCh38
rs1014270097 Chr.6: 29605909-29605909 on Build GRCh38
rs757082012 Chr.6: 29605591-29605591 on Build GRCh38
rs956175191 Chr.6: 29605957-29605957 on Build GRCh38
rs750314033 Chr.6: 29605599-29605599 on Build GRCh38
rs761241040 Chr.6: 29605732-29605732 on Build GRCh38
rs751938596 Chr.6: 29605613-29605613 on Build GRCh38
rs753934557 Chr.6: 29605553-29605553 on Build GRCh38
rs781159829 Chr.6: 29605593-29605593 on Build GRCh38
rs1000681872 Chr.6: 29605943-29605943 on Build GRCh38
rs1051840475 Chr.6: 29605695-29605695 on Build GRCh38
rs975380462 Chr.6: 29605762-29605762 on Build GRCh38
rs978959308 Chr.6: 29605514-29605514 on Build GRCh38
rs755363524 Chr.6: 29605945-29605945 on Build GRCh38
rs772364980 Chr.6: 29605727-29605727 on Build GRCh38
rs146059602 Chr.6: 29605540-29605540 on Build GRCh38

View Full Product Details