Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 30892988-30893491 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDR1
Entrez Gene ID
780
Gene Name
discoidin domain receptor tyrosine kinase 1
Gene Aliases
CAK, CD167, DDR, EDDR1, HGK2, MCK10, NEP, NTRK4, PTK3, PTK3A, RTK6, TRKE
UniGene
Hs.631988
Gene Chromosome Location
Chr.6: 30880909-30900156 on Build GRCh38
Transcripts
NM_001202521.1 NM_001202522.1 NM_001202523.1 NM_001297652.1 NM_001297653.1 NM_001297654.1 NM_001954.4 NM_013993.2 NM_013994.2 XM_006715185.2 XM_011514882.2 XM_011514883.1 XM_011514884.1 XM_011514885.1 XM_011514886.1 XM_011514887.1 XM_011514888.2 XM_017011268.1 XM_017011269.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6561325 c.1185C>T p.F395F Chr.6: 30893153-30893153 on Build GRCh38
4448712 c.1196-1G>A p.? Chr.6: 30893271-30893271 on Build GRCh38
3410923 c.1318C>T p.R440W Chr.6: 30893394-30893394 on Build GRCh38
6561326 c.378C>T p.F126F Chr.6: 30893153-30893153 on Build GRCh38
131719 c.1115C>T p.S372F Chr.6: 30893083-30893083 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775472685 Chr.6: 30893243-30893243 on Build GRCh38
rs768724096 Chr.6: 30893361-30893361 on Build GRCh38
rs929560502 Chr.6: 30893312-30893312 on Build GRCh38
rs985248289 Chr.6: 30893202-30893202 on Build GRCh38
rs756528806 Chr.6: 30893459-30893459 on Build GRCh38
rs149673892 Chr.6: 30893143-30893143 on Build GRCh38
rs914400120 Chr.6: 30893196-30893196 on Build GRCh38
rs765319320 Chr.6: 30893122-30893122 on Build GRCh38
rs1264320 Chr.6: 30893223-30893223 on Build GRCh38
rs200321415 Chr.6: 30893435-30893435 on Build GRCh38
rs971383731 Chr.6: 30893069-30893069 on Build GRCh38
rs781474400 Chr.6: 30893321-30893321 on Build GRCh38
rs201323442 Chr.6: 30893297-30893297 on Build GRCh38
rs202224714 Chr.6: 30893192-30893192 on Build GRCh38
rs151111058 Chr.6: 30893128-30893128 on Build GRCh38
rs767043024 Chr.6: 30893120-30893120 on Build GRCh38
rs762082578 Chr.6: 30893394-30893394 on Build GRCh38
rs752199108 Chr.6: 30893299-30893299 on Build GRCh38
rs932418304 Chr.6: 30893201-30893201 on Build GRCh38
rs773782485 Chr.6: 30893116-30893116 on Build GRCh38
rs201573522 Chr.6: 30893079-30893079 on Build GRCh38
rs764108148 Chr.6: 30893251-30893251 on Build GRCh38
rs777380145 Chr.6: 30893309-30893309 on Build GRCh38
rs757303566 Chr.6: 30893159-30893172 on Build GRCh38
rs747068783 Chr.6: 30893143-30893145 on Build GRCh38
rs3213644 Chr.6: 30893448-30893448 on Build GRCh38
rs763712134 Chr.6: 30893444-30893444 on Build GRCh38
rs375127720 Chr.6: 30893091-30893091 on Build GRCh38
rs754991222 Chr.6: 30893072-30893072 on Build GRCh38
rs766177606 Chr.6: 30893275-30893275 on Build GRCh38
rs892169406 Chr.6: 30893028-30893028 on Build GRCh38
rs56123736 Chr.6: 30893336-30893336 on Build GRCh38
rs751122894 Chr.6: 30893351-30893351 on Build GRCh38
rs759356928 Chr.6: 30893411-30893411 on Build GRCh38
rs752806738 Chr.6: 30893173-30893173 on Build GRCh38
rs987949112 Chr.6: 30893468-30893468 on Build GRCh38
rs567947293 Chr.6: 30893138-30893138 on Build GRCh38
rs200571528 Chr.6: 30893089-30893089 on Build GRCh38
rs764268212 Chr.6: 30893177-30893177 on Build GRCh38
rs573519555 Chr.6: 30893215-30893215 on Build GRCh38
rs554564581 Chr.6: 30893062-30893062 on Build GRCh38
rs751107390 Chr.6: 30893445-30893445 on Build GRCh38
rs754889257 Chr.6: 30893191-30893191 on Build GRCh38
rs750234098 Chr.6: 30893209-30893209 on Build GRCh38
rs766209632 Chr.6: 30893324-30893324 on Build GRCh38
rs778348398 Chr.6: 30893198-30893198 on Build GRCh38
rs770328052 Chr.6: 30893239-30893239 on Build GRCh38
rs758005462 Chr.6: 30893308-30893308 on Build GRCh38
rs560023700 Chr.6: 30893463-30893463 on Build GRCh38
rs765312737 Chr.6: 30893075-30893075 on Build GRCh38
rs765244466 Chr.6: 30893171-30893171 on Build GRCh38
rs780212379 Chr.6: 30893461-30893461 on Build GRCh38
rs146690504 Chr.6: 30893144-30893144 on Build GRCh38
rs553526036 Chr.6: 30893064-30893064 on Build GRCh38
rs536980714 Chr.6: 30893189-30893189 on Build GRCh38
rs777070953 Chr.6: 30893231-30893231 on Build GRCh38
rs761953617 Chr.6: 30893273-30893273 on Build GRCh38
rs754011805 Chr.6: 30893057-30893057 on Build GRCh38
rs758586403 Chr.6: 30893078-30893078 on Build GRCh38
rs767725360 Chr.6: 30893406-30893406 on Build GRCh38
rs773655721 Chr.6: 30893046-30893046 on Build GRCh38
rs925145979 Chr.6: 30893252-30893252 on Build GRCh38
rs778021074 Chr.6: 30893087-30893088 on Build GRCh38
rs571877438 Chr.6: 30893183-30893183 on Build GRCh38
rs779781682 Chr.6: 30893184-30893184 on Build GRCh38
rs756723773 Chr.6: 30893040-30893040 on Build GRCh38
rs1016470960 Chr.6: 30893395-30893395 on Build GRCh38
rs763325109 Chr.6: 30893343-30893343 on Build GRCh38
rs764892447 Chr.6: 30893417-30893417 on Build GRCh38
rs376389183 Chr.6: 30893249-30893249 on Build GRCh38
rs368281926 Chr.6: 30893436-30893436 on Build GRCh38
rs371183562 Chr.6: 30893110-30893110 on Build GRCh38
rs575792004 Chr.6: 30893310-30893310 on Build GRCh38
rs980718866 Chr.6: 30893455-30893455 on Build GRCh38
rs769140978 Chr.6: 30893470-30893470 on Build GRCh38
rs145370628 Chr.6: 30893090-30893090 on Build GRCh38
rs376202372 Chr.6: 30893118-30893118 on Build GRCh38
rs372114087 Chr.6: 30893376-30893376 on Build GRCh38
rs1002869977 Chr.6: 30893412-30893412 on Build GRCh38
rs781679716 Chr.6: 30893438-30893438 on Build GRCh38
rs747693382 Chr.6: 30893205-30893205 on Build GRCh38
rs771746527 Chr.6: 30893111-30893111 on Build GRCh38
rs960334684 Chr.6: 30893174-30893174 on Build GRCh38
rs770692392 Chr.6: 30893320-30893320 on Build GRCh38
rs751807233 Chr.6: 30893260-30893260 on Build GRCh38
rs563436031 Chr.6: 30893012-30893012 on Build GRCh38
rs113669350 Chr.6: 30893276-30893276 on Build GRCh38
rs993243737 Chr.6: 30893441-30893441 on Build GRCh38
rs746665157 Chr.6: 30893319-30893319 on Build GRCh38
rs1052211846 Chr.6: 30893287-30893287 on Build GRCh38
rs746211945 Chr.6: 30893323-30893323 on Build GRCh38
rs1044468980 Chr.6: 30893384-30893384 on Build GRCh38
rs371385997 Chr.6: 30893054-30893054 on Build GRCh38
rs760017700 Chr.6: 30893237-30893237 on Build GRCh38
rs149823598 Chr.6: 30893407-30893407 on Build GRCh38
rs780874884 Chr.6: 30893452-30893452 on Build GRCh38
rs752559723 Chr.6: 30893062-30893062 on Build GRCh38
rs778882518 Chr.6: 30893298-30893298 on Build GRCh38
rs752895954 Chr.6: 30893077-30893077 on Build GRCh38
rs540559152 Chr.6: 30893453-30893453 on Build GRCh38
rs746611882 Chr.6: 30893221-30893221 on Build GRCh38
rs953908673 Chr.6: 30893466-30893466 on Build GRCh38
rs1044922377 Chr.6: 30893235-30893235 on Build GRCh38
rs758615103 Chr.6: 30893176-30893176 on Build GRCh38
rs936513519 Chr.6: 30893467-30893467 on Build GRCh38
rs758609602 Chr.6: 30893070-30893072 on Build GRCh38
rs368372549 Chr.6: 30893056-30893056 on Build GRCh38
rs375731129 Chr.6: 30893018-30893018 on Build GRCh38
rs750264052 Chr.6: 30893180-30893180 on Build GRCh38

View Full Product Details