Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 109100248-109100749 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP57L1
Entrez Gene ID
285753
Gene Name
centrosomal protein 57 like 1
Gene Aliases
C6orf182, bA487F23.2, cep57R
UniGene
Hs.632616
Gene Chromosome Location
Chr.6: 109095130-109167696 on Build GRCh38
Transcripts
NM_001083535.1 NM_001271852.1 NM_001271853.1 XM_005266935.4 XM_005266936.4 XM_005266937.4 XM_005266938.4 XM_006715462.3 XM_011535759.2 XM_011535763.2 XM_017010764.1 XM_017010765.1 XM_017010767.1 XM_017010770.1 XM_017010772.1

SNP Detail

SNP ID Chromosome Location
rs1027894306 Chr.6: 109100552-109100552 on Build GRCh38
rs183509131 Chr.6: 109100547-109100547 on Build GRCh38
rs900886788 Chr.6: 109100450-109100450 on Build GRCh38
rs191372013 Chr.6: 109100384-109100384 on Build GRCh38
rs1016191659 Chr.6: 109100535-109100535 on Build GRCh38
rs6913480 Chr.6: 109100397-109100397 on Build GRCh38
rs147628274 Chr.6: 109100484-109100484 on Build GRCh38
rs945209207 Chr.6: 109100706-109100706 on Build GRCh38
rs58571680 Chr.6: 109100692-109100693 on Build GRCh38
rs570942203 Chr.6: 109100504-109100504 on Build GRCh38
rs980079705 Chr.6: 109100571-109100571 on Build GRCh38
rs750877091 Chr.6: 109100452-109100452 on Build GRCh38
rs1035630732 Chr.6: 109100673-109100681 on Build GRCh38
rs913727010 Chr.6: 109100705-109100705 on Build GRCh38
rs755726501 Chr.6: 109100483-109100483 on Build GRCh38
rs757595143 Chr.6: 109100307-109100307 on Build GRCh38
rs558189603 Chr.6: 109100564-109100564 on Build GRCh38
rs892268797 Chr.6: 109100396-109100396 on Build GRCh38
rs1040730104 Chr.6: 109100417-109100417 on Build GRCh38
rs115060876 Chr.6: 109100313-109100313 on Build GRCh38
rs888678888 Chr.6: 109100491-109100491 on Build GRCh38
rs946722435 Chr.6: 109100382-109100384 on Build GRCh38
rs1005779837 Chr.6: 109100503-109100503 on Build GRCh38
rs961535904 Chr.6: 109100536-109100536 on Build GRCh38
rs779537782 Chr.6: 109100489-109100489 on Build GRCh38
rs931822585 Chr.6: 109100435-109100435 on Build GRCh38
rs766967151 Chr.6: 109100329-109100329 on Build GRCh38
rs1024385898 Chr.6: 109100548-109100548 on Build GRCh38
rs980715788 Chr.6: 109100311-109100311 on Build GRCh38
rs539993641 Chr.6: 109100672-109100673 on Build GRCh38
rs939469970 Chr.6: 109100341-109100341 on Build GRCh38
rs559763137 Chr.6: 109100627-109100627 on Build GRCh38
rs571934565 Chr.6: 109100579-109100579 on Build GRCh38
rs1054045415 Chr.6: 109100376-109100376 on Build GRCh38
rs925951091 Chr.6: 109100589-109100589 on Build GRCh38
rs1050711662 Chr.6: 109100438-109100438 on Build GRCh38
rs897467758 Chr.6: 109100696-109100696 on Build GRCh38
rs936540071 Chr.6: 109100340-109100340 on Build GRCh38
rs868456547 Chr.6: 109100693-109100693 on Build GRCh38
rs758946863 Chr.6: 109100480-109100480 on Build GRCh38
rs980960258 Chr.6: 109100629-109100629 on Build GRCh38
rs890211092 Chr.6: 109100451-109100451 on Build GRCh38
rs537649815 Chr.6: 109100590-109100590 on Build GRCh38
rs960912199 Chr.6: 109100689-109100689 on Build GRCh38
rs970073448 Chr.6: 109100561-109100561 on Build GRCh38
rs989378045 Chr.6: 109100690-109100690 on Build GRCh38

View Full Product Details