Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 110610509-110611015 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK19
Entrez Gene ID
23097
Gene Name
cyclin dependent kinase 19
Gene Aliases
CDC2L6, CDK11, bA346C16.3
UniGene
Hs.744895
Gene Chromosome Location
Chr.6: 110609978-110816531 on Build GRCh38
Transcripts
NM_001300960.1 NM_001300963.1 NM_001300964.1 NM_015076.4 XM_005266871.3 XM_011535630.2 XM_011535631.2 XM_017010587.1 XM_017010588.1

SNP Detail

SNP ID Chromosome Location
rs1036681896 Chr.6: 110610596-110610596 on Build GRCh38
rs940968940 Chr.6: 110610590-110610590 on Build GRCh38
rs1019430209 Chr.6: 110610838-110610838 on Build GRCh38
rs905297922 Chr.6: 110610697-110610697 on Build GRCh38
rs1036524623 Chr.6: 110610601-110610601 on Build GRCh38
rs972928836 Chr.6: 110610859-110610859 on Build GRCh38
rs149853215 Chr.6: 110610663-110610663 on Build GRCh38
rs1049787705 Chr.6: 110610646-110610646 on Build GRCh38
rs930903871 Chr.6: 110610637-110610638 on Build GRCh38
rs1543991 Chr.6: 110610829-110610829 on Build GRCh38
rs754651648 Chr.6: 110610870-110610870 on Build GRCh38
rs185286169 Chr.6: 110610921-110610921 on Build GRCh38
rs144872213 Chr.6: 110610771-110610771 on Build GRCh38
rs952594342 Chr.6: 110610894-110610894 on Build GRCh38
rs765725672 Chr.6: 110610560-110610563 on Build GRCh38
rs1045093153 Chr.6: 110610680-110610680 on Build GRCh38
rs1028198918 Chr.6: 110610818-110610818 on Build GRCh38
rs1000755568 Chr.6: 110610807-110610807 on Build GRCh38
rs777783826 Chr.6: 110610588-110610588 on Build GRCh38
rs9386925 Chr.6: 110610554-110610554 on Build GRCh38
rs4269406 Chr.6: 110610575-110610575 on Build GRCh38
rs949525743 Chr.6: 110610962-110610962 on Build GRCh38
rs1056627229 Chr.6: 110610959-110610959 on Build GRCh38
rs1002392841 Chr.6: 110610675-110610676 on Build GRCh38
rs938272454 Chr.6: 110610948-110610948 on Build GRCh38
rs753614069 Chr.6: 110610869-110610869 on Build GRCh38
rs564936439 Chr.6: 110610555-110610555 on Build GRCh38
rs760585879 Chr.6: 110610720-110610720 on Build GRCh38
rs980923383 Chr.6: 110610971-110610971 on Build GRCh38
rs74927541 Chr.6: 110610911-110610911 on Build GRCh38
rs774177801 Chr.6: 110610991-110610991 on Build GRCh38
rs918124471 Chr.6: 110610623-110610623 on Build GRCh38
rs77990092 Chr.6: 110610900-110610900 on Build GRCh38
rs12216071 Chr.6: 110610568-110610568 on Build GRCh38
rs996697403 Chr.6: 110610824-110610824 on Build GRCh38
rs1000848662 Chr.6: 110610742-110610742 on Build GRCh38
rs546428083 Chr.6: 110610837-110610837 on Build GRCh38
rs1054219434 Chr.6: 110610750-110610750 on Build GRCh38
rs13220055 Chr.6: 110610561-110610561 on Build GRCh38
rs892368402 Chr.6: 110610764-110610764 on Build GRCh38
rs939795674 Chr.6: 110610595-110610595 on Build GRCh38
rs965175193 Chr.6: 110610785-110610785 on Build GRCh38
rs972249222 Chr.6: 110610931-110610931 on Build GRCh38
rs202222620 Chr.6: 110610753-110610754 on Build GRCh38
rs919571351 Chr.6: 110610886-110610886 on Build GRCh38
rs968488180 Chr.6: 110610794-110610794 on Build GRCh38
rs926708463 Chr.6: 110610990-110610990 on Build GRCh38
rs868745382 Chr.6: 110610961-110610961 on Build GRCh38
rs552346196 Chr.6: 110610913-110610913 on Build GRCh38
rs537807719 Chr.6: 110610942-110610942 on Build GRCh38
rs961151641 Chr.6: 110610846-110610847 on Build GRCh38
rs1010087436 Chr.6: 110610766-110610766 on Build GRCh38
rs1019548205 Chr.6: 110610780-110610780 on Build GRCh38
rs919625717 Chr.6: 110610603-110610603 on Build GRCh38

View Full Product Details