Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 110612858-110613369 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK19
Entrez Gene ID
23097
Gene Name
cyclin dependent kinase 19
Gene Aliases
CDC2L6, CDK11, bA346C16.3
UniGene
Hs.744895
Gene Chromosome Location
Chr.6: 110609978-110816531 on Build GRCh38
Transcripts
NM_001300960.1 NM_001300963.1 NM_001300964.1 NM_015076.4 XM_005266871.3 XM_011535630.2 XM_011535631.2 XM_017010587.1 XM_017010588.1

SNP Detail

SNP ID Chromosome Location
rs1016911217 Chr.6: 110613256-110613256 on Build GRCh38
rs1054049030 Chr.6: 110613082-110613082 on Build GRCh38
rs183240054 Chr.6: 110613295-110613295 on Build GRCh38
rs755768167 Chr.6: 110613334-110613334 on Build GRCh38
rs909906574 Chr.6: 110613017-110613017 on Build GRCh38
rs151338573 Chr.6: 110613300-110613300 on Build GRCh38
rs767831740 Chr.6: 110613067-110613067 on Build GRCh38
rs971750949 Chr.6: 110613310-110613310 on Build GRCh38
rs568496463 Chr.6: 110613333-110613333 on Build GRCh38
rs767025013 Chr.6: 110612894-110612894 on Build GRCh38
rs984124333 Chr.6: 110612978-110612978 on Build GRCh38
rs990407353 Chr.6: 110613251-110613251 on Build GRCh38
rs755674873 Chr.6: 110613098-110613098 on Build GRCh38
rs201908334 Chr.6: 110613226-110613226 on Build GRCh38
rs569087696 Chr.6: 110613280-110613280 on Build GRCh38
rs561789959 Chr.6: 110612929-110612929 on Build GRCh38
rs914308621 Chr.6: 110613257-110613257 on Build GRCh38
rs768707615 Chr.6: 110613158-110613158 on Build GRCh38
rs888813844 Chr.6: 110613118-110613118 on Build GRCh38
rs192336044 Chr.6: 110613076-110613076 on Build GRCh38
rs5879077 Chr.6: 110613225-110613226 on Build GRCh38
rs917048419 Chr.6: 110613065-110613065 on Build GRCh38
rs967559939 Chr.6: 110613266-110613266 on Build GRCh38
rs1051260526 Chr.6: 110612953-110612953 on Build GRCh38
rs144315117 Chr.6: 110613201-110613201 on Build GRCh38
rs373604414 Chr.6: 110612934-110612934 on Build GRCh38
rs922969109 Chr.6: 110613304-110613304 on Build GRCh38
rs917185335 Chr.6: 110613308-110613308 on Build GRCh38
rs148773159 Chr.6: 110613202-110613202 on Build GRCh38
rs977200698 Chr.6: 110612954-110612954 on Build GRCh38
rs759963278 Chr.6: 110612960-110612960 on Build GRCh38
rs935775054 Chr.6: 110613079-110613079 on Build GRCh38
rs971287156 Chr.6: 110613196-110613196 on Build GRCh38
rs932974378 Chr.6: 110613335-110613335 on Build GRCh38
rs991413296 Chr.6: 110613057-110613057 on Build GRCh38
rs1026261047 Chr.6: 110613223-110613223 on Build GRCh38
rs994118295 Chr.6: 110613151-110613151 on Build GRCh38
rs774790102 Chr.6: 110612927-110612927 on Build GRCh38
rs552497419 Chr.6: 110613215-110613215 on Build GRCh38
rs937234249 Chr.6: 110613045-110613045 on Build GRCh38
rs1005678027 Chr.6: 110613244-110613244 on Build GRCh38
rs898600239 Chr.6: 110613088-110613088 on Build GRCh38
rs879598212 Chr.6: 110612959-110612959 on Build GRCh38
rs546939830 Chr.6: 110613091-110613091 on Build GRCh38
rs752393765 Chr.6: 110613077-110613077 on Build GRCh38

View Full Product Details