Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 111674357-111674857 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FYN
Entrez Gene ID
2534
Gene Name
FYN proto-oncogene, Src family tyrosine kinase
Gene Aliases
SLK, SYN, p59-FYN
UniGene
Hs.390567
Gene Chromosome Location
Chr.6: 111660332-111873452 on Build GRCh38
Transcripts
NM_002037.5 NM_153047.3 NM_153048.3 XM_005266890.3 XM_005266892.3 XM_017010650.1 XM_017010651.1 XM_017010652.1 XM_017010653.1 XM_017010654.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3857635 c.1315C>A p.L439M Chr.6: 111674589-111674589 on Build GRCh38
1697765 c.1298C>T p.T433M Chr.6: 111674606-111674606 on Build GRCh38
3857637 c.1283T>G p.F428C Chr.6: 111674621-111674621 on Build GRCh38
4685940 c.1335C>T p.D445D Chr.6: 111674560-111674560 on Build GRCh38
3619585 c.1343C>T p.S448F Chr.6: 111674552-111674552 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374678602 Chr.6: 111674815-111674815 on Build GRCh38
rs575332330 Chr.6: 111674662-111674662 on Build GRCh38
rs368455702 Chr.6: 111674584-111674584 on Build GRCh38
rs869240076 Chr.6: 111674718-111674718 on Build GRCh38
rs577639834 Chr.6: 111674816-111674816 on Build GRCh38
rs776900760 Chr.6: 111674521-111674521 on Build GRCh38
rs1028715991 Chr.6: 111674690-111674690 on Build GRCh38
rs1000887355 Chr.6: 111674427-111674427 on Build GRCh38
rs982437538 Chr.6: 111674792-111674792 on Build GRCh38
rs758528682 Chr.6: 111674626-111674626 on Build GRCh38
rs3730353 Chr.6: 111674463-111674463 on Build GRCh38
rs1008879174 Chr.6: 111674492-111674492 on Build GRCh38
rs1801121 Chr.6: 111674571-111674571 on Build GRCh38
rs943887188 Chr.6: 111674391-111674391 on Build GRCh38
rs767344335 Chr.6: 111674578-111674578 on Build GRCh38
rs780676443 Chr.6: 111674529-111674529 on Build GRCh38
rs1045182574 Chr.6: 111674381-111674381 on Build GRCh38
rs766184115 Chr.6: 111674559-111674559 on Build GRCh38
rs951152746 Chr.6: 111674781-111674781 on Build GRCh38
rs747793778 Chr.6: 111674523-111674523 on Build GRCh38
rs758911070 Chr.6: 111674478-111674478 on Build GRCh38
rs975241436 Chr.6: 111674409-111674409 on Build GRCh38
rs181693633 Chr.6: 111674383-111674383 on Build GRCh38
rs769043283 Chr.6: 111674486-111674486 on Build GRCh38
rs1801109 Chr.6: 111674591-111674591 on Build GRCh38
rs780643960 Chr.6: 111674480-111674480 on Build GRCh38
rs962026371 Chr.6: 111674782-111674782 on Build GRCh38
rs755605763 Chr.6: 111674731-111674731 on Build GRCh38
rs975252841 Chr.6: 111674825-111674825 on Build GRCh38
rs757623530 Chr.6: 111674472-111674472 on Build GRCh38
rs200659869 Chr.6: 111674471-111674471 on Build GRCh38
rs186311726 Chr.6: 111674716-111674716 on Build GRCh38
rs755502848 Chr.6: 111674677-111674677 on Build GRCh38
rs780046906 Chr.6: 111674640-111674640 on Build GRCh38
rs576515269 Chr.6: 111674777-111674777 on Build GRCh38
rs896061537 Chr.6: 111674676-111674676 on Build GRCh38
rs371443548 Chr.6: 111674678-111674678 on Build GRCh38
rs747288114 Chr.6: 111674485-111674485 on Build GRCh38
rs1019582682 Chr.6: 111674814-111674814 on Build GRCh38
rs921361484 Chr.6: 111674415-111674415 on Build GRCh38
rs765319412 Chr.6: 111674722-111674722 on Build GRCh38
rs996928563 Chr.6: 111674590-111674590 on Build GRCh38
rs370092420 Chr.6: 111674666-111674666 on Build GRCh38
rs752601385 Chr.6: 111674583-111674583 on Build GRCh38
rs142448026 Chr.6: 111674527-111674527 on Build GRCh38
rs762451123 Chr.6: 111674536-111674536 on Build GRCh38
rs367543106 Chr.6: 111674557-111674557 on Build GRCh38
rs773268663 Chr.6: 111674396-111674396 on Build GRCh38
rs745961377 Chr.6: 111674477-111674477 on Build GRCh38
rs750493333 Chr.6: 111674605-111674605 on Build GRCh38
rs765563112 Chr.6: 111674599-111674599 on Build GRCh38
rs759436970 Chr.6: 111674566-111674566 on Build GRCh38
rs1023048219 Chr.6: 111674706-111674706 on Build GRCh38
rs995643269 Chr.6: 111674747-111674747 on Build GRCh38
rs555142306 Chr.6: 111674681-111674681 on Build GRCh38
rs764169408 Chr.6: 111674465-111674465 on Build GRCh38
rs779126245 Chr.6: 111674476-111674476 on Build GRCh38
rs536389321 Chr.6: 111674641-111674641 on Build GRCh38
rs747740382 Chr.6: 111674723-111674723 on Build GRCh38
rs774364411 Chr.6: 111674560-111674560 on Build GRCh38
rs751243713 Chr.6: 111674462-111674462 on Build GRCh38
rs1043073752 Chr.6: 111674453-111674453 on Build GRCh38
rs951768698 Chr.6: 111674691-111674691 on Build GRCh38
rs558770725 Chr.6: 111674783-111674783 on Build GRCh38

View Full Product Details