Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 2731423-2731930 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMZ1
Entrez Gene ID
155185
Gene Name
archaelysin family metallopeptidase 1
Gene Aliases
-
UniGene
Hs.42221
Gene Chromosome Location
Chr.7: 2679522-2767486 on Build GRCh38
Transcripts
NM_001321766.1 XM_011515151.2 XM_017011774.1 NM_001282440.1 NM_001282441.1 NM_001293092.1 NM_007353.2 XM_011515288.2 XM_017012008.1

Gene Symbol
GNA12
Entrez Gene ID
2768
Gene Name
G protein subunit alpha 12
Gene Aliases
NNX3, RMP, gep
UniGene
Hs.487341
Gene Chromosome Location
Chr.7: 2728105-2870656 on Build GRCh38
Transcripts
NM_001321766.1 XM_011515151.2 XM_017011774.1 NM_001282440.1 NM_001282441.1 NM_001293092.1 NM_007353.2 XM_011515288.2 XM_017012008.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
74769 c.704G>A p.R235H Chr.7: 2731623-2731623 on Build GRCh38
3663127 c.720G>T p.Q240H Chr.7: 2731607-2731607 on Build GRCh38
4866716 c.321C>T p.S107S Chr.7: 2731562-2731562 on Build GRCh38
3027515 c.389A>T p.E130V Chr.7: 2731494-2731494 on Build GRCh38
3637769 c.718C>T p.Q240* Chr.7: 2731609-2731609 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs918756975 Chr.7: 2731706-2731706 on Build GRCh38
rs932431919 Chr.7: 2731489-2731489 on Build GRCh38
rs766643469 Chr.7: 2731725-2731725 on Build GRCh38
rs980878134 Chr.7: 2731633-2731633 on Build GRCh38
rs200282933 Chr.7: 2731495-2731495 on Build GRCh38
rs779040599 Chr.7: 2731515-2731515 on Build GRCh38
rs952696779 Chr.7: 2731703-2731703 on Build GRCh38
rs771969963 Chr.7: 2731462-2731462 on Build GRCh38
rs906720953 Chr.7: 2731887-2731887 on Build GRCh38
rs199650069 Chr.7: 2731487-2731487 on Build GRCh38
rs748851227 Chr.7: 2731791-2731791 on Build GRCh38
rs774944056 Chr.7: 2731568-2731568 on Build GRCh38
rs1052830518 Chr.7: 2731856-2731856 on Build GRCh38
rs758966311 Chr.7: 2731483-2731483 on Build GRCh38
rs1049481827 Chr.7: 2731824-2731824 on Build GRCh38
rs748012133 Chr.7: 2731454-2731454 on Build GRCh38
rs191664263 Chr.7: 2731467-2731467 on Build GRCh38
rs375880168 Chr.7: 2731663-2731663 on Build GRCh38
rs748726279 Chr.7: 2731676-2731676 on Build GRCh38
rs755763128 Chr.7: 2731787-2731787 on Build GRCh38
rs766596673 Chr.7: 2731601-2731601 on Build GRCh38
rs370690250 Chr.7: 2731679-2731679 on Build GRCh38
rs994423247 Chr.7: 2731867-2731867 on Build GRCh38
rs535681947 Chr.7: 2731576-2731576 on Build GRCh38
rs961146959 Chr.7: 2731882-2731882 on Build GRCh38
rs774700519 Chr.7: 2731582-2731582 on Build GRCh38
rs757182218 Chr.7: 2731764-2731764 on Build GRCh38
rs748067229 Chr.7: 2731519-2731519 on Build GRCh38
rs750171093 Chr.7: 2731786-2731786 on Build GRCh38
rs879159666 Chr.7: 2731784-2731784 on Build GRCh38
rs760828167 Chr.7: 2731723-2731723 on Build GRCh38
rs762286637 Chr.7: 2731697-2731697 on Build GRCh38
rs999964514 Chr.7: 2731444-2731444 on Build GRCh38
rs772291731 Chr.7: 2731712-2731712 on Build GRCh38
rs773037986 Chr.7: 2731463-2731463 on Build GRCh38
rs761117102 Chr.7: 2731798-2731798 on Build GRCh38
rs751429360 Chr.7: 2731640-2731640 on Build GRCh38
rs751868693 Chr.7: 2731491-2731491 on Build GRCh38
rs9655213 Chr.7: 2731761-2731761 on Build GRCh38
rs1016959261 Chr.7: 2731848-2731848 on Build GRCh38
rs769392159 Chr.7: 2731654-2731654 on Build GRCh38
rs750586131 Chr.7: 2731598-2731598 on Build GRCh38
rs756132903 Chr.7: 2731512-2731512 on Build GRCh38
rs780940374 Chr.7: 2731645-2731645 on Build GRCh38
rs889897945 Chr.7: 2731840-2731840 on Build GRCh38
rs371474667 Chr.7: 2731616-2731616 on Build GRCh38
rs149751425 Chr.7: 2731499-2731499 on Build GRCh38
rs41305918 Chr.7: 2731795-2731795 on Build GRCh38
rs1051600150 Chr.7: 2731839-2731839 on Build GRCh38
rs11772508 Chr.7: 2731870-2731870 on Build GRCh38
rs745435606 Chr.7: 2731564-2731564 on Build GRCh38
rs746042429 Chr.7: 2731524-2731524 on Build GRCh38
rs201475988 Chr.7: 2731678-2731678 on Build GRCh38
rs1033732554 Chr.7: 2731688-2731688 on Build GRCh38
rs891449530 Chr.7: 2731866-2731866 on Build GRCh38
rs779512222 Chr.7: 2731794-2731794 on Build GRCh38
rs779674921 Chr.7: 2731789-2731789 on Build GRCh38
rs201962977 Chr.7: 2731549-2731549 on Build GRCh38
rs372227846 Chr.7: 2731783-2731783 on Build GRCh38
rs764109887 Chr.7: 2731818-2731818 on Build GRCh38
rs773585278 Chr.7: 2731721-2731721 on Build GRCh38
rs767565658 Chr.7: 2731510-2731510 on Build GRCh38
rs754038385 Chr.7: 2731738-2731738 on Build GRCh38
rs904215612 Chr.7: 2731521-2731521 on Build GRCh38
rs776211969 Chr.7: 2731482-2731482 on Build GRCh38
rs908533586 Chr.7: 2731762-2731762 on Build GRCh38
rs777850035 Chr.7: 2731632-2731632 on Build GRCh38
rs540650945 Chr.7: 2731472-2731472 on Build GRCh38
rs745542867 Chr.7: 2731649-2731649 on Build GRCh38
rs1046666701 Chr.7: 2731880-2731880 on Build GRCh38
rs764178346 Chr.7: 2731750-2731750 on Build GRCh38
rs199802606 Chr.7: 2731459-2731459 on Build GRCh38
rs925672791 Chr.7: 2731831-2731831 on Build GRCh38
rs759677261 Chr.7: 2731742-2731742 on Build GRCh38
rs776135196 Chr.7: 2731562-2731562 on Build GRCh38
rs566672345 Chr.7: 2731574-2731574 on Build GRCh38
rs762077179 Chr.7: 2731585-2731585 on Build GRCh38
rs932463218 Chr.7: 2731807-2731807 on Build GRCh38
rs756257195 Chr.7: 2731600-2731600 on Build GRCh38
rs756982785 Chr.7: 2731643-2731643 on Build GRCh38
rs570479193 Chr.7: 2731490-2731490 on Build GRCh38
rs954175741 Chr.7: 2731531-2731531 on Build GRCh38
rs767837122 Chr.7: 2731590-2731590 on Build GRCh38
rs770685700 Chr.7: 2731551-2731551 on Build GRCh38
rs45606633 Chr.7: 2731603-2731603 on Build GRCh38
rs138638605 Chr.7: 2731631-2731631 on Build GRCh38
rs768130678 Chr.7: 2731792-2731792 on Build GRCh38
rs574070992 Chr.7: 2731842-2731842 on Build GRCh38
rs769160245 Chr.7: 2731565-2731565 on Build GRCh38
rs999835892 Chr.7: 2731555-2731555 on Build GRCh38
rs992363540 Chr.7: 2731896-2731896 on Build GRCh38
rs150519840 Chr.7: 2731486-2731486 on Build GRCh38
rs778710393 Chr.7: 2731448-2731448 on Build GRCh38
rs758369709 Chr.7: 2731530-2731530 on Build GRCh38

View Full Product Details