Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 5527917-5528424 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTB
Entrez Gene ID
60
Gene Name
actin beta
Gene Aliases
BRWS1, PS1TP5BP1
UniGene
Hs.520640
Gene Chromosome Location
Chr.7: 5527148-5530601 on Build GRCh38
Transcripts
NM_001101.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1755317 c.669C>A p.F223L Chr.7: 5528414-5528414 on Build GRCh38
4504263 c.660C>T p.A220A Chr.7: 5528423-5528423 on Build GRCh38
6178037 c.719A>G p.Y240C Chr.7: 5528364-5528364 on Build GRCh38
1741423 c.757G>A p.E253K Chr.7: 5528326-5528326 on Build GRCh38
1741422 c.775G>A p.E259K Chr.7: 5528308-5528308 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370842886 Chr.7: 5528175-5528175 on Build GRCh38
rs1024438680 Chr.7: 5528303-5528303 on Build GRCh38
rs375076505 Chr.7: 5528232-5528232 on Build GRCh38
rs751030505 Chr.7: 5527976-5527976 on Build GRCh38
rs748432427 Chr.7: 5528263-5528263 on Build GRCh38
rs749038548 Chr.7: 5528262-5528262 on Build GRCh38
rs769182426 Chr.7: 5528100-5528100 on Build GRCh38
rs759978214 Chr.7: 5527966-5527966 on Build GRCh38
rs774758801 Chr.7: 5528106-5528106 on Build GRCh38
rs372828516 Chr.7: 5528237-5528237 on Build GRCh38
rs774251900 Chr.7: 5528327-5528327 on Build GRCh38
rs760228668 Chr.7: 5528205-5528205 on Build GRCh38
rs765988813 Chr.7: 5528206-5528206 on Build GRCh38
rs549072877 Chr.7: 5527982-5527982 on Build GRCh38
rs878948364 Chr.7: 5528003-5528003 on Build GRCh38
rs780418020 Chr.7: 5528233-5528233 on Build GRCh38
rs374997048 Chr.7: 5527973-5527973 on Build GRCh38
rs766044090 Chr.7: 5528124-5528124 on Build GRCh38
rs200270797 Chr.7: 5528267-5528267 on Build GRCh38
rs199962715 Chr.7: 5528235-5528235 on Build GRCh38
rs878971183 Chr.7: 5528001-5528001 on Build GRCh38
rs11546937 Chr.7: 5528107-5528107 on Build GRCh38
rs754810018 Chr.7: 5528154-5528154 on Build GRCh38
rs745402722 Chr.7: 5528318-5528318 on Build GRCh38
rs747085370 Chr.7: 5527957-5527957 on Build GRCh38
rs11546896 Chr.7: 5528157-5528157 on Build GRCh38
rs570862769 Chr.7: 5528381-5528381 on Build GRCh38
rs201400527 Chr.7: 5528082-5528082 on Build GRCh38
rs879102499 Chr.7: 5528279-5528279 on Build GRCh38
rs538560302 Chr.7: 5527943-5527943 on Build GRCh38
rs374794805 Chr.7: 5528243-5528243 on Build GRCh38
rs371153506 Chr.7: 5528097-5528097 on Build GRCh38
rs754862802 Chr.7: 5528004-5528004 on Build GRCh38
rs372551192 Chr.7: 5527989-5527989 on Build GRCh38
rs201113744 Chr.7: 5528378-5528378 on Build GRCh38
rs375036292 Chr.7: 5528207-5528207 on Build GRCh38
rs376569032 Chr.7: 5527940-5527940 on Build GRCh38
rs539831938 Chr.7: 5528063-5528063 on Build GRCh38
rs13447406 Chr.7: 5528088-5528088 on Build GRCh38
rs148969043 Chr.7: 5528345-5528345 on Build GRCh38
rs766880763 Chr.7: 5528112-5528112 on Build GRCh38
rs758868066 Chr.7: 5528269-5528269 on Build GRCh38
rs760723174 Chr.7: 5527956-5527956 on Build GRCh38
rs765697376 Chr.7: 5527997-5527997 on Build GRCh38
rs779757174 Chr.7: 5527992-5527992 on Build GRCh38
rs774299670 Chr.7: 5528357-5528357 on Build GRCh38
rs552269338 Chr.7: 5528351-5528351 on Build GRCh38
rs760283548 Chr.7: 5528118-5528118 on Build GRCh38
rs745973745 Chr.7: 5528178-5528178 on Build GRCh38
rs556846750 Chr.7: 5528399-5528399 on Build GRCh38
rs369091281 Chr.7: 5527974-5527974 on Build GRCh38
rs34792388 Chr.7: 5528058-5528058 on Build GRCh38
rs762851719 Chr.7: 5528256-5528256 on Build GRCh38
rs372898189 Chr.7: 5528216-5528216 on Build GRCh38
rs145793213 Chr.7: 5528363-5528363 on Build GRCh38
rs765395832 Chr.7: 5528233-5528234 on Build GRCh38
rs745673720 Chr.7: 5528384-5528384 on Build GRCh38
rs765751796 Chr.7: 5527967-5527967 on Build GRCh38
rs764970402 Chr.7: 5528252-5528252 on Build GRCh38
rs767166581 Chr.7: 5527988-5527988 on Build GRCh38
rs191425263 Chr.7: 5528211-5528211 on Build GRCh38
rs764886447 Chr.7: 5528221-5528221 on Build GRCh38
rs377004519 Chr.7: 5528290-5528290 on Build GRCh38
rs778767698 Chr.7: 5528019-5528019 on Build GRCh38
rs372973178 Chr.7: 5528261-5528261 on Build GRCh38
rs879226771 Chr.7: 5528187-5528187 on Build GRCh38
rs150105166 Chr.7: 5528130-5528130 on Build GRCh38
rs35311107 Chr.7: 5528070-5528070 on Build GRCh38
rs777412747 Chr.7: 5528248-5528248 on Build GRCh38
rs756248568 Chr.7: 5528259-5528259 on Build GRCh38
rs770490362 Chr.7: 5528247-5528247 on Build GRCh38
rs766398197 Chr.7: 5527991-5527992 on Build GRCh38
rs11546915 Chr.7: 5528109-5528109 on Build GRCh38
rs369552329 Chr.7: 5527961-5527961 on Build GRCh38
rs187270726 Chr.7: 5527987-5527987 on Build GRCh38
rs13447408 Chr.7: 5528037-5528037 on Build GRCh38
rs759039997 Chr.7: 5528250-5528250 on Build GRCh38
rs780471227 Chr.7: 5528192-5528192 on Build GRCh38
rs145818896 Chr.7: 5528321-5528321 on Build GRCh38
rs775096086 Chr.7: 5528254-5528254 on Build GRCh38
rs377390140 Chr.7: 5528166-5528166 on Build GRCh38
rs770460552 Chr.7: 5528285-5528285 on Build GRCh38
rs771436794 Chr.7: 5528288-5528288 on Build GRCh38
rs759740649 Chr.7: 5528232-5528236 on Build GRCh38
rs375065288 Chr.7: 5528193-5528193 on Build GRCh38
rs753983599 Chr.7: 5528260-5528260 on Build GRCh38
rs770089307 Chr.7: 5528184-5528184 on Build GRCh38
rs757404770 Chr.7: 5527991-5527991 on Build GRCh38
rs749113962 Chr.7: 5527995-5527995 on Build GRCh38
rs372006500 Chr.7: 5528231-5528231 on Build GRCh38
rs879203462 Chr.7: 5528278-5528278 on Build GRCh38
rs368944901 Chr.7: 5528257-5528257 on Build GRCh38
rs768587113 Chr.7: 5527942-5527942 on Build GRCh38
rs749940556 Chr.7: 5528115-5528115 on Build GRCh38
rs878878454 Chr.7: 5528277-5528277 on Build GRCh38
rs763430371 Chr.7: 5528098-5528098 on Build GRCh38
rs753950600 Chr.7: 5528111-5528111 on Build GRCh38
rs11546925 Chr.7: 5528326-5528326 on Build GRCh38
rs746616854 Chr.7: 5528238-5528239 on Build GRCh38
rs370628413 Chr.7: 5528203-5528203 on Build GRCh38
rs879165552 Chr.7: 5527999-5527999 on Build GRCh38
rs369349916 Chr.7: 5528236-5528236 on Build GRCh38
rs778241923 Chr.7: 5528275-5528275 on Build GRCh38
rs1057517888 Chr.7: 5528120-5528120 on Build GRCh38
rs11546939 Chr.7: 5528046-5528046 on Build GRCh38
rs852424 Chr.7: 5527965-5527965 on Build GRCh38
rs149428488 Chr.7: 5528223-5528223 on Build GRCh38
rs138499594 Chr.7: 5528396-5528396 on Build GRCh38
rs11546899 Chr.7: 5528355-5528355 on Build GRCh38
rs879092158 Chr.7: 5528000-5528000 on Build GRCh38
rs749580588 Chr.7: 5528234-5528234 on Build GRCh38
rs752537146 Chr.7: 5528222-5528222 on Build GRCh38
rs367990633 Chr.7: 5528194-5528194 on Build GRCh38
rs780741199 Chr.7: 5528191-5528191 on Build GRCh38
rs886726090 Chr.7: 5528271-5528271 on Build GRCh38
rs11546930 Chr.7: 5528313-5528313 on Build GRCh38
rs747456624 Chr.7: 5528229-5528229 on Build GRCh38
rs373362107 Chr.7: 5528163-5528163 on Build GRCh38
rs763891719 Chr.7: 5527977-5527977 on Build GRCh38
rs13447407 Chr.7: 5528085-5528085 on Build GRCh38

View Full Product Details