Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 6409436-6409944 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DAGLB
Entrez Gene ID
221955
Gene Name
diacylglycerol lipase beta
Gene Aliases
DAGLBETA, KCCR13L
UniGene
Hs.487498
Gene Chromosome Location
Chr.7: 6409116-6448012 on Build GRCh38
Transcripts
NM_001142936.1 NM_139179.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3882033 c.2012T>C p.V671A Chr.7: 6409844-6409844 on Build GRCh38
5725904 c.1938G>A p.R646R Chr.7: 6409918-6409918 on Build GRCh38
3882034 c.2011G>A p.V671M Chr.7: 6409845-6409845 on Build GRCh38
6876466 c.2007G>A p.V669V Chr.7: 6409849-6409849 on Build GRCh38
3982246 c.1991A>G p.Q664R Chr.7: 6409865-6409865 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774711011 Chr.7: 6409852-6409852 on Build GRCh38
rs534855020 Chr.7: 6409867-6409867 on Build GRCh38
rs943853600 Chr.7: 6409471-6409472 on Build GRCh38
rs35074640 Chr.7: 6409470-6409471 on Build GRCh38
rs769057175 Chr.7: 6409791-6409791 on Build GRCh38
rs1035826116 Chr.7: 6409553-6409553 on Build GRCh38
rs1038438904 Chr.7: 6409613-6409613 on Build GRCh38
rs778915640 Chr.7: 6409811-6409811 on Build GRCh38
rs371655886 Chr.7: 6409549-6409549 on Build GRCh38
rs758421152 Chr.7: 6409821-6409821 on Build GRCh38
rs765010300 Chr.7: 6409915-6409915 on Build GRCh38
rs182599475 Chr.7: 6409747-6409747 on Build GRCh38
rs145993691 Chr.7: 6409890-6409890 on Build GRCh38
rs757156328 Chr.7: 6409921-6409921 on Build GRCh38
rs760064650 Chr.7: 6409805-6409805 on Build GRCh38
rs1003723084 Chr.7: 6409542-6409542 on Build GRCh38
rs1015285692 Chr.7: 6409515-6409515 on Build GRCh38
rs376036204 Chr.7: 6409872-6409872 on Build GRCh38
rs836560 Chr.7: 6409598-6409598 on Build GRCh38
rs888104439 Chr.7: 6409493-6409493 on Build GRCh38
rs777519827 Chr.7: 6409829-6409829 on Build GRCh38
rs73341299 Chr.7: 6409568-6409568 on Build GRCh38
rs765885126 Chr.7: 6409806-6409806 on Build GRCh38
rs113378075 Chr.7: 6409569-6409569 on Build GRCh38
rs540007522 Chr.7: 6409795-6409795 on Build GRCh38
rs201251636 Chr.7: 6409468-6409469 on Build GRCh38
rs371750309 Chr.7: 6409871-6409871 on Build GRCh38
rs770074373 Chr.7: 6409773-6409773 on Build GRCh38
rs771398959 Chr.7: 6409906-6409906 on Build GRCh38
rs142539301 Chr.7: 6409864-6409864 on Build GRCh38
rs765187284 Chr.7: 6409817-6409818 on Build GRCh38
rs34950695 Chr.7: 6409823-6409824 on Build GRCh38
rs768801364 Chr.7: 6409849-6409849 on Build GRCh38
rs377670236 Chr.7: 6409828-6409828 on Build GRCh38
rs953333019 Chr.7: 6409474-6409474 on Build GRCh38
rs749309835 Chr.7: 6409892-6409892 on Build GRCh38
rs745875847 Chr.7: 6409772-6409772 on Build GRCh38
rs531365221 Chr.7: 6409726-6409726 on Build GRCh38
rs1044794039 Chr.7: 6409573-6409573 on Build GRCh38
rs763713902 Chr.7: 6409538-6409538 on Build GRCh38
rs745990465 Chr.7: 6409836-6409836 on Build GRCh38
rs535515389 Chr.7: 6409499-6409499 on Build GRCh38
rs201087247 Chr.7: 6409919-6409919 on Build GRCh38
rs702485 Chr.7: 6409641-6409641 on Build GRCh38
rs2303361 Chr.7: 6409865-6409865 on Build GRCh38
rs779525101 Chr.7: 6409740-6409740 on Build GRCh38
rs1008834302 Chr.7: 6409708-6409708 on Build GRCh38
rs763916685 Chr.7: 6409882-6409882 on Build GRCh38
rs911744727 Chr.7: 6409498-6409498 on Build GRCh38
rs566223923 Chr.7: 6409526-6409526 on Build GRCh38
rs769803026 Chr.7: 6409838-6409838 on Build GRCh38
rs899680461 Chr.7: 6409472-6409472 on Build GRCh38
rs939954813 Chr.7: 6409647-6409647 on Build GRCh38
rs752148097 Chr.7: 6409459-6409459 on Build GRCh38
rs190886886 Chr.7: 6409692-6409692 on Build GRCh38
rs201627327 Chr.7: 6409470-6409470 on Build GRCh38
rs1003263146 Chr.7: 6409725-6409725 on Build GRCh38
rs775320101 Chr.7: 6409917-6409917 on Build GRCh38
rs760092073 Chr.7: 6409809-6409809 on Build GRCh38
rs773635530 Chr.7: 6409861-6409861 on Build GRCh38
rs150406824 Chr.7: 6409912-6409912 on Build GRCh38
rs760294556 Chr.7: 6409491-6409491 on Build GRCh38
rs148435637 Chr.7: 6409845-6409845 on Build GRCh38
rs781332327 Chr.7: 6409768-6409768 on Build GRCh38
rs386709825 Chr.7: 6409640-6409641 on Build GRCh38
rs138713047 Chr.7: 6409920-6409920 on Build GRCh38
rs200840062 Chr.7: 6409472-6409473 on Build GRCh38
rs566182147 Chr.7: 6409781-6409781 on Build GRCh38
rs545821078 Chr.7: 6409667-6409667 on Build GRCh38
rs375537082 Chr.7: 6409853-6409853 on Build GRCh38
rs528991002 Chr.7: 6409755-6409755 on Build GRCh38
rs112505366 Chr.7: 6409545-6409545 on Build GRCh38
rs956556175 Chr.7: 6409720-6409720 on Build GRCh38
rs773617603 Chr.7: 6409897-6409897 on Build GRCh38
rs573809815 Chr.7: 6409624-6409624 on Build GRCh38
rs778833804 Chr.7: 6409751-6409751 on Build GRCh38
rs149594736 Chr.7: 6409808-6409808 on Build GRCh38
rs749972928 Chr.7: 6409801-6409801 on Build GRCh38
rs372529111 Chr.7: 6409797-6409797 on Build GRCh38
rs749569067 Chr.7: 6409846-6409846 on Build GRCh38
rs758542007 Chr.7: 6409666-6409666 on Build GRCh38
rs932753389 Chr.7: 6409856-6409856 on Build GRCh38
rs1008674513 Chr.7: 6409874-6409874 on Build GRCh38
rs1018858160 Chr.7: 6409876-6409876 on Build GRCh38
rs368954918 Chr.7: 6409886-6409886 on Build GRCh38
rs753487007 Chr.7: 6409807-6409807 on Build GRCh38
rs779069551 Chr.7: 6409894-6409894 on Build GRCh38
rs944110923 Chr.7: 6409602-6409602 on Build GRCh38
rs372178009 Chr.7: 6409619-6409619 on Build GRCh38
rs755806403 Chr.7: 6409737-6409737 on Build GRCh38
rs776181684 Chr.7: 6409719-6409719 on Build GRCh38
rs749836830 Chr.7: 6409733-6409733 on Build GRCh38
rs996799345 Chr.7: 6409621-6409621 on Build GRCh38
rs926824234 Chr.7: 6409583-6409583 on Build GRCh38
rs555311320 Chr.7: 6409601-6409601 on Build GRCh38
rs924712236 Chr.7: 6409464-6409464 on Build GRCh38
rs547424301 Chr.7: 6409767-6409767 on Build GRCh38
rs977446936 Chr.7: 6409590-6409590 on Build GRCh38
rs747539498 Chr.7: 6409905-6409905 on Build GRCh38
rs547688890 Chr.7: 6409524-6409524 on Build GRCh38
rs576061069 Chr.7: 6409584-6409584 on Build GRCh38
rs116243689 Chr.7: 6409881-6409881 on Build GRCh38
rs397964891 Chr.7: 6409475-6409476 on Build GRCh38
rs372411793 Chr.7: 6409909-6409909 on Build GRCh38
rs757280051 Chr.7: 6409588-6409588 on Build GRCh38
rs74672824 Chr.7: 6409640-6409640 on Build GRCh38
rs138190412 Chr.7: 6409752-6409752 on Build GRCh38
rs1038981882 Chr.7: 6409819-6409819 on Build GRCh38
rs143795237 Chr.7: 6409888-6409888 on Build GRCh38
rs776945676 Chr.7: 6409771-6409772 on Build GRCh38
rs140707721 Chr.7: 6409896-6409896 on Build GRCh38
rs199951557 Chr.7: 6409860-6409860 on Build GRCh38
rs570032882 Chr.7: 6409824-6409824 on Build GRCh38
rs757205077 Chr.7: 6409889-6409889 on Build GRCh38
rs754862251 Chr.7: 6409749-6409749 on Build GRCh38
rs541472579 Chr.7: 6409635-6409635 on Build GRCh38

View Full Product Details