Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 6412875-6413347 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DAGLB
Entrez Gene ID
221955
Gene Name
diacylglycerol lipase beta
Gene Aliases
DAGLBETA, KCCR13L
UniGene
Hs.487498
Gene Chromosome Location
Chr.7: 6409116-6448012 on Build GRCh38
Transcripts
NM_001142936.1 NM_139179.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5504966 c.1453A>G p.S485G Chr.7: 6413009-6413009 on Build GRCh38
6421191 c.1457T>C p.F486S Chr.7: 6413005-6413005 on Build GRCh38
1755337 c.1435C>G p.L479V Chr.7: 6413027-6413027 on Build GRCh38
4789189 c.1496+1G>A p.? Chr.7: 6412965-6412965 on Build GRCh38
4448956 c.1500_1501delCA p.S501fs*20 Chr.7: 6412879-6412880 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs991605147 Chr.7: 6413032-6413032 on Build GRCh38
rs999653892 Chr.7: 6413083-6413083 on Build GRCh38
rs836496 Chr.7: 6413162-6413162 on Build GRCh38
rs771501898 Chr.7: 6412916-6412916 on Build GRCh38
rs149543113 Chr.7: 6413223-6413223 on Build GRCh38
rs182599874 Chr.7: 6413179-6413179 on Build GRCh38
rs943973363 Chr.7: 6413138-6413138 on Build GRCh38
rs545831721 Chr.7: 6413278-6413278 on Build GRCh38
rs931662499 Chr.7: 6413246-6413246 on Build GRCh38
rs771820883 Chr.7: 6413029-6413029 on Build GRCh38
rs374456646 Chr.7: 6413156-6413156 on Build GRCh38
rs527540719 Chr.7: 6413230-6413230 on Build GRCh38
rs374335337 Chr.7: 6413071-6413071 on Build GRCh38
rs996546237 Chr.7: 6413154-6413154 on Build GRCh38
rs570581666 Chr.7: 6412977-6412977 on Build GRCh38
rs780865660 Chr.7: 6412920-6412920 on Build GRCh38
rs747589147 Chr.7: 6412906-6412906 on Build GRCh38
rs1052611639 Chr.7: 6413145-6413145 on Build GRCh38
rs777350065 Chr.7: 6413054-6413054 on Build GRCh38
rs757864497 Chr.7: 6412989-6412989 on Build GRCh38
rs373049215 Chr.7: 6413066-6413066 on Build GRCh38
rs754688760 Chr.7: 6413085-6413085 on Build GRCh38
rs747896238 Chr.7: 6413021-6413021 on Build GRCh38
rs771795599 Chr.7: 6412936-6412937 on Build GRCh38
rs138867213 Chr.7: 6413050-6413050 on Build GRCh38
rs199759442 Chr.7: 6412953-6412953 on Build GRCh38
rs758540406 Chr.7: 6412975-6412975 on Build GRCh38
rs758841943 Chr.7: 6412898-6412898 on Build GRCh38
rs773180405 Chr.7: 6413034-6413034 on Build GRCh38
rs1023654752 Chr.7: 6412945-6412945 on Build GRCh38
rs921710797 Chr.7: 6413221-6413221 on Build GRCh38
rs1014726277 Chr.7: 6413211-6413211 on Build GRCh38
rs189978233 Chr.7: 6412956-6412956 on Build GRCh38
rs1052013040 Chr.7: 6413057-6413057 on Build GRCh38
rs568596616 Chr.7: 6413104-6413104 on Build GRCh38
rs369997708 Chr.7: 6413070-6413070 on Build GRCh38
rs748696546 Chr.7: 6413080-6413080 on Build GRCh38
rs963204058 Chr.7: 6413213-6413213 on Build GRCh38
rs762492032 Chr.7: 6412935-6412935 on Build GRCh38
rs117352842 Chr.7: 6413147-6413147 on Build GRCh38
rs369479950 Chr.7: 6413043-6413043 on Build GRCh38
rs576137004 Chr.7: 6413187-6413187 on Build GRCh38
rs757159181 Chr.7: 6413087-6413087 on Build GRCh38
rs1015106189 Chr.7: 6413180-6413180 on Build GRCh38
rs377671019 Chr.7: 6413074-6413074 on Build GRCh38
rs115329544 Chr.7: 6413198-6413198 on Build GRCh38
rs779418437 Chr.7: 6413000-6413000 on Build GRCh38
rs548461410 Chr.7: 6412952-6412952 on Build GRCh38
rs941800217 Chr.7: 6413010-6413010 on Build GRCh38
rs373578617 Chr.7: 6412947-6412947 on Build GRCh38
rs143863613 Chr.7: 6413258-6413258 on Build GRCh38
rs148059863 Chr.7: 6412988-6412988 on Build GRCh38
rs987544786 Chr.7: 6413247-6413247 on Build GRCh38
rs375202452 Chr.7: 6412979-6412979 on Build GRCh38
rs757737978 Chr.7: 6413064-6413064 on Build GRCh38
rs1052711869 Chr.7: 6413277-6413277 on Build GRCh38
rs552188554 Chr.7: 6413314-6413314 on Build GRCh38
rs1024169117 Chr.7: 6413149-6413149 on Build GRCh38
rs145080190 Chr.7: 6412954-6412954 on Build GRCh38
rs369107448 Chr.7: 6413048-6413048 on Build GRCh38
rs537908568 Chr.7: 6412991-6412991 on Build GRCh38
rs745325623 Chr.7: 6412923-6412923 on Build GRCh38
rs752006642 Chr.7: 6413061-6413061 on Build GRCh38
rs116766804 Chr.7: 6413293-6413293 on Build GRCh38
rs758391168 Chr.7: 6413001-6413001 on Build GRCh38
rs777982248 Chr.7: 6412904-6412904 on Build GRCh38
rs962249443 Chr.7: 6413188-6413188 on Build GRCh38
rs141701599 Chr.7: 6413062-6413062 on Build GRCh38
rs138070509 Chr.7: 6413207-6413207 on Build GRCh38
rs772050079 Chr.7: 6413241-6413241 on Build GRCh38
rs755699927 Chr.7: 6412992-6412992 on Build GRCh38
rs146247471 Chr.7: 6413168-6413168 on Build GRCh38
rs973161684 Chr.7: 6413217-6413217 on Build GRCh38
rs187254386 Chr.7: 6413205-6413205 on Build GRCh38
rs968841720 Chr.7: 6413235-6413235 on Build GRCh38
rs543464621 Chr.7: 6413195-6413195 on Build GRCh38
rs192020988 Chr.7: 6413316-6413316 on Build GRCh38
rs924474491 Chr.7: 6413244-6413244 on Build GRCh38
rs774151497 Chr.7: 6413018-6413018 on Build GRCh38
rs758995609 Chr.7: 6412966-6412966 on Build GRCh38
rs893966695 Chr.7: 6413291-6413291 on Build GRCh38
rs768738450 Chr.7: 6413044-6413047 on Build GRCh38
rs1003513586 Chr.7: 6413318-6413318 on Build GRCh38
rs893866066 Chr.7: 6413146-6413146 on Build GRCh38
rs935582883 Chr.7: 6413271-6413271 on Build GRCh38
rs774065317 Chr.7: 6412941-6412941 on Build GRCh38
rs778821486 Chr.7: 6412930-6412930 on Build GRCh38
rs769364597 Chr.7: 6412931-6412931 on Build GRCh38
rs376018766 Chr.7: 6413052-6413052 on Build GRCh38
rs753017277 Chr.7: 6412967-6412967 on Build GRCh38
rs760440789 Chr.7: 6413317-6413317 on Build GRCh38
rs958744692 Chr.7: 6413027-6413027 on Build GRCh38
rs1007875741 Chr.7: 6412933-6412933 on Build GRCh38
rs750601576 Chr.7: 6412927-6412927 on Build GRCh38

View Full Product Details