Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 7381453-7381968 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL28A1
Entrez Gene ID
340267
Gene Name
collagen type XXVIII alpha 1 chain
Gene Aliases
COL28
UniGene
Hs.491104
Gene Chromosome Location
Chr.7: 7338543-7542062 on Build GRCh38
Transcripts
NM_001037763.2 XM_011515358.2 XM_011515359.2 XM_011515360.2 XM_011515362.2 XM_017012131.1 XM_017012132.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7336967 c.2205+2T>C p.? Chr.7: 7381542-7381542 on Build GRCh38
7352415 c.2174G>A p.G725D Chr.7: 7381575-7381575 on Build GRCh38
4673756 c.2190C>T p.G730G Chr.7: 7381559-7381559 on Build GRCh38
108564 c.2184C>T p.G728G Chr.7: 7381565-7381565 on Build GRCh38
5407886 c.2194C>T p.P732S Chr.7: 7381555-7381555 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369032731 Chr.7: 7381610-7381610 on Build GRCh38
rs376179370 Chr.7: 7381563-7381563 on Build GRCh38
rs769189367 Chr.7: 7381593-7381593 on Build GRCh38
rs758237340 Chr.7: 7381580-7381580 on Build GRCh38
rs540754087 Chr.7: 7381675-7381675 on Build GRCh38
rs544762951 Chr.7: 7381863-7381863 on Build GRCh38
rs774900530 Chr.7: 7381736-7381737 on Build GRCh38
rs747931025 Chr.7: 7381751-7381751 on Build GRCh38
rs1033483740 Chr.7: 7381498-7381498 on Build GRCh38
rs771066060 Chr.7: 7381505-7381505 on Build GRCh38
rs547436883 Chr.7: 7381690-7381691 on Build GRCh38
rs749893839 Chr.7: 7381543-7381543 on Build GRCh38
rs774956623 Chr.7: 7381535-7381535 on Build GRCh38
rs550821423 Chr.7: 7381500-7381500 on Build GRCh38
rs749209613 Chr.7: 7381937-7381937 on Build GRCh38
rs114343166 Chr.7: 7381692-7381692 on Build GRCh38
rs777589199 Chr.7: 7381657-7381657 on Build GRCh38
rs1039254090 Chr.7: 7381515-7381515 on Build GRCh38
rs766033847 Chr.7: 7381547-7381547 on Build GRCh38
rs138688284 Chr.7: 7381606-7381606 on Build GRCh38
rs769941061 Chr.7: 7381585-7381585 on Build GRCh38
rs944906562 Chr.7: 7381556-7381556 on Build GRCh38
rs776059681 Chr.7: 7381528-7381528 on Build GRCh38
rs962059056 Chr.7: 7381519-7381519 on Build GRCh38
rs143382163 Chr.7: 7381565-7381565 on Build GRCh38
rs1018305832 Chr.7: 7381777-7381777 on Build GRCh38
rs773141129 Chr.7: 7381864-7381864 on Build GRCh38
rs1002490424 Chr.7: 7381921-7381921 on Build GRCh38
rs377230561 Chr.7: 7381532-7381532 on Build GRCh38
rs1038476820 Chr.7: 7381845-7381845 on Build GRCh38
rs202213416 Chr.7: 7381592-7381592 on Build GRCh38
rs972064258 Chr.7: 7381522-7381522 on Build GRCh38
rs976679658 Chr.7: 7381869-7381869 on Build GRCh38
rs752430303 Chr.7: 7381651-7381651 on Build GRCh38
rs769476996 Chr.7: 7381809-7381809 on Build GRCh38
rs1024376614 Chr.7: 7381690-7381690 on Build GRCh38
rs758420131 Chr.7: 7381653-7381653 on Build GRCh38
rs267601567 Chr.7: 7381603-7381603 on Build GRCh38
rs573610613 Chr.7: 7381625-7381625 on Build GRCh38
rs373271322 Chr.7: 7381560-7381560 on Build GRCh38
rs986341432 Chr.7: 7381763-7381763 on Build GRCh38
rs765122403 Chr.7: 7381621-7381621 on Build GRCh38
rs779126199 Chr.7: 7381614-7381614 on Build GRCh38
rs966223527 Chr.7: 7381801-7381801 on Build GRCh38
rs773594268 Chr.7: 7381732-7381732 on Build GRCh38
rs749503209 Chr.7: 7381914-7381914 on Build GRCh38
rs1294627 Chr.7: 7381679-7381679 on Build GRCh38
rs993558675 Chr.7: 7381693-7381693 on Build GRCh38
rs534435778 Chr.7: 7381832-7381835 on Build GRCh38
rs76149511 Chr.7: 7381549-7381549 on Build GRCh38
rs765833313 Chr.7: 7381612-7381612 on Build GRCh38
rs532246400 Chr.7: 7381650-7381650 on Build GRCh38
rs35753638 Chr.7: 7381479-7381479 on Build GRCh38
rs745925326 Chr.7: 7381584-7381584 on Build GRCh38
rs770136538 Chr.7: 7381518-7381518 on Build GRCh38
rs184719460 Chr.7: 7381581-7381581 on Build GRCh38
rs780430419 Chr.7: 7381589-7381589 on Build GRCh38
rs555065828 Chr.7: 7381613-7381613 on Build GRCh38
rs746142342 Chr.7: 7381511-7381511 on Build GRCh38
rs762446039 Chr.7: 7381598-7381598 on Build GRCh38
rs755821980 Chr.7: 7381546-7381546 on Build GRCh38
rs200187116 Chr.7: 7381501-7381501 on Build GRCh38
rs1033563575 Chr.7: 7381929-7381929 on Build GRCh38
rs751479364 Chr.7: 7381660-7381660 on Build GRCh38
rs1003869537 Chr.7: 7381834-7381834 on Build GRCh38
rs770065447 Chr.7: 7381545-7381545 on Build GRCh38
rs75350369 Chr.7: 7381575-7381575 on Build GRCh38
rs1027580852 Chr.7: 7381718-7381718 on Build GRCh38
rs754709749 Chr.7: 7381496-7381496 on Build GRCh38
rs898409010 Chr.7: 7381902-7381902 on Build GRCh38
rs896313976 Chr.7: 7381678-7381678 on Build GRCh38
rs375609798 Chr.7: 7381533-7381533 on Build GRCh38
rs1004305202 Chr.7: 7381539-7381539 on Build GRCh38
rs952047471 Chr.7: 7381740-7381740 on Build GRCh38
rs762201820 Chr.7: 7381537-7381537 on Build GRCh38
rs796559921 Chr.7: 7381747-7381748 on Build GRCh38
rs61544045 Chr.7: 7381735-7381736 on Build GRCh38
rs937872246 Chr.7: 7381618-7381618 on Build GRCh38
rs528705287 Chr.7: 7381668-7381668 on Build GRCh38
rs986356220 Chr.7: 7381548-7381548 on Build GRCh38
rs778632121 Chr.7: 7381568-7381568 on Build GRCh38
rs776553065 Chr.7: 7381510-7381510 on Build GRCh38

View Full Product Details