Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 12689851-12690355 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARL4A
Entrez Gene ID
10124
Gene Name
ADP ribosylation factor like GTPase 4A
Gene Aliases
ARL4
UniGene
Hs.245540
Gene Chromosome Location
Chr.7: 12686827-12690934 on Build GRCh38
Transcripts
NM_001037164.2 NM_001195396.1 NM_005738.4 NM_212460.3

SNP Detail

SNP ID Chromosome Location
rs73053491 Chr.7: 12690308-12690308 on Build GRCh38
rs966051346 Chr.7: 12690246-12690246 on Build GRCh38
rs970382299 Chr.7: 12690043-12690044 on Build GRCh38
rs538619784 Chr.7: 12690220-12690220 on Build GRCh38
rs146659511 Chr.7: 12690301-12690301 on Build GRCh38
rs956555692 Chr.7: 12690034-12690034 on Build GRCh38
rs869084944 Chr.7: 12690313-12690314 on Build GRCh38
rs1024867608 Chr.7: 12690234-12690234 on Build GRCh38
rs1012446342 Chr.7: 12690186-12690186 on Build GRCh38
rs186180920 Chr.7: 12689888-12689888 on Build GRCh38
rs1032160051 Chr.7: 12690029-12690029 on Build GRCh38
rs941289042 Chr.7: 12690122-12690122 on Build GRCh38
rs559561062 Chr.7: 12690303-12690303 on Build GRCh38
rs922431940 Chr.7: 12690070-12690070 on Build GRCh38
rs183395893 Chr.7: 12690203-12690203 on Build GRCh38
rs540978308 Chr.7: 12690039-12690039 on Build GRCh38
rs896816802 Chr.7: 12690157-12690157 on Build GRCh38
rs929448824 Chr.7: 12690308-12690309 on Build GRCh38
rs1000324874 Chr.7: 12690015-12690015 on Build GRCh38
rs34958926 Chr.7: 12689956-12689957 on Build GRCh38
rs976135254 Chr.7: 12690261-12690261 on Build GRCh38
rs904561716 Chr.7: 12690167-12690170 on Build GRCh38
rs1038270347 Chr.7: 12689879-12689879 on Build GRCh38
rs963086537 Chr.7: 12690304-12690304 on Build GRCh38
rs922042439 Chr.7: 12690264-12690264 on Build GRCh38
rs929480079 Chr.7: 12690125-12690125 on Build GRCh38
rs62449782 Chr.7: 12689979-12689979 on Build GRCh38
rs976145258 Chr.7: 12690051-12690051 on Build GRCh38
rs890716666 Chr.7: 12690141-12690141 on Build GRCh38
rs890431724 Chr.7: 12689903-12689903 on Build GRCh38
rs968438513 Chr.7: 12690012-12690012 on Build GRCh38
rs563405168 Chr.7: 12690150-12690150 on Build GRCh38
rs372626843 Chr.7: 12689937-12689937 on Build GRCh38
rs971035220 Chr.7: 12690288-12690301 on Build GRCh38
rs543521941 Chr.7: 12690084-12690084 on Build GRCh38
rs1034803333 Chr.7: 12690166-12690166 on Build GRCh38
rs551368390 Chr.7: 12690004-12690004 on Build GRCh38
rs772407322 Chr.7: 12690158-12690158 on Build GRCh38
rs993696523 Chr.7: 12689886-12689886 on Build GRCh38
rs898019937 Chr.7: 12689882-12689882 on Build GRCh38
rs73300797 Chr.7: 12689919-12689919 on Build GRCh38
rs557482380 Chr.7: 12690047-12690047 on Build GRCh38
rs1031767556 Chr.7: 12690197-12690197 on Build GRCh38
rs991064721 Chr.7: 12689977-12689977 on Build GRCh38
rs775796692 Chr.7: 12690211-12690248 on Build GRCh38
rs774868446 Chr.7: 12690234-12690234 on Build GRCh38
rs973088513 Chr.7: 12690307-12690307 on Build GRCh38
rs985232799 Chr.7: 12690292-12690292 on Build GRCh38
rs1030965215 Chr.7: 12690244-12690244 on Build GRCh38
rs917859909 Chr.7: 12690042-12690042 on Build GRCh38
rs116246574 Chr.7: 12690300-12690300 on Build GRCh38
rs535694046 Chr.7: 12689902-12689902 on Build GRCh38
rs1015747858 Chr.7: 12689878-12689878 on Build GRCh38
rs190589614 Chr.7: 12690076-12690076 on Build GRCh38
rs932472893 Chr.7: 12690275-12690275 on Build GRCh38
rs969199321 Chr.7: 12689996-12689996 on Build GRCh38
rs1037449420 Chr.7: 12690112-12690112 on Build GRCh38
rs748979890 Chr.7: 12690050-12690050 on Build GRCh38
rs888399624 Chr.7: 12690088-12690088 on Build GRCh38
rs66751850 Chr.7: 12690314-12690314 on Build GRCh38
rs1036676935 Chr.7: 12690127-12690127 on Build GRCh38
rs113067734 Chr.7: 12690223-12690223 on Build GRCh38
rs959537917 Chr.7: 12689921-12689921 on Build GRCh38
rs895315703 Chr.7: 12690183-12690183 on Build GRCh38

View Full Product Details