Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 16674281-16674813 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BZW2
Entrez Gene ID
28969
Gene Name
basic leucine zipper and W2 domains 2
Gene Aliases
HSPC028, MST017, MSTP017
UniGene
Hs.487635
Gene Chromosome Location
Chr.7: 16646134-16706523 on Build GRCh38
Transcripts
NM_001159767.1 NM_014038.2 NR_027624.1 XM_006715706.1 XM_006715707.1 XM_006715708.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6613765 c.98C>T p.T33I Chr.7: 16674451-16674451 on Build GRCh38
5023313 c.78A>T p.E26D Chr.7: 16674431-16674431 on Build GRCh38
6613766 c.208G>A p.D70N Chr.7: 16674561-16674561 on Build GRCh38
6176944 c.133C>T p.L45F Chr.7: 16674486-16674486 on Build GRCh38
6215112 c.59-4T>G p.? Chr.7: 16674408-16674408 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200769359 Chr.7: 16674625-16674625 on Build GRCh38
rs775814878 Chr.7: 16674392-16674392 on Build GRCh38
rs777968073 Chr.7: 16674319-16674319 on Build GRCh38
rs749609808 Chr.7: 16674416-16674416 on Build GRCh38
rs757585264 Chr.7: 16674428-16674428 on Build GRCh38
rs571928357 Chr.7: 16674488-16674488 on Build GRCh38
rs746385527 Chr.7: 16674391-16674391 on Build GRCh38
rs397707424 Chr.7: 16674316-16674317 on Build GRCh38
rs779812813 Chr.7: 16674762-16674762 on Build GRCh38
rs188990994 Chr.7: 16674775-16674775 on Build GRCh38
rs980522801 Chr.7: 16674690-16674690 on Build GRCh38
rs760636063 Chr.7: 16674464-16674464 on Build GRCh38
rs1006676778 Chr.7: 16674385-16674385 on Build GRCh38
rs751795372 Chr.7: 16674495-16674495 on Build GRCh38
rs531913002 Chr.7: 16674377-16674377 on Build GRCh38
rs397795477 Chr.7: 16674317-16674318 on Build GRCh38
rs776652258 Chr.7: 16674596-16674596 on Build GRCh38
rs766370529 Chr.7: 16674473-16674473 on Build GRCh38
rs927403945 Chr.7: 16674697-16674697 on Build GRCh38
rs199513488 Chr.7: 16674498-16674498 on Build GRCh38
rs536826949 Chr.7: 16674365-16674366 on Build GRCh38
rs753857266 Chr.7: 16674483-16674483 on Build GRCh38
rs1051115280 Chr.7: 16674383-16674383 on Build GRCh38
rs756244692 Chr.7: 16674524-16674524 on Build GRCh38
rs755653753 Chr.7: 16674619-16674622 on Build GRCh38
rs995791739 Chr.7: 16674500-16674500 on Build GRCh38
rs369300617 Chr.7: 16674597-16674597 on Build GRCh38
rs765120867 Chr.7: 16674603-16674603 on Build GRCh38
rs1024725956 Chr.7: 16674643-16674643 on Build GRCh38
rs143752887 Chr.7: 16674605-16674605 on Build GRCh38
rs768991557 Chr.7: 16674554-16674554 on Build GRCh38
rs747239471 Chr.7: 16674367-16674367 on Build GRCh38
rs754753791 Chr.7: 16674486-16674486 on Build GRCh38
rs946022298 Chr.7: 16674767-16674767 on Build GRCh38
rs911620721 Chr.7: 16674640-16674640 on Build GRCh38
rs11465103 Chr.7: 16674308-16674309 on Build GRCh38
rs76743558 Chr.7: 16674318-16674318 on Build GRCh38
rs990782024 Chr.7: 16674720-16674720 on Build GRCh38
rs576147150 Chr.7: 16674757-16674757 on Build GRCh38
rs35233079 Chr.7: 16674484-16674484 on Build GRCh38
rs202219703 Chr.7: 16674619-16674619 on Build GRCh38
rs762074973 Chr.7: 16674613-16674613 on Build GRCh38
rs550412063 Chr.7: 16674434-16674434 on Build GRCh38
rs767517929 Chr.7: 16674564-16674564 on Build GRCh38
rs750520509 Chr.7: 16674518-16674518 on Build GRCh38
rs991862126 Chr.7: 16674638-16674638 on Build GRCh38
rs747801736 Chr.7: 16674566-16674566 on Build GRCh38
rs1033606410 Chr.7: 16674496-16674496 on Build GRCh38
rs760062551 Chr.7: 16674573-16674573 on Build GRCh38
rs772636286 Chr.7: 16674463-16674463 on Build GRCh38
rs372220513 Chr.7: 16674460-16674460 on Build GRCh38
rs1010087484 Chr.7: 16674352-16674352 on Build GRCh38
rs780501687 Chr.7: 16674542-16674542 on Build GRCh38
rs936199630 Chr.7: 16674707-16674707 on Build GRCh38
rs770571445 Chr.7: 16674587-16674587 on Build GRCh38
rs3211210 Chr.7: 16674446-16674446 on Build GRCh38
rs539320736 Chr.7: 16674507-16674507 on Build GRCh38
rs772938890 Chr.7: 16674568-16674568 on Build GRCh38
rs771670267 Chr.7: 16674567-16674567 on Build GRCh38
rs139763605 Chr.7: 16674552-16674552 on Build GRCh38
rs778102314 Chr.7: 16674365-16674365 on Build GRCh38
rs779315221 Chr.7: 16674560-16674560 on Build GRCh38
rs750085179 Chr.7: 16674366-16674369 on Build GRCh38
rs900000218 Chr.7: 16674694-16674694 on Build GRCh38
rs117913417 Chr.7: 16674361-16674361 on Build GRCh38
rs761427539 Chr.7: 16674440-16674440 on Build GRCh38
rs781187973 Chr.7: 16674384-16674384 on Build GRCh38
rs781303797 Chr.7: 16674508-16674508 on Build GRCh38
rs767046038 Chr.7: 16674451-16674451 on Build GRCh38
rs750692811 Chr.7: 16674614-16674614 on Build GRCh38
rs958027414 Chr.7: 16674311-16674311 on Build GRCh38
rs370423915 Chr.7: 16674606-16674606 on Build GRCh38
rs559712082 Chr.7: 16674672-16674672 on Build GRCh38
rs893814266 Chr.7: 16674782-16674782 on Build GRCh38
rs913516068 Chr.7: 16674730-16674730 on Build GRCh38

View Full Product Details