Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 16682412-16682909 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BZW2
Entrez Gene ID
28969
Gene Name
basic leucine zipper and W2 domains 2
Gene Aliases
HSPC028, MST017, MSTP017
UniGene
Hs.487635
Gene Chromosome Location
Chr.7: 16646134-16706523 on Build GRCh38
Transcripts
NM_001159767.1 NM_014038.2 NR_027624.1 XM_006715706.1 XM_006715707.1 XM_006715708.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1088071 c.404A>G p.K135R Chr.7: 16682844-16682844 on Build GRCh38
485153 c.340-2A>T p.? Chr.7: 16682778-16682778 on Build GRCh38
485154 c.380A>G p.K127R Chr.7: 16682820-16682820 on Build GRCh38
6365028 c.340G>T p.V114F Chr.7: 16682780-16682780 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs564499984 Chr.7: 16682741-16682741 on Build GRCh38
rs200606385 Chr.7: 16682879-16682879 on Build GRCh38
rs748709268 Chr.7: 16682768-16682768 on Build GRCh38
rs818504 Chr.7: 16682631-16682631 on Build GRCh38
rs777937538 Chr.7: 16682768-16682769 on Build GRCh38
rs895490136 Chr.7: 16682532-16682533 on Build GRCh38
rs994775513 Chr.7: 16682709-16682709 on Build GRCh38
rs943432608 Chr.7: 16682493-16682493 on Build GRCh38
rs780955589 Chr.7: 16682829-16682829 on Build GRCh38
rs552687866 Chr.7: 16682614-16682614 on Build GRCh38
rs139259521 Chr.7: 16682476-16682476 on Build GRCh38
rs769557614 Chr.7: 16682767-16682767 on Build GRCh38
rs968050246 Chr.7: 16682775-16682775 on Build GRCh38
rs536603336 Chr.7: 16682530-16682530 on Build GRCh38
rs112719307 Chr.7: 16682529-16682529 on Build GRCh38
rs144311925 Chr.7: 16682869-16682869 on Build GRCh38
rs1050051231 Chr.7: 16682625-16682625 on Build GRCh38
rs924061103 Chr.7: 16682806-16682806 on Build GRCh38
rs897316993 Chr.7: 16682688-16682688 on Build GRCh38
rs963487910 Chr.7: 16682586-16682586 on Build GRCh38
rs1610686 Chr.7: 16682438-16682438 on Build GRCh38
rs1016596964 Chr.7: 16682866-16682866 on Build GRCh38
rs1014713405 Chr.7: 16682684-16682684 on Build GRCh38
rs866462656 Chr.7: 16682778-16682778 on Build GRCh38
rs59826374 Chr.7: 16682777-16682778 on Build GRCh38
rs765371736 Chr.7: 16682772-16682772 on Build GRCh38
rs818502 Chr.7: 16682745-16682745 on Build GRCh38
rs541938348 Chr.7: 16682580-16682580 on Build GRCh38
rs919471010 Chr.7: 16682638-16682638 on Build GRCh38
rs529885897 Chr.7: 16682767-16682767 on Build GRCh38
rs901792469 Chr.7: 16682585-16682585 on Build GRCh38
rs548375548 Chr.7: 16682818-16682818 on Build GRCh38
rs563391071 Chr.7: 16682842-16682842 on Build GRCh38
rs961859547 Chr.7: 16682439-16682439 on Build GRCh38
rs774259387 Chr.7: 16682868-16682869 on Build GRCh38
rs1005275064 Chr.7: 16682857-16682857 on Build GRCh38
rs749145105 Chr.7: 16682861-16682861 on Build GRCh38
rs34052310 Chr.7: 16682767-16682768 on Build GRCh38
rs386710757 Chr.7: 16682435-16682438 on Build GRCh38
rs149560698 Chr.7: 16682721-16682721 on Build GRCh38
rs12537678 Chr.7: 16682551-16682551 on Build GRCh38
rs776944988 Chr.7: 16682865-16682866 on Build GRCh38
rs990745027 Chr.7: 16682479-16682479 on Build GRCh38
rs754351432 Chr.7: 16682605-16682605 on Build GRCh38
rs1014371115 Chr.7: 16682533-16682533 on Build GRCh38
rs934447527 Chr.7: 16682761-16682761 on Build GRCh38
rs200567141 Chr.7: 16682820-16682820 on Build GRCh38
rs548164698 Chr.7: 16682477-16682477 on Build GRCh38
rs972257439 Chr.7: 16682462-16682462 on Build GRCh38
rs369984633 Chr.7: 16682821-16682821 on Build GRCh38
rs753131321 Chr.7: 16682782-16682782 on Build GRCh38
rs779678039 Chr.7: 16682858-16682858 on Build GRCh38
rs76482084 Chr.7: 16682734-16682734 on Build GRCh38
rs561172422 Chr.7: 16682591-16682591 on Build GRCh38
rs534405039 Chr.7: 16682581-16682581 on Build GRCh38
rs768490159 Chr.7: 16682867-16682868 on Build GRCh38
rs562191255 Chr.7: 16682758-16682758 on Build GRCh38
rs368856057 Chr.7: 16682752-16682752 on Build GRCh38
rs980943371 Chr.7: 16682447-16682447 on Build GRCh38
rs552154095 Chr.7: 16682535-16682535 on Build GRCh38
rs1610685 Chr.7: 16682435-16682435 on Build GRCh38
rs961204941 Chr.7: 16682726-16682726 on Build GRCh38
rs762626555 Chr.7: 16682867-16682870 on Build GRCh38
rs1023562199 Chr.7: 16682770-16682770 on Build GRCh38
rs369597008 Chr.7: 16682885-16682885 on Build GRCh38
rs77328149 Chr.7: 16682768-16682768 on Build GRCh38
rs1022625683 Chr.7: 16682534-16682534 on Build GRCh38
rs569954845 Chr.7: 16682527-16682527 on Build GRCh38
rs1052949307 Chr.7: 16682765-16682765 on Build GRCh38
rs747128208 Chr.7: 16682770-16682771 on Build GRCh38
rs759011029 Chr.7: 16682787-16682787 on Build GRCh38
rs552114788 Chr.7: 16682833-16682833 on Build GRCh38
rs114291907 Chr.7: 16682473-16682473 on Build GRCh38
rs923783562 Chr.7: 16682553-16682553 on Build GRCh38
rs1042935015 Chr.7: 16682526-16682526 on Build GRCh38
rs201583635 Chr.7: 16682750-16682750 on Build GRCh38
rs942313916 Chr.7: 16682454-16682454 on Build GRCh38
rs909365642 Chr.7: 16682450-16682450 on Build GRCh38
rs574672706 Chr.7: 16682628-16682628 on Build GRCh38
rs557338129 Chr.7: 16682657-16682657 on Build GRCh38
rs904374907 Chr.7: 16682756-16682756 on Build GRCh38
rs1014153412 Chr.7: 16682811-16682811 on Build GRCh38
rs78182563 Chr.7: 16682779-16682779 on Build GRCh38
rs818503 Chr.7: 16682728-16682728 on Build GRCh38

View Full Product Details