Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 90411755-90412250 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLDN12
Entrez Gene ID
9069
Gene Name
claudin 12
Gene Aliases
-
UniGene
Hs.731917
Gene Chromosome Location
Chr.7: 90403334-90415954 on Build GRCh38
Transcripts
NM_001185072.2 NM_001185073.2 NM_012129.4 XM_017012892.1 XM_017012893.1

Gene Symbol
LOC102723899
Entrez Gene ID
102723899
Gene Name
pituitary tumor-transforming gene 1 protein-interacting protein
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.7: 90403379-90513397 on Build GRCh38
Transcripts
NM_001185072.2 NM_001185073.2 NM_012129.4 XM_017012892.1 XM_017012893.1

SNP Detail

SNP ID Chromosome Location
rs966920424 Chr.7: 90411984-90411984 on Build GRCh38
rs1046575872 Chr.7: 90412097-90412097 on Build GRCh38
rs745698884 Chr.7: 90412121-90412121 on Build GRCh38
rs987824381 Chr.7: 90411826-90411826 on Build GRCh38
rs747236371 Chr.7: 90412208-90412208 on Build GRCh38
rs1051152137 Chr.7: 90411847-90411847 on Build GRCh38
rs778624493 Chr.7: 90412095-90412095 on Build GRCh38
rs925253464 Chr.7: 90412031-90412031 on Build GRCh38
rs539571059 Chr.7: 90412188-90412188 on Build GRCh38
rs1006856013 Chr.7: 90411880-90411880 on Build GRCh38
rs973340540 Chr.7: 90412134-90412134 on Build GRCh38
rs150731226 Chr.7: 90412118-90412118 on Build GRCh38
rs1020773675 Chr.7: 90411978-90411978 on Build GRCh38
rs568368853 Chr.7: 90412028-90412028 on Build GRCh38
rs1011180357 Chr.7: 90411976-90411976 on Build GRCh38
rs780463390 Chr.7: 90411888-90411888 on Build GRCh38
rs753554653 Chr.7: 90411815-90411815 on Build GRCh38
rs910163091 Chr.7: 90412096-90412096 on Build GRCh38
rs918776048 Chr.7: 90412143-90412143 on Build GRCh38
rs910451672 Chr.7: 90411849-90411849 on Build GRCh38
rs141519674 Chr.7: 90412064-90412064 on Build GRCh38
rs550071913 Chr.7: 90411986-90411986 on Build GRCh38
rs530391270 Chr.7: 90411896-90411896 on Build GRCh38
rs17862173 Chr.7: 90411810-90411810 on Build GRCh38
rs889808860 Chr.7: 90411857-90411857 on Build GRCh38
rs557654693 Chr.7: 90412150-90412150 on Build GRCh38
rs796678110 Chr.7: 90411808-90411808 on Build GRCh38
rs535700024 Chr.7: 90412045-90412045 on Build GRCh38
rs934122576 Chr.7: 90412171-90412171 on Build GRCh38
rs893721640 Chr.7: 90411957-90411957 on Build GRCh38
rs892656778 Chr.7: 90412077-90412077 on Build GRCh38
rs975399496 Chr.7: 90412017-90412017 on Build GRCh38
rs954562301 Chr.7: 90411816-90411816 on Build GRCh38
rs962972806 Chr.7: 90412117-90412117 on Build GRCh38
rs934050209 Chr.7: 90411846-90411846 on Build GRCh38
rs368462015 Chr.7: 90411872-90411872 on Build GRCh38
rs771727577 Chr.7: 90412168-90412168 on Build GRCh38
rs898831508 Chr.7: 90412200-90412200 on Build GRCh38
rs11760813 Chr.7: 90412216-90412216 on Build GRCh38
rs569574949 Chr.7: 90411852-90411852 on Build GRCh38
rs545400197 Chr.7: 90412042-90412042 on Build GRCh38
rs1034695011 Chr.7: 90412050-90412050 on Build GRCh38
rs1026052766 Chr.7: 90412228-90412228 on Build GRCh38

View Full Product Details