Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 130305731-130306235 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPA4
Entrez Gene ID
51200
Gene Name
carboxypeptidase A4
Gene Aliases
CPA3
UniGene
Hs.93764
Gene Chromosome Location
Chr.7: 130293134-130324180 on Build GRCh38
Transcripts
NM_001163446.1 NM_016352.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5406504 c.541C>T p.H181Y Chr.7: 130305870-130305870 on Build GRCh38
1699355 c.547C>T p.R183* Chr.7: 130305876-130305876 on Build GRCh38
1085520 c.517G>A p.A173T Chr.7: 130305846-130305846 on Build GRCh38
4674283 c.548G>A p.R183Q Chr.7: 130305877-130305877 on Build GRCh38
4674282 c.504C>T p.G168G Chr.7: 130305833-130305833 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369400058 Chr.7: 130305807-130305807 on Build GRCh38
rs373689363 Chr.7: 130305775-130305775 on Build GRCh38
rs568752294 Chr.7: 130306169-130306169 on Build GRCh38
rs374516163 Chr.7: 130305845-130305845 on Build GRCh38
rs933019563 Chr.7: 130306055-130306055 on Build GRCh38
rs561619635 Chr.7: 130305929-130305929 on Build GRCh38
rs375297697 Chr.7: 130305783-130305783 on Build GRCh38
rs918471510 Chr.7: 130305781-130305781 on Build GRCh38
rs1036078732 Chr.7: 130306129-130306129 on Build GRCh38
rs752100399 Chr.7: 130306021-130306021 on Build GRCh38
rs779181523 Chr.7: 130305940-130305940 on Build GRCh38
rs759281126 Chr.7: 130305825-130305825 on Build GRCh38
rs370716350 Chr.7: 130305961-130305961 on Build GRCh38
rs182983229 Chr.7: 130305794-130305794 on Build GRCh38
rs372218349 Chr.7: 130305776-130305776 on Build GRCh38
rs113946505 Chr.7: 130305928-130305928 on Build GRCh38
rs775976932 Chr.7: 130305849-130305849 on Build GRCh38
rs771255976 Chr.7: 130305894-130305894 on Build GRCh38
rs923191050 Chr.7: 130305839-130305839 on Build GRCh38
rs922953847 Chr.7: 130306051-130306051 on Build GRCh38
rs190768992 Chr.7: 130306097-130306097 on Build GRCh38
rs368402149 Chr.7: 130305805-130305805 on Build GRCh38
rs376557337 Chr.7: 130305830-130305830 on Build GRCh38
rs768716046 Chr.7: 130305797-130305797 on Build GRCh38
rs1013066578 Chr.7: 130305976-130305976 on Build GRCh38
rs937127818 Chr.7: 130306034-130306034 on Build GRCh38
rs750073332 Chr.7: 130306106-130306106 on Build GRCh38
rs779132743 Chr.7: 130305874-130305874 on Build GRCh38
rs771811846 Chr.7: 130306149-130306149 on Build GRCh38
rs763598487 Chr.7: 130305840-130305840 on Build GRCh38
rs775735959 Chr.7: 130305789-130305789 on Build GRCh38
rs779886656 Chr.7: 130305931-130305931 on Build GRCh38
rs931917887 Chr.7: 130306058-130306058 on Build GRCh38
rs376601819 Chr.7: 130305960-130305960 on Build GRCh38
rs751553332 Chr.7: 130305841-130305841 on Build GRCh38
rs897590585 Chr.7: 130306163-130306163 on Build GRCh38
rs529185630 Chr.7: 130305944-130305944 on Build GRCh38
rs780949237 Chr.7: 130305781-130305781 on Build GRCh38
rs1034645578 Chr.7: 130306201-130306201 on Build GRCh38
rs761324290 Chr.7: 130305856-130305868 on Build GRCh38
rs749469512 Chr.7: 130305857-130305857 on Build GRCh38
rs772634426 Chr.7: 130305814-130305814 on Build GRCh38
rs369670894 Chr.7: 130305908-130305908 on Build GRCh38
rs965317165 Chr.7: 130306028-130306028 on Build GRCh38
rs927249628 Chr.7: 130306026-130306026 on Build GRCh38
rs1021776283 Chr.7: 130305989-130305989 on Build GRCh38
rs148114709 Chr.7: 130306038-130306039 on Build GRCh38
rs760976885 Chr.7: 130305893-130305893 on Build GRCh38
rs769888943 Chr.7: 130305784-130305784 on Build GRCh38
rs752125645 Chr.7: 130305773-130305773 on Build GRCh38
rs962658708 Chr.7: 130305759-130305759 on Build GRCh38
rs562589354 Chr.7: 130306002-130306002 on Build GRCh38
rs972716000 Chr.7: 130305763-130305763 on Build GRCh38
rs375340609 Chr.7: 130305806-130305806 on Build GRCh38
rs989986967 Chr.7: 130306035-130306035 on Build GRCh38
rs755198740 Chr.7: 130305939-130305939 on Build GRCh38
rs35924272 Chr.7: 130305911-130305911 on Build GRCh38
rs3735053 Chr.7: 130305877-130305877 on Build GRCh38
rs780143875 Chr.7: 130305856-130305856 on Build GRCh38
rs373582879 Chr.7: 130305930-130305930 on Build GRCh38
rs575286796 Chr.7: 130305834-130305834 on Build GRCh38
rs971323542 Chr.7: 130306013-130306013 on Build GRCh38
rs536193059 Chr.7: 130306178-130306178 on Build GRCh38
rs945815363 Chr.7: 130306044-130306044 on Build GRCh38
rs977172899 Chr.7: 130306029-130306029 on Build GRCh38
rs77023166 Chr.7: 130306112-130306112 on Build GRCh38
rs1002887567 Chr.7: 130306199-130306199 on Build GRCh38
rs994240699 Chr.7: 130306133-130306133 on Build GRCh38
rs377447196 Chr.7: 130305900-130305900 on Build GRCh38
rs373418440 Chr.7: 130305812-130305812 on Build GRCh38
rs771257342 Chr.7: 130305962-130305962 on Build GRCh38
rs762369785 Chr.7: 130305835-130305835 on Build GRCh38
rs763658331 Chr.7: 130305910-130305910 on Build GRCh38
rs528468875 Chr.7: 130306111-130306111 on Build GRCh38
rs370825723 Chr.7: 130305858-130305858 on Build GRCh38
rs772237665 Chr.7: 130305948-130305948 on Build GRCh38
rs114997863 Chr.7: 130306072-130306072 on Build GRCh38
rs773805038 Chr.7: 130305886-130305886 on Build GRCh38
rs754076590 Chr.7: 130305934-130305934 on Build GRCh38
rs1025623840 Chr.7: 130306168-130306168 on Build GRCh38
rs34355198 Chr.7: 130306053-130306054 on Build GRCh38
rs766988313 Chr.7: 130306096-130306096 on Build GRCh38
rs1026099417 Chr.7: 130305997-130305997 on Build GRCh38
rs533155699 Chr.7: 130306038-130306038 on Build GRCh38
rs977380354 Chr.7: 130305803-130305803 on Build GRCh38
rs773678294 Chr.7: 130305824-130305824 on Build GRCh38
rs977514705 Chr.7: 130306009-130306009 on Build GRCh38
rs368665966 Chr.7: 130305844-130305844 on Build GRCh38

View Full Product Details