Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 130311947-130312451 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPA4
Entrez Gene ID
51200
Gene Name
carboxypeptidase A4
Gene Aliases
CPA3
UniGene
Hs.93764
Gene Chromosome Location
Chr.7: 130293134-130324180 on Build GRCh38
Transcripts
NM_001163446.1 NM_016352.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5406505 c.1007G>A p.R336K Chr.7: 130312051-130312051 on Build GRCh38
6361179 c.1012G>A p.A338T Chr.7: 130312056-130312056 on Build GRCh38
5938179 c.994G>T p.D332Y Chr.7: 130312038-130312038 on Build GRCh38
5551140 c.1068C>T p.C356C Chr.7: 130312112-130312112 on Build GRCh38
3633255 c.1077C>T p.V359V Chr.7: 130312121-130312121 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756338620 Chr.7: 130312041-130312041 on Build GRCh38
rs774341134 Chr.7: 130312109-130312109 on Build GRCh38
rs548810211 Chr.7: 130312411-130312411 on Build GRCh38
rs772339798 Chr.7: 130312004-130312005 on Build GRCh38
rs149582129 Chr.7: 130312408-130312408 on Build GRCh38
rs367928246 Chr.7: 130312157-130312157 on Build GRCh38
rs754309265 Chr.7: 130312187-130312187 on Build GRCh38
rs775082970 Chr.7: 130312082-130312082 on Build GRCh38
rs779579199 Chr.7: 130312055-130312055 on Build GRCh38
rs372282809 Chr.7: 130312081-130312081 on Build GRCh38
rs529274615 Chr.7: 130312173-130312173 on Build GRCh38
rs766717186 Chr.7: 130312138-130312138 on Build GRCh38
rs747430473 Chr.7: 130312166-130312166 on Build GRCh38
rs771132610 Chr.7: 130312172-130312172 on Build GRCh38
rs77010479 Chr.7: 130312052-130312052 on Build GRCh38
rs369255418 Chr.7: 130311978-130311978 on Build GRCh38
rs767342034 Chr.7: 130312035-130312035 on Build GRCh38
rs1047076061 Chr.7: 130312142-130312142 on Build GRCh38
rs773585979 Chr.7: 130311996-130311996 on Build GRCh38
rs778445407 Chr.7: 130312226-130312226 on Build GRCh38
rs73721866 Chr.7: 130312265-130312265 on Build GRCh38
rs1043313045 Chr.7: 130312304-130312304 on Build GRCh38
rs780090812 Chr.7: 130312044-130312044 on Build GRCh38
rs138167434 Chr.7: 130312245-130312245 on Build GRCh38
rs752177727 Chr.7: 130312003-130312003 on Build GRCh38
rs147548483 Chr.7: 130312038-130312038 on Build GRCh38
rs762452834 Chr.7: 130312096-130312096 on Build GRCh38
rs938673246 Chr.7: 130312184-130312184 on Build GRCh38
rs954394061 Chr.7: 130312070-130312070 on Build GRCh38
rs761780608 Chr.7: 130312123-130312123 on Build GRCh38
rs368498370 Chr.7: 130312048-130312048 on Build GRCh38
rs993720558 Chr.7: 130312405-130312405 on Build GRCh38
rs558258355 Chr.7: 130312098-130312098 on Build GRCh38
rs963541211 Chr.7: 130312043-130312043 on Build GRCh38
rs899027211 Chr.7: 130312353-130312353 on Build GRCh38
rs767265107 Chr.7: 130312130-130312130 on Build GRCh38
rs1007514441 Chr.7: 130312347-130312347 on Build GRCh38
rs745552935 Chr.7: 130312075-130312075 on Build GRCh38
rs756494169 Chr.7: 130312132-130312132 on Build GRCh38
rs73721865 Chr.7: 130312133-130312133 on Build GRCh38
rs776636570 Chr.7: 130312382-130312382 on Build GRCh38
rs986982791 Chr.7: 130312071-130312071 on Build GRCh38
rs148547868 Chr.7: 130312061-130312061 on Build GRCh38
rs563787834 Chr.7: 130312262-130312262 on Build GRCh38
rs754446628 Chr.7: 130312218-130312218 on Build GRCh38
rs1030049192 Chr.7: 130312373-130312373 on Build GRCh38
rs967568099 Chr.7: 130312024-130312024 on Build GRCh38
rs754058607 Chr.7: 130312141-130312141 on Build GRCh38
rs902924539 Chr.7: 130312319-130312319 on Build GRCh38
rs577880214 Chr.7: 130312058-130312058 on Build GRCh38
rs192320982 Chr.7: 130312148-130312148 on Build GRCh38
rs748649596 Chr.7: 130312057-130312057 on Build GRCh38
rs564645887 Chr.7: 130312158-130312158 on Build GRCh38
rs998172044 Chr.7: 130311981-130311981 on Build GRCh38
rs551594721 Chr.7: 130312286-130312286 on Build GRCh38
rs538098001 Chr.7: 130312026-130312026 on Build GRCh38
rs762399844 Chr.7: 130312005-130312005 on Build GRCh38
rs566220896 Chr.7: 130312349-130312349 on Build GRCh38
rs1038015711 Chr.7: 130312351-130312351 on Build GRCh38
rs934815290 Chr.7: 130312180-130312180 on Build GRCh38
rs764690016 Chr.7: 130311999-130311999 on Build GRCh38
rs567215061 Chr.7: 130312426-130312426 on Build GRCh38
rs763536036 Chr.7: 130312097-130312097 on Build GRCh38
rs77268930 Chr.7: 130312429-130312429 on Build GRCh38
rs933994521 Chr.7: 130312256-130312256 on Build GRCh38
rs184184193 Chr.7: 130312018-130312018 on Build GRCh38
rs755044764 Chr.7: 130312049-130312049 on Build GRCh38
rs998540789 Chr.7: 130312321-130312321 on Build GRCh38
rs187503457 Chr.7: 130312020-130312020 on Build GRCh38
rs371522367 Chr.7: 130312292-130312292 on Build GRCh38

View Full Product Details