Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 134442270-134442774 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKR1B1
Entrez Gene ID
231
Gene Name
aldo-keto reductase family 1 member B
Gene Aliases
ADR, ALDR1, ALR2, AR
UniGene
Hs.521212
Gene Chromosome Location
Chr.7: 134442352-134459136 on Build GRCh38
Transcripts
NM_001628.2 XM_005250234.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6321271 c.909-3C>A p.? Chr.7: 134442773-134442773 on Build GRCh38
1568621 c.927T>G p.D309E Chr.7: 134442752-134442752 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs752010122 Chr.7: 134442733-134442733 on Build GRCh38
rs571283804 Chr.7: 134442611-134442611 on Build GRCh38
rs781778959 Chr.7: 134442741-134442741 on Build GRCh38
rs760577936 Chr.7: 134442704-134442704 on Build GRCh38
rs764619605 Chr.7: 134442732-134442732 on Build GRCh38
rs11544690 Chr.7: 134442481-134442481 on Build GRCh38
rs377108391 Chr.7: 134442658-134442658 on Build GRCh38
rs2229543 Chr.7: 134442603-134442603 on Build GRCh38
rs191484045 Chr.7: 134442725-134442725 on Build GRCh38
rs947871253 Chr.7: 134442449-134442449 on Build GRCh38
rs993344719 Chr.7: 134442560-134442560 on Build GRCh38
rs530031579 Chr.7: 134442338-134442351 on Build GRCh38
rs1005992672 Chr.7: 134442596-134442596 on Build GRCh38
rs1803094 Chr.7: 134442419-134442419 on Build GRCh38
rs3087630 Chr.7: 134442519-134442519 on Build GRCh38
rs549872743 Chr.7: 134442395-134442395 on Build GRCh38
rs957131489 Chr.7: 134442633-134442633 on Build GRCh38
rs1803095 Chr.7: 134442645-134442645 on Build GRCh38
rs983507156 Chr.7: 134442618-134442618 on Build GRCh38
rs868158674 Chr.7: 134442540-134442540 on Build GRCh38
rs1030468797 Chr.7: 134442597-134442597 on Build GRCh38
rs888528603 Chr.7: 134442567-134442567 on Build GRCh38
rs915003451 Chr.7: 134442675-134442675 on Build GRCh38
rs1052470989 Chr.7: 134442533-134442533 on Build GRCh38
rs772920425 Chr.7: 134442382-134442382 on Build GRCh38
rs964142762 Chr.7: 134442580-134442580 on Build GRCh38
rs764583222 Chr.7: 134442730-134442730 on Build GRCh38
rs186296854 Chr.7: 134442630-134442630 on Build GRCh38
rs748434538 Chr.7: 134442365-134442365 on Build GRCh38
rs116049631 Chr.7: 134442743-134442743 on Build GRCh38
rs956862151 Chr.7: 134442340-134442340 on Build GRCh38
rs907763391 Chr.7: 134442367-134442368 on Build GRCh38
rs894037874 Chr.7: 134442534-134442534 on Build GRCh38
rs183787048 Chr.7: 134442453-134442453 on Build GRCh38
rs17167851 Chr.7: 134442309-134442309 on Build GRCh38
rs1040236478 Chr.7: 134442486-134442486 on Build GRCh38
rs1003807046 Chr.7: 134442619-134442619 on Build GRCh38
rs928129943 Chr.7: 134442322-134442322 on Build GRCh38
rs1037530388 Chr.7: 134442381-134442381 on Build GRCh38
rs1035232595 Chr.7: 134442621-134442621 on Build GRCh38
rs969612253 Chr.7: 134442304-134442304 on Build GRCh38
rs771790109 Chr.7: 134442319-134442319 on Build GRCh38
rs746497134 Chr.7: 134442562-134442562 on Build GRCh38
rs944642668 Chr.7: 134442479-134442479 on Build GRCh38
rs897660033 Chr.7: 134442506-134442506 on Build GRCh38
rs528058963 Chr.7: 134442484-134442484 on Build GRCh38
rs372374617 Chr.7: 134442710-134442710 on Build GRCh38
rs13306160 Chr.7: 134442648-134442648 on Build GRCh38
rs764730821 Chr.7: 134442712-134442712 on Build GRCh38
rs200286486 Chr.7: 134442709-134442709 on Build GRCh38
rs757815855 Chr.7: 134442737-134442737 on Build GRCh38
rs201593466 Chr.7: 134442700-134442700 on Build GRCh38
rs969349248 Chr.7: 134442612-134442612 on Build GRCh38
rs5055 Chr.7: 134442650-134442650 on Build GRCh38
rs536960470 Chr.7: 134442367-134442367 on Build GRCh38

View Full Product Details