Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 134448809-134449308 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKR1B1
Entrez Gene ID
231
Gene Name
aldo-keto reductase family 1 member B
Gene Aliases
ADR, ALDR1, ALR2, AR
UniGene
Hs.521212
Gene Chromosome Location
Chr.7: 134442352-134459136 on Build GRCh38
Transcripts
NM_001628.2 XM_005250234.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5729996 c.497A>G p.Q166R Chr.7: 134449052-134449052 on Build GRCh38
4153853 c.483C>T p.N161N Chr.7: 134449066-134449066 on Build GRCh38
6597938 c.430-1G>T p.? Chr.7: 134449120-134449120 on Build GRCh38
6499941 c.551A>T p.Q184L Chr.7: 134448998-134448998 on Build GRCh38
5463590 c.442C>T p.L148L Chr.7: 134449107-134449107 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759072459 Chr.7: 134448988-134448988 on Build GRCh38
rs150005764 Chr.7: 134448963-134448963 on Build GRCh38
rs370691275 Chr.7: 134448925-134448925 on Build GRCh38
rs1003776570 Chr.7: 134449037-134449037 on Build GRCh38
rs768726474 Chr.7: 134448952-134448953 on Build GRCh38
rs201824534 Chr.7: 134449143-134449143 on Build GRCh38
rs759780033 Chr.7: 134448996-134448996 on Build GRCh38
rs761706597 Chr.7: 134448971-134448971 on Build GRCh38
rs781297475 Chr.7: 134449148-134449148 on Build GRCh38
rs371330503 Chr.7: 134448969-134448969 on Build GRCh38
rs764636310 Chr.7: 134449138-134449138 on Build GRCh38
rs574032801 Chr.7: 134448964-134448964 on Build GRCh38
rs11544687 Chr.7: 134449028-134449028 on Build GRCh38
rs1056568915 Chr.7: 134449117-134449117 on Build GRCh38
rs1023543431 Chr.7: 134449095-134449095 on Build GRCh38
rs149138376 Chr.7: 134449153-134449153 on Build GRCh38
rs17660462 Chr.7: 134448868-134448868 on Build GRCh38
rs911654122 Chr.7: 134449243-134449245 on Build GRCh38
rs530032632 Chr.7: 134448858-134448858 on Build GRCh38
rs747992861 Chr.7: 134449082-134449082 on Build GRCh38
rs778745784 Chr.7: 134449076-134449076 on Build GRCh38
rs561198533 Chr.7: 134449193-134449193 on Build GRCh38
rs772694816 Chr.7: 134448976-134448976 on Build GRCh38
rs375815110 Chr.7: 134448965-134448965 on Build GRCh38
rs976100580 Chr.7: 134449203-134449203 on Build GRCh38
rs759209480 Chr.7: 134449128-134449128 on Build GRCh38
rs966374892 Chr.7: 134449199-134449199 on Build GRCh38
rs541305306 Chr.7: 134449183-134449183 on Build GRCh38
rs778492841 Chr.7: 134449018-134449018 on Build GRCh38
rs111372981 Chr.7: 134448851-134448851 on Build GRCh38
rs533778853 Chr.7: 134448934-134448934 on Build GRCh38
rs892598346 Chr.7: 134449141-134449141 on Build GRCh38
rs777980833 Chr.7: 134448951-134448951 on Build GRCh38
rs905427503 Chr.7: 134449029-134449029 on Build GRCh38
rs367750648 Chr.7: 134449058-134449058 on Build GRCh38
rs1036111927 Chr.7: 134449268-134449268 on Build GRCh38
rs1022676029 Chr.7: 134449185-134449185 on Build GRCh38
rs184881277 Chr.7: 134449233-134449233 on Build GRCh38
rs572585606 Chr.7: 134449182-134449182 on Build GRCh38
rs773198721 Chr.7: 134448941-134448941 on Build GRCh38
rs773147883 Chr.7: 134449100-134449100 on Build GRCh38
rs774302005 Chr.7: 134448984-134448984 on Build GRCh38
rs371927495 Chr.7: 134449145-134449145 on Build GRCh38
rs1017545692 Chr.7: 134449188-134449188 on Build GRCh38
rs192987063 Chr.7: 134448981-134448981 on Build GRCh38
rs751620614 Chr.7: 134449023-134449023 on Build GRCh38
rs757422740 Chr.7: 134449025-134449025 on Build GRCh38
rs771990751 Chr.7: 134449098-134449098 on Build GRCh38
rs764859838 Chr.7: 134449133-134449133 on Build GRCh38
rs773183131 Chr.7: 134448954-134448954 on Build GRCh38
rs770505903 Chr.7: 134449116-134449116 on Build GRCh38
rs763418044 Chr.7: 134449136-134449136 on Build GRCh38
rs13306162 Chr.7: 134448983-134448983 on Build GRCh38
rs758962979 Chr.7: 134449014-134449014 on Build GRCh38
rs748016824 Chr.7: 134449161-134449161 on Build GRCh38
rs770799641 Chr.7: 134449068-134449069 on Build GRCh38
rs986103331 Chr.7: 134448904-134448904 on Build GRCh38
rs569772783 Chr.7: 134448912-134448912 on Build GRCh38
rs898861518 Chr.7: 134449274-134449274 on Build GRCh38
rs760625485 Chr.7: 134449040-134449042 on Build GRCh38
rs765608056 Chr.7: 134449000-134449000 on Build GRCh38
rs919064348 Chr.7: 134448936-134448936 on Build GRCh38
rs921508518 Chr.7: 134449219-134449219 on Build GRCh38
rs764966557 Chr.7: 134449134-134449134 on Build GRCh38
rs199899097 Chr.7: 134449038-134449038 on Build GRCh38
rs750518284 Chr.7: 134449152-134449152 on Build GRCh38
rs145273429 Chr.7: 134448966-134448966 on Build GRCh38
rs139585031 Chr.7: 134448926-134448926 on Build GRCh38
rs772045887 Chr.7: 134448974-134448974 on Build GRCh38
rs781218518 Chr.7: 134449027-134449027 on Build GRCh38
rs142295480 Chr.7: 134449263-134449263 on Build GRCh38
rs369344364 Chr.7: 134448991-134448991 on Build GRCh38
rs571395236 Chr.7: 134448931-134448931 on Build GRCh38
rs756345099 Chr.7: 134449168-134449168 on Build GRCh38
rs746128224 Chr.7: 134449072-134449072 on Build GRCh38
rs747147279 Chr.7: 134448952-134448952 on Build GRCh38
rs934166547 Chr.7: 134449227-134449227 on Build GRCh38
rs760130465 Chr.7: 134449110-134449110 on Build GRCh38
rs780206121 Chr.7: 134449169-134449169 on Build GRCh38
rs769948905 Chr.7: 134449075-134449075 on Build GRCh38
rs995203338 Chr.7: 134449015-134449015 on Build GRCh38
rs139346970 Chr.7: 134449133-134449136 on Build GRCh38
rs765891820 Chr.7: 134448990-134448990 on Build GRCh38
rs975991520 Chr.7: 134449210-134449210 on Build GRCh38
rs187287203 Chr.7: 134449264-134449264 on Build GRCh38

View Full Product Details