Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 134933996-134934509 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CALD1
Entrez Gene ID
800
Gene Name
caldesmon 1
Gene Aliases
CDM, H-CAD, HCAD, L-CAD, LCAD, NAG22
UniGene
Hs.490203
Gene Chromosome Location
Chr.7: 134779413-134970729 on Build GRCh38
Transcripts
NM_004342.6 NM_033138.3 NM_033139.3 NM_033140.3 NM_033157.3 XM_017012650.1 XM_017012651.1 XM_017012652.1 XM_017012653.1 XM_017012654.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5406557 c.1233G>A p.Q411Q Chr.7: 134934002-134934002 on Build GRCh38
6619049 c.1302G>T p.K434N Chr.7: 134934071-134934071 on Build GRCh38
7102529 c.1276G>C p.D426H Chr.7: 134934045-134934045 on Build GRCh38
5467023 c.1291G>A p.A431T Chr.7: 134934060-134934060 on Build GRCh38
4463549 c.1297C>T p.L433L Chr.7: 134934066-134934066 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751500848 Chr.7: 134934100-134934100 on Build GRCh38
rs1022184088 Chr.7: 134934431-134934431 on Build GRCh38
rs987312431 Chr.7: 134934137-134934137 on Build GRCh38
rs930413973 Chr.7: 134934235-134934235 on Build GRCh38
rs762289688 Chr.7: 134934050-134934050 on Build GRCh38
rs1041225374 Chr.7: 134934110-134934110 on Build GRCh38
rs769014173 Chr.7: 134934042-134934042 on Build GRCh38
rs545049372 Chr.7: 134934288-134934288 on Build GRCh38
rs769398416 Chr.7: 134934021-134934021 on Build GRCh38
rs774921696 Chr.7: 134934046-134934046 on Build GRCh38
rs773207941 Chr.7: 134934059-134934059 on Build GRCh38
rs772171717 Chr.7: 134934058-134934058 on Build GRCh38
rs920232116 Chr.7: 134934220-134934220 on Build GRCh38
rs1043768779 Chr.7: 134934392-134934392 on Build GRCh38
rs985775193 Chr.7: 134934458-134934458 on Build GRCh38
rs888456742 Chr.7: 134934238-134934238 on Build GRCh38
rs892718947 Chr.7: 134934312-134934313 on Build GRCh38
rs559029235 Chr.7: 134934311-134934311 on Build GRCh38
rs763860435 Chr.7: 134934095-134934095 on Build GRCh38
rs777091370 Chr.7: 134934221-134934221 on Build GRCh38
rs749597641 Chr.7: 134934026-134934026 on Build GRCh38
rs756630579 Chr.7: 134934295-134934295 on Build GRCh38
rs749625165 Chr.7: 134934474-134934474 on Build GRCh38
rs926361231 Chr.7: 134934292-134934292 on Build GRCh38
rs376074647 Chr.7: 134934063-134934063 on Build GRCh38
rs780721505 Chr.7: 134934111-134934111 on Build GRCh38
rs972145318 Chr.7: 134934205-134934205 on Build GRCh38
rs1034815142 Chr.7: 134934351-134934351 on Build GRCh38
rs1045556386 Chr.7: 134934285-134934285 on Build GRCh38
rs968137456 Chr.7: 134934432-134934432 on Build GRCh38
rs145835307 Chr.7: 134934081-134934081 on Build GRCh38
rs939554605 Chr.7: 134934307-134934307 on Build GRCh38
rs930192703 Chr.7: 134934146-134934146 on Build GRCh38
rs773546022 Chr.7: 134934072-134934072 on Build GRCh38
rs942853792 Chr.7: 134934103-134934103 on Build GRCh38
rs945620953 Chr.7: 134934389-134934389 on Build GRCh38
rs74364650 Chr.7: 134934241-134934241 on Build GRCh38
rs866201523 Chr.7: 134934139-134934139 on Build GRCh38
rs6956245 Chr.7: 134934181-134934181 on Build GRCh38
rs371013211 Chr.7: 134934125-134934125 on Build GRCh38
rs529596635 Chr.7: 134934335-134934335 on Build GRCh38
rs572411379 Chr.7: 134934355-134934355 on Build GRCh38
rs536539795 Chr.7: 134934143-134934143 on Build GRCh38
rs751022050 Chr.7: 134934213-134934213 on Build GRCh38
rs192091573 Chr.7: 134934404-134934404 on Build GRCh38
rs757189115 Chr.7: 134934109-134934109 on Build GRCh38
rs12707192 Chr.7: 134934415-134934415 on Build GRCh38
rs35887182 Chr.7: 134934355-134934355 on Build GRCh38
rs567668088 Chr.7: 134934121-134934121 on Build GRCh38
rs1009886445 Chr.7: 134934429-134934429 on Build GRCh38
rs1003367502 Chr.7: 134934312-134934312 on Build GRCh38

View Full Product Details