Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 139374145-139374671 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C7orf55-LUC7L2
Entrez Gene ID
100996928
Gene Name
C7orf55-LUC7L2 readthrough
Gene Aliases
-
UniGene
Hs.731488
Gene Chromosome Location
Chr.7: 139341132-139423457 on Build GRCh38
Transcripts
NM_001244584.2 NM_001244585.1 NM_001270643.1 NM_016019.4

Gene Symbol
LUC7L2
Entrez Gene ID
51631
Gene Name
LUC7 like 2, pre-mRNA splicing factor
Gene Aliases
CGI-59, CGI-74, LUC7B2
UniGene
Hs.731488
Gene Chromosome Location
Chr.7: 139340359-139423457 on Build GRCh38
Transcripts
NM_001244584.2 NM_001244585.1 NM_001270643.1 NM_016019.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6230610 c.44C>T p.P15L Chr.7: 139374457-139374457 on Build GRCh38
7230713 c.41C>A p.A14D Chr.7: 139374454-139374454 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs948245517 Chr.7: 139374246-139374246 on Build GRCh38
rs971313212 Chr.7: 139374648-139374648 on Build GRCh38
rs749401623 Chr.7: 139374471-139374471 on Build GRCh38
rs183170981 Chr.7: 139374637-139374637 on Build GRCh38
rs570490531 Chr.7: 139374215-139374215 on Build GRCh38
rs777658960 Chr.7: 139374505-139374505 on Build GRCh38
rs939117714 Chr.7: 139374630-139374630 on Build GRCh38
rs954834621 Chr.7: 139374480-139374480 on Build GRCh38
rs200579985 Chr.7: 139374198-139374199 on Build GRCh38
rs539725368 Chr.7: 139374519-139374519 on Build GRCh38
rs558478343 Chr.7: 139374544-139374544 on Build GRCh38
rs187353705 Chr.7: 139374301-139374301 on Build GRCh38
rs957326460 Chr.7: 139374626-139374626 on Build GRCh38
rs920849402 Chr.7: 139374570-139374570 on Build GRCh38
rs561410429 Chr.7: 139374612-139374612 on Build GRCh38
rs1037947503 Chr.7: 139374202-139374202 on Build GRCh38
rs975280487 Chr.7: 139374360-139374360 on Build GRCh38
rs756287350 Chr.7: 139374430-139374430 on Build GRCh38
rs907850721 Chr.7: 139374166-139374166 on Build GRCh38
rs988510916 Chr.7: 139374628-139374628 on Build GRCh38
rs568219251 Chr.7: 139374258-139374258 on Build GRCh38
rs929995931 Chr.7: 139374365-139374365 on Build GRCh38
rs746416897 Chr.7: 139374414-139374414 on Build GRCh38
rs1018475901 Chr.7: 139374584-139374584 on Build GRCh38
rs891390041 Chr.7: 139374482-139374482 on Build GRCh38
rs940656141 Chr.7: 139374526-139374526 on Build GRCh38
rs537895198 Chr.7: 139374235-139374235 on Build GRCh38
rs973692354 Chr.7: 139374542-139374542 on Build GRCh38
rs899997324 Chr.7: 139374600-139374600 on Build GRCh38
rs572637547 Chr.7: 139374481-139374481 on Build GRCh38
rs780490989 Chr.7: 139374457-139374457 on Build GRCh38
rs1038446547 Chr.7: 139374281-139374281 on Build GRCh38
rs190670670 Chr.7: 139374546-139374546 on Build GRCh38
rs1022165491 Chr.7: 139374330-139374330 on Build GRCh38
rs753216686 Chr.7: 139374351-139374351 on Build GRCh38
rs539946622 Chr.7: 139374255-139374255 on Build GRCh38
rs1030643200 Chr.7: 139374293-139374293 on Build GRCh38
rs1052122381 Chr.7: 139374633-139374633 on Build GRCh38
rs1032768081 Chr.7: 139374615-139374615 on Build GRCh38
rs748312014 Chr.7: 139374512-139374512 on Build GRCh38
rs1010801803 Chr.7: 139374319-139374319 on Build GRCh38
rs536020126 Chr.7: 139374264-139374264 on Build GRCh38
rs1008470579 Chr.7: 139374564-139374564 on Build GRCh38

View Full Product Details