Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 139561153-139561692 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIPK2
Entrez Gene ID
28996
Gene Name
homeodomain interacting protein kinase 2
Gene Aliases
PRO0593
UniGene
Hs.731417
Gene Chromosome Location
Chr.7: 139561570-139777894 on Build GRCh38
Transcripts
NM_001113239.2 NM_022740.4 XM_006715935.2 XM_011516077.2 XM_011516078.2 XM_011516079.2 XM_011516080.2 XM_011516081.2 XM_017012069.1

SNP Detail

SNP ID Chromosome Location
rs1019339444 Chr.7: 139561315-139561315 on Build GRCh38
rs899321705 Chr.7: 139561207-139561207 on Build GRCh38
rs545787372 Chr.7: 139561268-139561268 on Build GRCh38
rs944914656 Chr.7: 139561604-139561604 on Build GRCh38
rs990564177 Chr.7: 139561426-139561426 on Build GRCh38
rs557033249 Chr.7: 139561226-139561226 on Build GRCh38
rs1020858879 Chr.7: 139561374-139561374 on Build GRCh38
rs984920700 Chr.7: 139561363-139561363 on Build GRCh38
rs1041990711 Chr.7: 139561482-139561482 on Build GRCh38
rs991688388 Chr.7: 139561419-139561419 on Build GRCh38
rs192548036 Chr.7: 139561212-139561212 on Build GRCh38
rs930785045 Chr.7: 139561460-139561460 on Build GRCh38
rs531416808 Chr.7: 139561318-139561318 on Build GRCh38
rs559755254 Chr.7: 139561193-139561193 on Build GRCh38
rs568240380 Chr.7: 139561402-139561402 on Build GRCh38
rs113451041 Chr.7: 139561566-139561566 on Build GRCh38
rs926382231 Chr.7: 139561367-139561367 on Build GRCh38
rs774466351 Chr.7: 139561580-139561581 on Build GRCh38
rs927582693 Chr.7: 139561403-139561403 on Build GRCh38
rs989299368 Chr.7: 139561364-139561364 on Build GRCh38
rs1048332444 Chr.7: 139561484-139561484 on Build GRCh38
rs776080595 Chr.7: 139561662-139561665 on Build GRCh38
rs1009164968 Chr.7: 139561289-139561289 on Build GRCh38
rs1054558289 Chr.7: 139561267-139561268 on Build GRCh38
rs549635272 Chr.7: 139561396-139561396 on Build GRCh38
rs1054886752 Chr.7: 139561204-139561204 on Build GRCh38
rs547574352 Chr.7: 139561554-139561554 on Build GRCh38
rs920365806 Chr.7: 139561459-139561459 on Build GRCh38
rs967002593 Chr.7: 139561381-139561381 on Build GRCh38
rs958857089 Chr.7: 139561412-139561412 on Build GRCh38
rs917542136 Chr.7: 139561188-139561188 on Build GRCh38
rs1033091656 Chr.7: 139561328-139561328 on Build GRCh38
rs982216717 Chr.7: 139561192-139561192 on Build GRCh38
rs1026245245 Chr.7: 139561652-139561652 on Build GRCh38
rs937716104 Chr.7: 139561201-139561201 on Build GRCh38
rs1005923658 Chr.7: 139561658-139561658 on Build GRCh38
rs372066835 Chr.7: 139561327-139561327 on Build GRCh38
rs539366655 Chr.7: 139561586-139561586 on Build GRCh38
rs951766361 Chr.7: 139561351-139561351 on Build GRCh38
rs528404833 Chr.7: 139561362-139561362 on Build GRCh38
rs957865285 Chr.7: 139561331-139561331 on Build GRCh38
rs1018816427 Chr.7: 139561298-139561298 on Build GRCh38
rs996174223 Chr.7: 139561669-139561669 on Build GRCh38
rs993436581 Chr.7: 139561325-139561325 on Build GRCh38
rs951791615 Chr.7: 139561655-139561655 on Build GRCh38
rs936044643 Chr.7: 139561495-139561495 on Build GRCh38
rs763752452 Chr.7: 139561326-139561326 on Build GRCh38
rs541702424 Chr.7: 139561187-139561187 on Build GRCh38
rs780241610 Chr.7: 139561545-139561546 on Build GRCh38
rs556583638 Chr.7: 139561669-139561670 on Build GRCh38
rs993800682 Chr.7: 139561646-139561647 on Build GRCh38
rs1040639006 Chr.7: 139561612-139561612 on Build GRCh38
rs950241503 Chr.7: 139561475-139561478 on Build GRCh38
rs901689510 Chr.7: 139561614-139561614 on Build GRCh38
rs900494262 Chr.7: 139561619-139561619 on Build GRCh38
rs924577619 Chr.7: 139561482-139561482 on Build GRCh38
rs1047988667 Chr.7: 139561236-139561236 on Build GRCh38
rs184845600 Chr.7: 139561425-139561425 on Build GRCh38
rs888934906 Chr.7: 139561267-139561267 on Build GRCh38
rs917427719 Chr.7: 139561432-139561432 on Build GRCh38
rs1026363721 Chr.7: 139561348-139561348 on Build GRCh38
rs564086557 Chr.7: 139561278-139561278 on Build GRCh38
rs965968645 Chr.7: 139561319-139561320 on Build GRCh38

View Full Product Details