Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 139564225-139564728 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIPK2
Entrez Gene ID
28996
Gene Name
homeodomain interacting protein kinase 2
Gene Aliases
PRO0593
UniGene
Hs.731417
Gene Chromosome Location
Chr.7: 139561570-139777894 on Build GRCh38
Transcripts
NM_001113239.2 NM_022740.4 XM_006715935.2 XM_011516077.2 XM_011516078.2 XM_011516079.2 XM_011516080.2 XM_011516081.2 XM_017012069.1

SNP Detail

SNP ID Chromosome Location
rs978800022 Chr.7: 139564269-139564269 on Build GRCh38
rs888669524 Chr.7: 139564306-139564306 on Build GRCh38
rs866651825 Chr.7: 139564296-139564296 on Build GRCh38
rs546459700 Chr.7: 139564624-139564624 on Build GRCh38
rs548502520 Chr.7: 139564469-139564469 on Build GRCh38
rs760781453 Chr.7: 139564620-139564620 on Build GRCh38
rs913875431 Chr.7: 139564411-139564411 on Build GRCh38
rs865820018 Chr.7: 139564425-139564425 on Build GRCh38
rs897168133 Chr.7: 139564474-139564474 on Build GRCh38
rs988816221 Chr.7: 139564408-139564408 on Build GRCh38
rs1003036957 Chr.7: 139564542-139564543 on Build GRCh38
rs1037371684 Chr.7: 139564502-139564502 on Build GRCh38
rs146196505 Chr.7: 139564286-139564286 on Build GRCh38
rs141513603 Chr.7: 139564285-139564285 on Build GRCh38
rs979468317 Chr.7: 139564439-139564439 on Build GRCh38
rs546172475 Chr.7: 139564510-139564510 on Build GRCh38
rs28570373 Chr.7: 139564476-139564476 on Build GRCh38
rs955759628 Chr.7: 139564574-139564574 on Build GRCh38
rs79697516 Chr.7: 139564557-139564557 on Build GRCh38
rs961173381 Chr.7: 139564576-139564576 on Build GRCh38
rs1051451132 Chr.7: 139564273-139564273 on Build GRCh38
rs995239809 Chr.7: 139564335-139564335 on Build GRCh38
rs968999306 Chr.7: 139564521-139564521 on Build GRCh38
rs868229630 Chr.7: 139564623-139564623 on Build GRCh38
rs760161900 Chr.7: 139564480-139564480 on Build GRCh38
rs921388394 Chr.7: 139564468-139564468 on Build GRCh38
rs148004934 Chr.7: 139564478-139564478 on Build GRCh38
rs1035604839 Chr.7: 139564394-139564394 on Build GRCh38
rs774758186 Chr.7: 139564406-139564406 on Build GRCh38
rs186411867 Chr.7: 139564592-139564592 on Build GRCh38
rs73156835 Chr.7: 139564680-139564680 on Build GRCh38
rs562931851 Chr.7: 139564371-139564371 on Build GRCh38
rs934381100 Chr.7: 139564278-139564278 on Build GRCh38
rs752887273 Chr.7: 139564618-139564618 on Build GRCh38
rs924302787 Chr.7: 139564277-139564277 on Build GRCh38
rs575251945 Chr.7: 139564664-139564664 on Build GRCh38
rs978718691 Chr.7: 139564549-139564549 on Build GRCh38
rs969540753 Chr.7: 139564361-139564361 on Build GRCh38
rs376180398 Chr.7: 139564358-139564358 on Build GRCh38
rs947046321 Chr.7: 139564260-139564260 on Build GRCh38
rs1005805969 Chr.7: 139564338-139564338 on Build GRCh38
rs530273996 Chr.7: 139564431-139564431 on Build GRCh38
rs1616786 Chr.7: 139564518-139564518 on Build GRCh38
rs905575263 Chr.7: 139564536-139564536 on Build GRCh38
rs898143104 Chr.7: 139564508-139564508 on Build GRCh38
rs1044664069 Chr.7: 139564302-139564302 on Build GRCh38
rs868710819 Chr.7: 139564307-139564307 on Build GRCh38
rs17160987 Chr.7: 139564668-139564668 on Build GRCh38
rs980996224 Chr.7: 139564373-139564373 on Build GRCh38
rs939931277 Chr.7: 139564489-139564489 on Build GRCh38

View Full Product Details