Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 139566516-139567020 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIPK2
Entrez Gene ID
28996
Gene Name
homeodomain interacting protein kinase 2
Gene Aliases
PRO0593
UniGene
Hs.731417
Gene Chromosome Location
Chr.7: 139561570-139777894 on Build GRCh38
Transcripts
NM_001113239.2 NM_022740.4 XM_006715935.2 XM_011516077.2 XM_011516078.2 XM_011516079.2 XM_011516080.2 XM_011516081.2 XM_017012069.1

SNP Detail

SNP ID Chromosome Location
rs1001477779 Chr.7: 139566869-139566869 on Build GRCh38
rs917927098 Chr.7: 139566702-139566702 on Build GRCh38
rs1018370469 Chr.7: 139566630-139566630 on Build GRCh38
rs1020079311 Chr.7: 139566602-139566602 on Build GRCh38
rs761977244 Chr.7: 139566592-139566592 on Build GRCh38
rs17261671 Chr.7: 139566810-139566810 on Build GRCh38
rs531902897 Chr.7: 139566625-139566625 on Build GRCh38
rs767445351 Chr.7: 139566913-139566913 on Build GRCh38
rs556055563 Chr.7: 139566748-139566748 on Build GRCh38
rs1041505114 Chr.7: 139566914-139566914 on Build GRCh38
rs974276637 Chr.7: 139566986-139566986 on Build GRCh38
rs145349975 Chr.7: 139566653-139566653 on Build GRCh38
rs909865392 Chr.7: 139566852-139566852 on Build GRCh38
rs545994183 Chr.7: 139566559-139566559 on Build GRCh38
rs899998141 Chr.7: 139566865-139566865 on Build GRCh38
rs974014316 Chr.7: 139566779-139566779 on Build GRCh38
rs956724777 Chr.7: 139566655-139566655 on Build GRCh38
rs375418564 Chr.7: 139566613-139566613 on Build GRCh38
rs535443939 Chr.7: 139566949-139566949 on Build GRCh38
rs965926381 Chr.7: 139566809-139566809 on Build GRCh38
rs950188849 Chr.7: 139566895-139566895 on Build GRCh38
rs373603779 Chr.7: 139566802-139566802 on Build GRCh38
rs71543337 Chr.7: 139566727-139566727 on Build GRCh38
rs888772293 Chr.7: 139566970-139566970 on Build GRCh38
rs903045357 Chr.7: 139566916-139566916 on Build GRCh38
rs571072699 Chr.7: 139566870-139566870 on Build GRCh38
rs970745839 Chr.7: 139566929-139566929 on Build GRCh38
rs564390154 Chr.7: 139566585-139566585 on Build GRCh38
rs967236258 Chr.7: 139566620-139566620 on Build GRCh38
rs937195630 Chr.7: 139566769-139566769 on Build GRCh38
rs1032618544 Chr.7: 139566665-139566665 on Build GRCh38
rs6959392 Chr.7: 139566707-139566707 on Build GRCh38
rs895938309 Chr.7: 139566891-139566891 on Build GRCh38
rs1007933063 Chr.7: 139566629-139566629 on Build GRCh38
rs568789940 Chr.7: 139566692-139566692 on Build GRCh38
rs71520060 Chr.7: 139566689-139566689 on Build GRCh38
rs980833955 Chr.7: 139566951-139566951 on Build GRCh38
rs964590329 Chr.7: 139566646-139566646 on Build GRCh38
rs1046672414 Chr.7: 139566811-139566811 on Build GRCh38
rs1040913807 Chr.7: 139566607-139566607 on Build GRCh38
rs74864132 Chr.7: 139566741-139566741 on Build GRCh38
rs892673263 Chr.7: 139566879-139566879 on Build GRCh38
rs898142349 Chr.7: 139566608-139566608 on Build GRCh38
rs919869660 Chr.7: 139566808-139566808 on Build GRCh38
rs765488488 Chr.7: 139566650-139566650 on Build GRCh38
rs1046789145 Chr.7: 139566624-139566624 on Build GRCh38
rs958556179 Chr.7: 139566976-139566976 on Build GRCh38
rs895624441 Chr.7: 139566578-139566578 on Build GRCh38
rs970778862 Chr.7: 139566706-139566706 on Build GRCh38
rs1033802869 Chr.7: 139566969-139566969 on Build GRCh38
rs927098211 Chr.7: 139566750-139566750 on Build GRCh38
rs1039454009 Chr.7: 139566847-139566847 on Build GRCh38
rs1024950638 Chr.7: 139566927-139566927 on Build GRCh38
rs890808869 Chr.7: 139566815-139566815 on Build GRCh38
rs943778325 Chr.7: 139566833-139566833 on Build GRCh38

View Full Product Details