Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 139568059-139568564 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIPK2
Entrez Gene ID
28996
Gene Name
homeodomain interacting protein kinase 2
Gene Aliases
PRO0593
UniGene
Hs.731417
Gene Chromosome Location
Chr.7: 139561570-139777894 on Build GRCh38
Transcripts
NM_001113239.2 NM_022740.4 XM_006715935.2 XM_011516077.2 XM_011516078.2 XM_011516079.2 XM_011516080.2 XM_011516081.2 XM_017012069.1

SNP Detail

SNP ID Chromosome Location
rs965120372 Chr.7: 139568299-139568299 on Build GRCh38
rs907864155 Chr.7: 139568341-139568341 on Build GRCh38
rs17261727 Chr.7: 139568522-139568522 on Build GRCh38
rs777965460 Chr.7: 139568388-139568388 on Build GRCh38
rs1040461536 Chr.7: 139568084-139568084 on Build GRCh38
rs1040427342 Chr.7: 139568359-139568359 on Build GRCh38
rs749311776 Chr.7: 139568188-139568188 on Build GRCh38
rs893685319 Chr.7: 139568115-139568115 on Build GRCh38
rs376666834 Chr.7: 139568372-139568372 on Build GRCh38
rs990859013 Chr.7: 139568431-139568431 on Build GRCh38
rs184327908 Chr.7: 139568468-139568468 on Build GRCh38
rs929186356 Chr.7: 139568102-139568102 on Build GRCh38
rs541813128 Chr.7: 139568119-139568119 on Build GRCh38
rs115766907 Chr.7: 139568208-139568208 on Build GRCh38
rs763027551 Chr.7: 139568334-139568342 on Build GRCh38
rs751711373 Chr.7: 139568474-139568474 on Build GRCh38
rs535754827 Chr.7: 139568446-139568446 on Build GRCh38
rs759357603 Chr.7: 139568300-139568300 on Build GRCh38
rs1020601216 Chr.7: 139568148-139568148 on Build GRCh38
rs757483564 Chr.7: 139568518-139568518 on Build GRCh38
rs920781744 Chr.7: 139568277-139568277 on Build GRCh38
rs928794031 Chr.7: 139568514-139568514 on Build GRCh38
rs1049554486 Chr.7: 139568413-139568413 on Build GRCh38
rs934863185 Chr.7: 139568517-139568517 on Build GRCh38
rs188984492 Chr.7: 139568523-139568523 on Build GRCh38
rs900549077 Chr.7: 139568381-139568381 on Build GRCh38
rs932061821 Chr.7: 139568411-139568411 on Build GRCh38
rs564295814 Chr.7: 139568249-139568249 on Build GRCh38
rs544178607 Chr.7: 139568242-139568242 on Build GRCh38
rs531364648 Chr.7: 139568262-139568262 on Build GRCh38
rs552346154 Chr.7: 139568241-139568241 on Build GRCh38
rs990966326 Chr.7: 139568256-139568256 on Build GRCh38
rs949133332 Chr.7: 139568463-139568463 on Build GRCh38
rs916261937 Chr.7: 139568434-139568434 on Build GRCh38
rs562776101 Chr.7: 139568507-139568507 on Build GRCh38
rs944747435 Chr.7: 139568342-139568342 on Build GRCh38
rs901009215 Chr.7: 139568099-139568099 on Build GRCh38
rs1046289631 Chr.7: 139568185-139568188 on Build GRCh38
rs767349529 Chr.7: 139568520-139568520 on Build GRCh38
rs949171679 Chr.7: 139568165-139568165 on Build GRCh38
rs188097975 Chr.7: 139568106-139568106 on Build GRCh38
rs1028582561 Chr.7: 139568110-139568110 on Build GRCh38
rs376212354 Chr.7: 139568116-139568116 on Build GRCh38
rs1017605943 Chr.7: 139568295-139568295 on Build GRCh38
rs1010810667 Chr.7: 139568481-139568481 on Build GRCh38
rs191747147 Chr.7: 139568322-139568322 on Build GRCh38
rs140559824 Chr.7: 139568164-139568164 on Build GRCh38
rs941961652 Chr.7: 139568417-139568417 on Build GRCh38
rs1031912230 Chr.7: 139568213-139568213 on Build GRCh38
rs73485514 Chr.7: 139568306-139568306 on Build GRCh38

View Full Product Details