Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 141929105-141929599 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLEC5A
Entrez Gene ID
23601
Gene Name
C-type lectin domain family 5 member A
Gene Aliases
CLECSF5, MDL-1, MDL1
UniGene
Hs.446235
Gene Chromosome Location
Chr.7: 141927356-141947009 on Build GRCh38
Transcripts
NM_001301167.1 NM_013252.2 XM_011515995.2 XM_017011915.1 XM_017011917.1

SNP Detail

SNP ID Chromosome Location
rs968930899 Chr.7: 141929330-141929330 on Build GRCh38
rs551116468 Chr.7: 141929273-141929273 on Build GRCh38
rs1007229701 Chr.7: 141929301-141929301 on Build GRCh38
rs1029949334 Chr.7: 141929198-141929198 on Build GRCh38
rs915908801 Chr.7: 141929437-141929437 on Build GRCh38
rs953082574 Chr.7: 141929281-141929281 on Build GRCh38
rs1021375527 Chr.7: 141929302-141929302 on Build GRCh38
rs566727825 Chr.7: 141929566-141929566 on Build GRCh38
rs139844008 Chr.7: 141929169-141929169 on Build GRCh38
rs371843436 Chr.7: 141929343-141929343 on Build GRCh38
rs573318123 Chr.7: 141929403-141929403 on Build GRCh38
rs782578284 Chr.7: 141929567-141929567 on Build GRCh38
rs909986920 Chr.7: 141929516-141929516 on Build GRCh38
rs78522560 Chr.7: 141929141-141929141 on Build GRCh38
rs977292947 Chr.7: 141929349-141929349 on Build GRCh38
rs782553757 Chr.7: 141929139-141929139 on Build GRCh38
rs901439999 Chr.7: 141929165-141929165 on Build GRCh38
rs540239524 Chr.7: 141929450-141929450 on Build GRCh38
rs948710734 Chr.7: 141929467-141929467 on Build GRCh38
rs10267314 Chr.7: 141929500-141929500 on Build GRCh38
rs371734368 Chr.7: 141929133-141929133 on Build GRCh38
rs959967939 Chr.7: 141929410-141929410 on Build GRCh38
rs1007195468 Chr.7: 141929573-141929573 on Build GRCh38
rs888743428 Chr.7: 141929568-141929568 on Build GRCh38
rs940156497 Chr.7: 141929521-141929521 on Build GRCh38
rs924467678 Chr.7: 141929405-141929405 on Build GRCh38
rs1048469638 Chr.7: 141929510-141929510 on Build GRCh38
rs1037190110 Chr.7: 141929522-141929522 on Build GRCh38
rs1037666541 Chr.7: 141929148-141929148 on Build GRCh38
rs764463810 Chr.7: 141929432-141929432 on Build GRCh38
rs561239137 Chr.7: 141929175-141929175 on Build GRCh38
rs901376620 Chr.7: 141929524-141929524 on Build GRCh38

View Full Product Details