Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 141931627-141932168 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLEC5A
Entrez Gene ID
23601
Gene Name
C-type lectin domain family 5 member A
Gene Aliases
CLECSF5, MDL-1, MDL1
UniGene
Hs.446235
Gene Chromosome Location
Chr.7: 141927356-141947009 on Build GRCh38
Transcripts
NM_001301167.1 NM_013252.2 XM_011515995.2 XM_017011915.1 XM_017011916.1 XM_017011917.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3698225 c.403C>T p.R135C Chr.7: 141931769-141931769 on Build GRCh38
4892697 c.352C>T p.L118F Chr.7: 141931820-141931820 on Build GRCh38
3832087 c.371C>T p.A124V Chr.7: 141931801-141931801 on Build GRCh38
3634369 c.429C>T p.I143I Chr.7: 141931743-141931743 on Build GRCh38
6222516 c.421C>T p.R141C Chr.7: 141931751-141931751 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374427602 Chr.7: 141931886-141931886 on Build GRCh38
rs201368100 Chr.7: 141931651-141931651 on Build GRCh38
rs1025761712 Chr.7: 141931927-141931927 on Build GRCh38
rs35942193 Chr.7: 141931750-141931750 on Build GRCh38
rs782346936 Chr.7: 141931813-141931813 on Build GRCh38
rs975698263 Chr.7: 141931951-141931951 on Build GRCh38
rs141690341 Chr.7: 141932039-141932039 on Build GRCh38
rs782807070 Chr.7: 141931755-141931755 on Build GRCh38
rs782012621 Chr.7: 141931842-141931842 on Build GRCh38
rs782811200 Chr.7: 141931841-141931841 on Build GRCh38
rs73525937 Chr.7: 141931707-141931707 on Build GRCh38
rs781867395 Chr.7: 141931756-141931756 on Build GRCh38
rs949440725 Chr.7: 141932066-141932066 on Build GRCh38
rs566356201 Chr.7: 141931751-141931751 on Build GRCh38
rs782127214 Chr.7: 141931844-141931844 on Build GRCh38
rs925236071 Chr.7: 141931974-141931974 on Build GRCh38
rs782720629 Chr.7: 141931978-141931978 on Build GRCh38
rs782759856 Chr.7: 141931729-141931729 on Build GRCh38
rs41275006 Chr.7: 141931910-141931910 on Build GRCh38
rs781992335 Chr.7: 141931797-141931797 on Build GRCh38
rs560818346 Chr.7: 141931762-141931762 on Build GRCh38
rs73525940 Chr.7: 141931848-141931848 on Build GRCh38
rs201568466 Chr.7: 141931844-141931845 on Build GRCh38
rs151000385 Chr.7: 141931768-141931768 on Build GRCh38
rs78381987 Chr.7: 141931716-141931716 on Build GRCh38
rs112606557 Chr.7: 141931829-141931829 on Build GRCh38
rs782088076 Chr.7: 141931745-141931745 on Build GRCh38
rs782717206 Chr.7: 141931749-141931749 on Build GRCh38
rs781948107 Chr.7: 141931712-141931712 on Build GRCh38
rs1046524380 Chr.7: 141932124-141932124 on Build GRCh38
rs564782128 Chr.7: 141931702-141931702 on Build GRCh38
rs782392773 Chr.7: 141931786-141931786 on Build GRCh38
rs974191404 Chr.7: 141931693-141931693 on Build GRCh38
rs12673916 Chr.7: 141931770-141931770 on Build GRCh38
rs147061815 Chr.7: 141932042-141932042 on Build GRCh38
rs781966787 Chr.7: 141931835-141931835 on Build GRCh38
rs573500737 Chr.7: 141931726-141931726 on Build GRCh38
rs781995322 Chr.7: 141931834-141931834 on Build GRCh38
rs138215469 Chr.7: 141931812-141931812 on Build GRCh38
rs373132565 Chr.7: 141931691-141931691 on Build GRCh38
rs782716381 Chr.7: 141931840-141931840 on Build GRCh38
rs782465892 Chr.7: 141931663-141931663 on Build GRCh38
rs184165682 Chr.7: 141931926-141931926 on Build GRCh38
rs782441523 Chr.7: 141931715-141931715 on Build GRCh38
rs527950204 Chr.7: 141932133-141932133 on Build GRCh38
rs782184547 Chr.7: 141931714-141931714 on Build GRCh38
rs368177582 Chr.7: 141932008-141932008 on Build GRCh38
rs782813264 Chr.7: 141931650-141931650 on Build GRCh38
rs200934951 Chr.7: 141931782-141931782 on Build GRCh38
rs782211501 Chr.7: 141931692-141931692 on Build GRCh38
rs782709275 Chr.7: 141931867-141931867 on Build GRCh38
rs753505917 Chr.7: 141931850-141931850 on Build GRCh38
rs913941467 Chr.7: 141932063-141932063 on Build GRCh38
rs576650864 Chr.7: 141931928-141931928 on Build GRCh38
rs782355724 Chr.7: 141931708-141931708 on Build GRCh38
rs139154114 Chr.7: 141931930-141931930 on Build GRCh38
rs781827617 Chr.7: 141931661-141931661 on Build GRCh38
rs535824964 Chr.7: 141931845-141931845 on Build GRCh38
rs973778857 Chr.7: 141931921-141931921 on Build GRCh38
rs12673939 Chr.7: 141931946-141931946 on Build GRCh38
rs782508050 Chr.7: 141931652-141931652 on Build GRCh38
rs986583304 Chr.7: 141931746-141931746 on Build GRCh38
rs912389268 Chr.7: 141931943-141931943 on Build GRCh38
rs140946328 Chr.7: 141931769-141931769 on Build GRCh38
rs782509233 Chr.7: 141931757-141931757 on Build GRCh38
rs782400649 Chr.7: 141931705-141931705 on Build GRCh38
rs112316966 Chr.7: 141931695-141931695 on Build GRCh38
rs1285937 Chr.7: 141932013-141932013 on Build GRCh38
rs1002422258 Chr.7: 141931883-141931883 on Build GRCh38
rs201042036 Chr.7: 141931834-141931835 on Build GRCh38
rs1034422024 Chr.7: 141931680-141931680 on Build GRCh38
rs375407444 Chr.7: 141931659-141931659 on Build GRCh38
rs139367147 Chr.7: 141931737-141931737 on Build GRCh38

View Full Product Details