Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 143397456-143397955 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPHA1
Entrez Gene ID
2041
Gene Name
EPH receptor A1
Gene Aliases
EPH, EPHT, EPHT1
UniGene
Hs.89839
Gene Chromosome Location
Chr.7: 143390813-143408892 on Build GRCh38
Transcripts
NM_005232.4 XM_006715880.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1661675 c.1672G>A p.G558S Chr.7: 143397601-143397601 on Build GRCh38
1548900 c.1693A>C p.I565L Chr.7: 143397580-143397580 on Build GRCh38
3635254 c.1610C>T p.P537L Chr.7: 143397925-143397925 on Build GRCh38
3878959 c.1706G>A p.R569Q Chr.7: 143397567-143397567 on Build GRCh38
364760 c.1651G>T p.V551F Chr.7: 143397622-143397622 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs905061964 Chr.7: 143397793-143397793 on Build GRCh38
rs776663355 Chr.7: 143397659-143397659 on Build GRCh38
rs185706866 Chr.7: 143397545-143397545 on Build GRCh38
rs564687025 Chr.7: 143397490-143397490 on Build GRCh38
rs879691431 Chr.7: 143397806-143397806 on Build GRCh38
rs754713753 Chr.7: 143397933-143397933 on Build GRCh38
rs1046462373 Chr.7: 143397714-143397714 on Build GRCh38
rs111778416 Chr.7: 143397600-143397600 on Build GRCh38
rs766683764 Chr.7: 143397540-143397540 on Build GRCh38
rs752138390 Chr.7: 143397690-143397690 on Build GRCh38
rs925917171 Chr.7: 143397715-143397715 on Build GRCh38
rs45497499 Chr.7: 143397542-143397542 on Build GRCh38
rs775980315 Chr.7: 143397677-143397677 on Build GRCh38
rs555414886 Chr.7: 143397574-143397574 on Build GRCh38
rs781324039 Chr.7: 143397883-143397883 on Build GRCh38
rs201508170 Chr.7: 143397541-143397541 on Build GRCh38
rs748133352 Chr.7: 143397872-143397872 on Build GRCh38
rs1016338802 Chr.7: 143397569-143397569 on Build GRCh38
rs142999380 Chr.7: 143397566-143397566 on Build GRCh38
rs774390706 Chr.7: 143397648-143397648 on Build GRCh38
rs759805828 Chr.7: 143397663-143397663 on Build GRCh38
rs532718317 Chr.7: 143397900-143397900 on Build GRCh38
rs760467922 Chr.7: 143397538-143397539 on Build GRCh38
rs762216093 Chr.7: 143397809-143397809 on Build GRCh38
rs779732304 Chr.7: 143397701-143397701 on Build GRCh38
rs773268230 Chr.7: 143397560-143397560 on Build GRCh38
rs776282653 Chr.7: 143397513-143397513 on Build GRCh38
rs768560395 Chr.7: 143397647-143397647 on Build GRCh38
rs910648566 Chr.7: 143397535-143397535 on Build GRCh38
rs202068423 Chr.7: 143397538-143397538 on Build GRCh38
rs201716947 Chr.7: 143397930-143397930 on Build GRCh38
rs776947009 Chr.7: 143397567-143397567 on Build GRCh38
rs373850526 Chr.7: 143397526-143397526 on Build GRCh38
rs754801581 Chr.7: 143397704-143397704 on Build GRCh38
rs774012786 Chr.7: 143397538-143397538 on Build GRCh38
rs200563282 Chr.7: 143397537-143397538 on Build GRCh38
rs200212760 Chr.7: 143397568-143397568 on Build GRCh38
rs562294624 Chr.7: 143397790-143397790 on Build GRCh38
rs367732092 Chr.7: 143397700-143397700 on Build GRCh38
rs780668174 Chr.7: 143397615-143397615 on Build GRCh38
rs374126260 Chr.7: 143397885-143397885 on Build GRCh38
rs764663016 Chr.7: 143397685-143397685 on Build GRCh38
rs770846462 Chr.7: 143397881-143397881 on Build GRCh38
rs372030742 Chr.7: 143397542-143397542 on Build GRCh38
rs778778160 Chr.7: 143397934-143397934 on Build GRCh38
rs199646340 Chr.7: 143397898-143397898 on Build GRCh38
rs138347396 Chr.7: 143397623-143397623 on Build GRCh38
rs767968483 Chr.7: 143397914-143397914 on Build GRCh38
rs113338478 Chr.7: 143397773-143397773 on Build GRCh38
rs747022455 Chr.7: 143397649-143397649 on Build GRCh38
rs576721214 Chr.7: 143397774-143397774 on Build GRCh38
rs765617275 Chr.7: 143397671-143397671 on Build GRCh38
rs1005386856 Chr.7: 143397830-143397830 on Build GRCh38
rs769337411 Chr.7: 143397582-143397599 on Build GRCh38
rs368300129 Chr.7: 143397897-143397897 on Build GRCh38
rs770115977 Chr.7: 143397888-143397888 on Build GRCh38
rs763458946 Chr.7: 143397678-143397678 on Build GRCh38
rs765726381 Chr.7: 143397564-143397564 on Build GRCh38
rs780762492 Chr.7: 143397539-143397540 on Build GRCh38
rs765978732 Chr.7: 143397928-143397928 on Build GRCh38
rs777423438 Chr.7: 143397543-143397543 on Build GRCh38
rs370950145 Chr.7: 143397519-143397519 on Build GRCh38
rs775349109 Chr.7: 143397546-143397546 on Build GRCh38
rs776242488 Chr.7: 143397760-143397760 on Build GRCh38
rs112594963 Chr.7: 143397708-143397708 on Build GRCh38
rs774662756 Chr.7: 143397909-143397909 on Build GRCh38
rs904925689 Chr.7: 143397548-143397548 on Build GRCh38
rs543988201 Chr.7: 143397779-143397779 on Build GRCh38
rs775445317 Chr.7: 143397528-143397528 on Build GRCh38
rs766091682 Chr.7: 143397730-143397730 on Build GRCh38
rs771069813 Chr.7: 143397655-143397655 on Build GRCh38
rs756672063 Chr.7: 143397696-143397696 on Build GRCh38
rs759566707 Chr.7: 143397816-143397816 on Build GRCh38
rs148751554 Chr.7: 143397921-143397921 on Build GRCh38
rs779820434 Chr.7: 143397624-143397624 on Build GRCh38
rs144350036 Chr.7: 143397839-143397839 on Build GRCh38
rs987993560 Chr.7: 143397752-143397752 on Build GRCh38
rs761841029 Chr.7: 143397531-143397531 on Build GRCh38
rs868237069 Chr.7: 143397581-143397581 on Build GRCh38
rs1055061418 Chr.7: 143397794-143397794 on Build GRCh38
rs898806841 Chr.7: 143397865-143397865 on Build GRCh38
rs936860521 Chr.7: 143397632-143397632 on Build GRCh38
rs79446026 Chr.7: 143397544-143397544 on Build GRCh38
rs542834261 Chr.7: 143397711-143397711 on Build GRCh38
rs750578123 Chr.7: 143397539-143397539 on Build GRCh38
rs749142175 Chr.7: 143397644-143397644 on Build GRCh38
rs181565708 Chr.7: 143397504-143397504 on Build GRCh38
rs535509887 Chr.7: 143397622-143397622 on Build GRCh38
rs760220358 Chr.7: 143397922-143397922 on Build GRCh38
rs754480002 Chr.7: 143397857-143397857 on Build GRCh38
rs551069772 Chr.7: 143397912-143397912 on Build GRCh38
rs758966923 Chr.7: 143397575-143397575 on Build GRCh38
rs993182669 Chr.7: 143397877-143397877 on Build GRCh38
rs45453497 Chr.7: 143397722-143397722 on Build GRCh38
rs937716142 Chr.7: 143397821-143397821 on Build GRCh38
rs954869349 Chr.7: 143397494-143397494 on Build GRCh38
rs754945454 Chr.7: 143397526-143397526 on Build GRCh38
rs760861491 Chr.7: 143397563-143397563 on Build GRCh38
rs978908441 Chr.7: 143397712-143397712 on Build GRCh38
rs763454805 Chr.7: 143397702-143397702 on Build GRCh38
rs116896514 Chr.7: 143397846-143397846 on Build GRCh38
rs896939396 Chr.7: 143397845-143397845 on Build GRCh38

View Full Product Details