Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 39610252-39610755 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAM18
Entrez Gene ID
8749
Gene Name
ADAM metallopeptidase domain 18
Gene Aliases
ADAM27, tMDCIII
UniGene
Hs.127930
Gene Chromosome Location
Chr.8: 39584568-39730064 on Build GRCh38
Transcripts
NM_001190956.1 NM_001320313.1 NM_014237.2 NR_135201.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3699084 c.452C>A p.S151Y Chr.8: 39610636-39610636 on Build GRCh38
308869 c.493C>A p.Q165K Chr.8: 39610677-39610677 on Build GRCh38
3649013 c.509C>T p.P170L Chr.8: 39610693-39610693 on Build GRCh38
7262127 c.459A>C p.L153F Chr.8: 39610643-39610643 on Build GRCh38
5509071 c.516C>A p.N172K Chr.8: 39610700-39610700 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs919480438 Chr.8: 39610346-39610346 on Build GRCh38
rs376919508 Chr.8: 39610727-39610727 on Build GRCh38
rs768452253 Chr.8: 39610473-39610473 on Build GRCh38
rs1052762054 Chr.8: 39610283-39610283 on Build GRCh38
rs777830988 Chr.8: 39610730-39610730 on Build GRCh38
rs148690264 Chr.8: 39610633-39610633 on Build GRCh38
rs929487906 Chr.8: 39610656-39610656 on Build GRCh38
rs367919364 Chr.8: 39610566-39610566 on Build GRCh38
rs996597862 Chr.8: 39610381-39610381 on Build GRCh38
rs780206354 Chr.8: 39610657-39610657 on Build GRCh38
rs969508898 Chr.8: 39610729-39610729 on Build GRCh38
rs142986500 Chr.8: 39610622-39610622 on Build GRCh38
rs200924990 Chr.8: 39610378-39610378 on Build GRCh38
rs992252905 Chr.8: 39610440-39610440 on Build GRCh38
rs761055219 Chr.8: 39610693-39610693 on Build GRCh38
rs899542596 Chr.8: 39610371-39610371 on Build GRCh38
rs747999507 Chr.8: 39610664-39610664 on Build GRCh38
rs936713316 Chr.8: 39610280-39610280 on Build GRCh38
rs779347108 Chr.8: 39610648-39610648 on Build GRCh38
rs750624408 Chr.8: 39610713-39610713 on Build GRCh38
rs1049364395 Chr.8: 39610377-39610377 on Build GRCh38
rs779970792 Chr.8: 39610724-39610724 on Build GRCh38
rs561528576 Chr.8: 39610452-39610452 on Build GRCh38
rs1003907480 Chr.8: 39610417-39610417 on Build GRCh38
rs757629778 Chr.8: 39610711-39610711 on Build GRCh38
rs1003751135 Chr.8: 39610412-39610412 on Build GRCh38
rs201725440 Chr.8: 39610598-39610598 on Build GRCh38
rs139114737 Chr.8: 39610551-39610551 on Build GRCh38
rs749224382 Chr.8: 39610731-39610731 on Build GRCh38
rs918088460 Chr.8: 39610449-39610449 on Build GRCh38
rs770563977 Chr.8: 39610680-39610680 on Build GRCh38
rs773923013 Chr.8: 39610681-39610681 on Build GRCh38
rs769431916 Chr.8: 39610597-39610597 on Build GRCh38
rs759110523 Chr.8: 39610630-39610630 on Build GRCh38
rs369574720 Chr.8: 39610479-39610479 on Build GRCh38
rs967717832 Chr.8: 39610318-39610318 on Build GRCh38
rs369509127 Chr.8: 39610599-39610599 on Build GRCh38
rs147248851 Chr.8: 39610554-39610554 on Build GRCh38
rs1044870332 Chr.8: 39610385-39610385 on Build GRCh38
rs549488812 Chr.8: 39610588-39610588 on Build GRCh38
rs776516988 Chr.8: 39610549-39610549 on Build GRCh38
rs962283561 Chr.8: 39610430-39610430 on Build GRCh38
rs375908521 Chr.8: 39610647-39610647 on Build GRCh38
rs758728874 Chr.8: 39610716-39610716 on Build GRCh38
rs768711287 Chr.8: 39610559-39610559 on Build GRCh38
rs575240108 Chr.8: 39610587-39610587 on Build GRCh38
rs766072613 Chr.8: 39610621-39610621 on Build GRCh38
rs751113624 Chr.8: 39610490-39610490 on Build GRCh38
rs769814741 Chr.8: 39610670-39610670 on Build GRCh38
rs752297916 Chr.8: 39610384-39610384 on Build GRCh38
rs778208283 Chr.8: 39610527-39610527 on Build GRCh38
rs574473254 Chr.8: 39610356-39610356 on Build GRCh38
rs976286138 Chr.8: 39610342-39610342 on Build GRCh38
rs887764272 Chr.8: 39610377-39610378 on Build GRCh38
rs952471124 Chr.8: 39610390-39610390 on Build GRCh38
rs756666331 Chr.8: 39610635-39610635 on Build GRCh38
rs771309353 Chr.8: 39610541-39610541 on Build GRCh38
rs1034826645 Chr.8: 39610420-39610420 on Build GRCh38
rs370884759 Chr.8: 39610686-39610686 on Build GRCh38
rs1015569491 Chr.8: 39610423-39610423 on Build GRCh38
rs543254937 Chr.8: 39610385-39610385 on Build GRCh38
rs143967659 Chr.8: 39610685-39610685 on Build GRCh38
rs756363697 Chr.8: 39610522-39610522 on Build GRCh38
rs753098867 Chr.8: 39610507-39610507 on Build GRCh38
rs932531267 Chr.8: 39610658-39610658 on Build GRCh38
rs929562916 Chr.8: 39610366-39610366 on Build GRCh38
rs369562039 Chr.8: 39610602-39610602 on Build GRCh38
rs749421107 Chr.8: 39610531-39610531 on Build GRCh38
rs761823550 Chr.8: 39610593-39610593 on Build GRCh38
rs775095784 Chr.8: 39610689-39610689 on Build GRCh38
rs576732402 Chr.8: 39610478-39610478 on Build GRCh38
rs986277076 Chr.8: 39610330-39610330 on Build GRCh38
rs151185191 Chr.8: 39610651-39610651 on Build GRCh38
rs201110239 Chr.8: 39610448-39610448 on Build GRCh38
rs267601916 Chr.8: 39610692-39610692 on Build GRCh38
rs1008033832 Chr.8: 39610341-39610341 on Build GRCh38
rs73605945 Chr.8: 39610609-39610609 on Build GRCh38
rs372919027 Chr.8: 39610712-39610712 on Build GRCh38
rs755458692 Chr.8: 39610493-39610493 on Build GRCh38
rs754252814 Chr.8: 39610709-39610709 on Build GRCh38
rs202232780 Chr.8: 39610496-39610496 on Build GRCh38
rs764709336 Chr.8: 39610703-39610703 on Build GRCh38
rs749142259 Chr.8: 39610678-39610678 on Build GRCh38
rs772863227 Chr.8: 39610677-39610677 on Build GRCh38
rs181566641 Chr.8: 39610340-39610340 on Build GRCh38
rs142169302 Chr.8: 39610636-39610636 on Build GRCh38
rs147230372 Chr.8: 39610632-39610632 on Build GRCh38
rs925413610 Chr.8: 39610502-39610502 on Build GRCh38
rs760347966 Chr.8: 39610471-39610471 on Build GRCh38
rs758494889 Chr.8: 39610654-39610654 on Build GRCh38

View Full Product Details