Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 39637393-39637943 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAM18
Entrez Gene ID
8749
Gene Name
ADAM metallopeptidase domain 18
Gene Aliases
ADAM27, tMDCIII
UniGene
Hs.127930
Gene Chromosome Location
Chr.8: 39584568-39730064 on Build GRCh38
Transcripts
NM_001320313.1 NM_014237.2 NR_135201.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3649017 c.731C>T p.S244F Chr.8: 39637607-39637607 on Build GRCh38
1723841 c.751G>A p.D251N Chr.8: 39637627-39637627 on Build GRCh38
6516110 c.669C>A p.T223T Chr.8: 39637545-39637545 on Build GRCh38
3649018 c.732C>T p.S244S Chr.8: 39637608-39637608 on Build GRCh38
3649019 c.741G>A p.G247G Chr.8: 39637617-39637617 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759564897 Chr.8: 39637658-39637658 on Build GRCh38
rs1010854900 Chr.8: 39637872-39637872 on Build GRCh38
rs141094631 Chr.8: 39637657-39637657 on Build GRCh38
rs544054266 Chr.8: 39637443-39637443 on Build GRCh38
rs773453427 Chr.8: 39637742-39637742 on Build GRCh38
rs538445152 Chr.8: 39637777-39637777 on Build GRCh38
rs191138498 Chr.8: 39637494-39637494 on Build GRCh38
rs778498150 Chr.8: 39637686-39637686 on Build GRCh38
rs771884090 Chr.8: 39637565-39637565 on Build GRCh38
rs538327994 Chr.8: 39637873-39637873 on Build GRCh38
rs760378572 Chr.8: 39637676-39637676 on Build GRCh38
rs768922590 Chr.8: 39637710-39637710 on Build GRCh38
rs766288632 Chr.8: 39637510-39637510 on Build GRCh38
rs775409625 Chr.8: 39637755-39637755 on Build GRCh38
rs962513394 Chr.8: 39637786-39637786 on Build GRCh38
rs763710692 Chr.8: 39637680-39637680 on Build GRCh38
rs780150173 Chr.8: 39637516-39637520 on Build GRCh38
rs143506696 Chr.8: 39637856-39637856 on Build GRCh38
rs962175607 Chr.8: 39637682-39637682 on Build GRCh38
rs999457374 Chr.8: 39637629-39637629 on Build GRCh38
rs865959777 Chr.8: 39637627-39637627 on Build GRCh38
rs375099212 Chr.8: 39637664-39637664 on Build GRCh38
rs928120961 Chr.8: 39637912-39637912 on Build GRCh38
rs199835295 Chr.8: 39637539-39637539 on Build GRCh38
rs369670957 Chr.8: 39637674-39637674 on Build GRCh38
rs535022743 Chr.8: 39637844-39637844 on Build GRCh38
rs772182455 Chr.8: 39637748-39637748 on Build GRCh38
rs1024749886 Chr.8: 39637462-39637465 on Build GRCh38
rs751369846 Chr.8: 39637517-39637517 on Build GRCh38
rs541966827 Chr.8: 39637689-39637689 on Build GRCh38
rs753383322 Chr.8: 39637681-39637681 on Build GRCh38
rs761869049 Chr.8: 39637648-39637648 on Build GRCh38
rs866192798 Chr.8: 39637712-39637712 on Build GRCh38
rs756888023 Chr.8: 39637448-39637448 on Build GRCh38
rs973748696 Chr.8: 39637478-39637478 on Build GRCh38
rs377707835 Chr.8: 39637740-39637740 on Build GRCh38
rs3824144 Chr.8: 39637806-39637806 on Build GRCh38
rs10112294 Chr.8: 39637555-39637555 on Build GRCh38
rs375864755 Chr.8: 39637561-39637561 on Build GRCh38
rs577489796 Chr.8: 39637482-39637482 on Build GRCh38
rs367547346 Chr.8: 39637520-39637520 on Build GRCh38
rs201445727 Chr.8: 39637693-39637693 on Build GRCh38
rs781208386 Chr.8: 39637722-39637722 on Build GRCh38
rs369119694 Chr.8: 39637696-39637696 on Build GRCh38
rs1017903859 Chr.8: 39637465-39637465 on Build GRCh38
rs371887913 Chr.8: 39637497-39637497 on Build GRCh38
rs920825106 Chr.8: 39637481-39637481 on Build GRCh38
rs1029531928 Chr.8: 39637436-39637436 on Build GRCh38
rs768810673 Chr.8: 39637617-39637617 on Build GRCh38
rs778580768 Chr.8: 39637519-39637519 on Build GRCh38
rs983673177 Chr.8: 39637908-39637908 on Build GRCh38
rs552094015 Chr.8: 39637778-39637778 on Build GRCh38
rs747201163 Chr.8: 39637614-39637614 on Build GRCh38
rs267601917 Chr.8: 39637559-39637559 on Build GRCh38
rs920806043 Chr.8: 39637703-39637703 on Build GRCh38
rs759511494 Chr.8: 39637747-39637747 on Build GRCh38
rs776762461 Chr.8: 39637642-39637642 on Build GRCh38
rs780374847 Chr.8: 39637701-39637701 on Build GRCh38
rs529706001 Chr.8: 39637724-39637724 on Build GRCh38
rs552662865 Chr.8: 39637466-39637466 on Build GRCh38
rs946698650 Chr.8: 39637602-39637602 on Build GRCh38
rs962683907 Chr.8: 39637468-39637468 on Build GRCh38
rs772364556 Chr.8: 39637604-39637604 on Build GRCh38
rs972265993 Chr.8: 39637692-39637692 on Build GRCh38
rs34064543 Chr.8: 39637543-39637543 on Build GRCh38
rs752576579 Chr.8: 39637675-39637675 on Build GRCh38
rs753394104 Chr.8: 39637441-39637441 on Build GRCh38
rs369932234 Chr.8: 39637595-39637595 on Build GRCh38
rs950942797 Chr.8: 39637861-39637861 on Build GRCh38
rs752345891 Chr.8: 39637537-39637537 on Build GRCh38
rs1006758787 Chr.8: 39637715-39637715 on Build GRCh38
rs747432229 Chr.8: 39637702-39637702 on Build GRCh38
rs903287547 Chr.8: 39637792-39637792 on Build GRCh38
rs564249507 Chr.8: 39637568-39637568 on Build GRCh38
rs780429788 Chr.8: 39637612-39637612 on Build GRCh38
rs767437563 Chr.8: 39637522-39637522 on Build GRCh38
rs781666226 Chr.8: 39637690-39637690 on Build GRCh38
rs569543406 Chr.8: 39637754-39637754 on Build GRCh38
rs915940296 Chr.8: 39637775-39637775 on Build GRCh38

View Full Product Details