Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 41654272-41654775 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANK1
Entrez Gene ID
286
Gene Name
ankyrin 1
Gene Aliases
ANK, SPH1, SPH2
UniGene
Hs.654438
Gene Chromosome Location
Chr.8: 41653225-41896762 on Build GRCh38
Transcripts
NM_000037.3 NM_001142445.1 NM_001142446.1 NM_020475.2 NM_020476.2 NM_020477.2 NM_020478.4 NM_020480.4 XM_005273476.4 XM_011544490.2 XM_011544491.2 XM_011544494.2 XM_011544495.2 XM_011544496.2 XM_011544500.2 XM_011544501.2 XM_011544502.2 XM_011544503.2 XM_011544504.2 XM_011544505.2 XM_017013319.1 XM_017013320.1 XM_017013321.1 XM_017013322.1 XM_017013323.1 XM_017013324.1 XM_017013325.1 XM_017013326.1 XM_017013327.1 XM_017013328.1 XM_017013329.1

SNP Detail

SNP ID Chromosome Location
rs527279141 Chr.8: 41654565-41654565 on Build GRCh38
rs908741238 Chr.8: 41654725-41654725 on Build GRCh38
rs1003032028 Chr.8: 41654504-41654504 on Build GRCh38
rs542907914 Chr.8: 41654327-41654327 on Build GRCh38
rs1050071864 Chr.8: 41654358-41654358 on Build GRCh38
rs956602087 Chr.8: 41654526-41654526 on Build GRCh38
rs934162069 Chr.8: 41654365-41654365 on Build GRCh38
rs753724860 Chr.8: 41654320-41654320 on Build GRCh38
rs901344844 Chr.8: 41654390-41654390 on Build GRCh38
rs974321679 Chr.8: 41654746-41654746 on Build GRCh38
rs942946556 Chr.8: 41654380-41654380 on Build GRCh38
rs985849931 Chr.8: 41654571-41654572 on Build GRCh38
rs987022893 Chr.8: 41654378-41654378 on Build GRCh38
rs546957844 Chr.8: 41654594-41654594 on Build GRCh38
rs977041324 Chr.8: 41654670-41654670 on Build GRCh38
rs914969086 Chr.8: 41654323-41654323 on Build GRCh38
rs920140011 Chr.8: 41654749-41654749 on Build GRCh38
rs1047655448 Chr.8: 41654721-41654721 on Build GRCh38
rs567278502 Chr.8: 41654663-41654663 on Build GRCh38
rs116137861 Chr.8: 41654388-41654388 on Build GRCh38
rs551057596 Chr.8: 41654503-41654503 on Build GRCh38
rs769931738 Chr.8: 41654441-41654441 on Build GRCh38
rs955524434 Chr.8: 41654688-41654688 on Build GRCh38
rs1044176046 Chr.8: 41654429-41654429 on Build GRCh38
rs946434661 Chr.8: 41654334-41654334 on Build GRCh38
rs1038620920 Chr.8: 41654391-41654391 on Build GRCh38
rs554562729 Chr.8: 41654405-41654405 on Build GRCh38
rs921309516 Chr.8: 41654347-41654347 on Build GRCh38
rs562500715 Chr.8: 41654382-41654382 on Build GRCh38
rs891609842 Chr.8: 41654528-41654528 on Build GRCh38
rs1023609649 Chr.8: 41654442-41654442 on Build GRCh38
rs968024672 Chr.8: 41654573-41654573 on Build GRCh38
rs1047176428 Chr.8: 41654395-41654395 on Build GRCh38
rs886062934 Chr.8: 41654728-41654728 on Build GRCh38
rs556825982 Chr.8: 41654743-41654743 on Build GRCh38
rs536499240 Chr.8: 41654735-41654735 on Build GRCh38
rs905171905 Chr.8: 41654497-41654497 on Build GRCh38
rs941452720 Chr.8: 41654748-41654748 on Build GRCh38
rs112189561 Chr.8: 41654521-41654521 on Build GRCh38
rs922836487 Chr.8: 41654673-41654673 on Build GRCh38
rs1011505496 Chr.8: 41654532-41654532 on Build GRCh38
rs886062933 Chr.8: 41654716-41654716 on Build GRCh38
rs773270510 Chr.8: 41654700-41654700 on Build GRCh38
rs975352104 Chr.8: 41654577-41654577 on Build GRCh38
rs886062932 Chr.8: 41654548-41654548 on Build GRCh38
rs944315677 Chr.8: 41654642-41654642 on Build GRCh38
rs898180614 Chr.8: 41654402-41654402 on Build GRCh38
rs1028274507 Chr.8: 41654607-41654607 on Build GRCh38

View Full Product Details