Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 43168998-43169535 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HGSNAT
Entrez Gene ID
138050
Gene Name
heparan-alpha-glucosaminide N-acetyltransferase
Gene Aliases
HGNAT, MPS3C, RP73, TMEM76
UniGene
Hs.600384
Gene Chromosome Location
Chr.8: 43140449-43202827 on Build GRCh38
Transcripts
NM_152419.2 XM_005273409.1 XM_005273410.1 XM_005273411.1 XM_005273412.3 XM_017013088.1 XM_017013089.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7223887 c.648T>A p.S216R Chr.8: 43169173-43169173 on Build GRCh38
307666 c.652G>T p.D218Y Chr.8: 43169177-43169177 on Build GRCh38
4976167 c.686G>C p.S229T Chr.8: 43169211-43169211 on Build GRCh38
2962189 c.704G>A p.R235H Chr.8: 43169229-43169229 on Build GRCh38
1100113 c.648T>C p.S216S Chr.8: 43169173-43169173 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1024182962 Chr.8: 43169104-43169104 on Build GRCh38
rs202173801 Chr.8: 43169058-43169061 on Build GRCh38
rs553018464 Chr.8: 43169466-43169466 on Build GRCh38
rs1038109591 Chr.8: 43169073-43169073 on Build GRCh38
rs1012749298 Chr.8: 43169100-43169100 on Build GRCh38
rs748100708 Chr.8: 43169273-43169273 on Build GRCh38
rs188402900 Chr.8: 43169370-43169370 on Build GRCh38
rs1038077414 Chr.8: 43169455-43169455 on Build GRCh38
rs955830864 Chr.8: 43169447-43169447 on Build GRCh38
rs192297314 Chr.8: 43169229-43169229 on Build GRCh38
rs981366355 Chr.8: 43169414-43169414 on Build GRCh38
rs4272427 Chr.8: 43169482-43169482 on Build GRCh38
rs894260781 Chr.8: 43169080-43169080 on Build GRCh38
rs957369804 Chr.8: 43169336-43169336 on Build GRCh38
rs769542626 Chr.8: 43169288-43169288 on Build GRCh38
rs568730607 Chr.8: 43169301-43169301 on Build GRCh38
rs759673019 Chr.8: 43169440-43169440 on Build GRCh38
rs761748617 Chr.8: 43169493-43169493 on Build GRCh38
rs144188129 Chr.8: 43169262-43169262 on Build GRCh38
rs767944176 Chr.8: 43169135-43169135 on Build GRCh38
rs753221555 Chr.8: 43169146-43169146 on Build GRCh38
rs754185531 Chr.8: 43169207-43169207 on Build GRCh38
rs528466699 Chr.8: 43169243-43169243 on Build GRCh38
rs746972888 Chr.8: 43169062-43169065 on Build GRCh38
rs914361795 Chr.8: 43169469-43169469 on Build GRCh38
rs199861011 Chr.8: 43169228-43169228 on Build GRCh38
rs1023628464 Chr.8: 43169387-43169387 on Build GRCh38
rs976141808 Chr.8: 43169382-43169382 on Build GRCh38
rs1045514026 Chr.8: 43169506-43169506 on Build GRCh38
rs75461309 Chr.8: 43169366-43169366 on Build GRCh38
rs983700240 Chr.8: 43169439-43169439 on Build GRCh38
rs532600953 Chr.8: 43169045-43169045 on Build GRCh38
rs546298041 Chr.8: 43169102-43169102 on Build GRCh38
rs779371714 Chr.8: 43169143-43169143 on Build GRCh38
rs964805469 Chr.8: 43169377-43169377 on Build GRCh38
rs367799225 Chr.8: 43169225-43169225 on Build GRCh38
rs909479985 Chr.8: 43169451-43169451 on Build GRCh38
rs73631580 Chr.8: 43169452-43169452 on Build GRCh38
rs763403091 Chr.8: 43169203-43169203 on Build GRCh38
rs929477640 Chr.8: 43169412-43169412 on Build GRCh38
rs182504686 Chr.8: 43169338-43169338 on Build GRCh38
rs1035953203 Chr.8: 43169432-43169432 on Build GRCh38
rs748560542 Chr.8: 43169165-43169166 on Build GRCh38
rs756486644 Chr.8: 43169161-43169161 on Build GRCh38
rs758473075 Chr.8: 43169234-43169234 on Build GRCh38
rs1005650855 Chr.8: 43169054-43169054 on Build GRCh38
rs774305899 Chr.8: 43169277-43169277 on Build GRCh38
rs923430133 Chr.8: 43169407-43169407 on Build GRCh38
rs946995268 Chr.8: 43169039-43169039 on Build GRCh38
rs192234778 Chr.8: 43169478-43169478 on Build GRCh38

View Full Product Details