Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 43172218-43172728 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HGSNAT
Entrez Gene ID
138050
Gene Name
heparan-alpha-glucosaminide N-acetyltransferase
Gene Aliases
HGNAT, MPS3C, RP73, TMEM76
UniGene
Hs.600384
Gene Chromosome Location
Chr.8: 43140449-43202827 on Build GRCh38
Transcripts
NM_152419.2 XM_005273409.1 XM_005273410.1 XM_005273411.1 XM_005273412.3 XM_017013088.1 XM_017013089.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1100114 c.848T>C p.V283A Chr.8: 43172330-43172330 on Build GRCh38
7264287 c.828-10_828-8delCTT p.? Chr.8: 43172300-43172302 on Build GRCh38
1569042 c.885C>T p.F295F Chr.8: 43172367-43172367 on Build GRCh38
223253 c.865G>A p.G289R Chr.8: 43172347-43172347 on Build GRCh38
5492377 c.890A>G p.H297R Chr.8: 43172372-43172372 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762706228 Chr.8: 43172702-43172702 on Build GRCh38
rs981101102 Chr.8: 43172242-43172242 on Build GRCh38
rs758094310 Chr.8: 43172377-43172377 on Build GRCh38
rs988574197 Chr.8: 43172695-43172695 on Build GRCh38
rs771179693 Chr.8: 43172425-43172425 on Build GRCh38
rs1035369901 Chr.8: 43172603-43172603 on Build GRCh38
rs777334573 Chr.8: 43172305-43172305 on Build GRCh38
rs994670542 Chr.8: 43172543-43172543 on Build GRCh38
rs765032039 Chr.8: 43172368-43172368 on Build GRCh38
rs34496674 Chr.8: 43172471-43172472 on Build GRCh38
rs563079121 Chr.8: 43172451-43172451 on Build GRCh38
rs776162529 Chr.8: 43172300-43172302 on Build GRCh38
rs778768331 Chr.8: 43172279-43172279 on Build GRCh38
rs190447162 Chr.8: 43172434-43172434 on Build GRCh38
rs11998092 Chr.8: 43172509-43172509 on Build GRCh38
rs754600448 Chr.8: 43172394-43172394 on Build GRCh38
rs1034995606 Chr.8: 43172251-43172251 on Build GRCh38
rs545140890 Chr.8: 43172460-43172460 on Build GRCh38
rs780853640 Chr.8: 43172407-43172407 on Build GRCh38
rs202161501 Chr.8: 43172408-43172408 on Build GRCh38
rs779528959 Chr.8: 43172286-43172286 on Build GRCh38
rs145743135 Chr.8: 43172638-43172638 on Build GRCh38
rs866778133 Chr.8: 43172426-43172426 on Build GRCh38
rs905036493 Chr.8: 43172580-43172580 on Build GRCh38
rs567437818 Chr.8: 43172624-43172624 on Build GRCh38
rs766657364 Chr.8: 43172326-43172326 on Build GRCh38
rs757103938 Chr.8: 43172277-43172277 on Build GRCh38
rs554146681 Chr.8: 43172378-43172378 on Build GRCh38
rs773638310 Chr.8: 43172314-43172314 on Build GRCh38
rs531408990 Chr.8: 43172454-43172454 on Build GRCh38
rs186434281 Chr.8: 43172285-43172285 on Build GRCh38
rs374716613 Chr.8: 43172269-43172269 on Build GRCh38
rs143545798 Chr.8: 43172595-43172595 on Build GRCh38
rs768829150 Chr.8: 43172615-43172615 on Build GRCh38
rs773552088 Chr.8: 43172422-43172422 on Build GRCh38
rs753802710 Chr.8: 43172637-43172641 on Build GRCh38
rs376365655 Chr.8: 43172343-43172343 on Build GRCh38
rs371534501 Chr.8: 43172421-43172421 on Build GRCh38
rs753843717 Chr.8: 43172342-43172342 on Build GRCh38
rs527272516 Chr.8: 43172547-43172547 on Build GRCh38
rs772667253 Chr.8: 43172423-43172423 on Build GRCh38
rs774695207 Chr.8: 43172431-43172431 on Build GRCh38
rs1036100461 Chr.8: 43172497-43172497 on Build GRCh38
rs796874154 Chr.8: 43172554-43172554 on Build GRCh38
rs1033944171 Chr.8: 43172629-43172629 on Build GRCh38
rs751701983 Chr.8: 43172328-43172328 on Build GRCh38
rs750259126 Chr.8: 43172373-43172373 on Build GRCh38
rs1030332261 Chr.8: 43172488-43172488 on Build GRCh38
rs767664133 Chr.8: 43172435-43172435 on Build GRCh38
rs1027452249 Chr.8: 43172574-43172574 on Build GRCh38
rs762402992 Chr.8: 43172308-43172308 on Build GRCh38
rs946871556 Chr.8: 43172355-43172355 on Build GRCh38
rs549517998 Chr.8: 43172669-43172669 on Build GRCh38
rs761147174 Chr.8: 43172276-43172277 on Build GRCh38
rs959202881 Chr.8: 43172642-43172642 on Build GRCh38
rs770357315 Chr.8: 43172311-43172311 on Build GRCh38
rs890098520 Chr.8: 43172472-43172472 on Build GRCh38
rs137963729 Chr.8: 43172331-43172331 on Build GRCh38
rs770410584 Chr.8: 43172278-43172279 on Build GRCh38
rs768255849 Chr.8: 43172294-43172294 on Build GRCh38
rs767574122 Chr.8: 43172339-43172339 on Build GRCh38
rs746644852 Chr.8: 43172392-43172392 on Build GRCh38
rs757193957 Chr.8: 43172346-43172346 on Build GRCh38
rs962973696 Chr.8: 43172573-43172573 on Build GRCh38
rs746567359 Chr.8: 43172289-43172289 on Build GRCh38
rs763179425 Chr.8: 43172322-43172322 on Build GRCh38
rs763954864 Chr.8: 43172493-43172493 on Build GRCh38
rs868630625 Chr.8: 43172473-43172473 on Build GRCh38

View Full Product Details