Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 6732483-6732987 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGPAT5
Entrez Gene ID
55326
Gene Name
1-acylglycerol-3-phosphate O-acyltransferase 5
Gene Aliases
1AGPAT5, LPAATE
UniGene
Hs.624002
Gene Chromosome Location
Chr.8: 6708357-6761503 on Build GRCh38
Transcripts
NM_018361.3 XM_011534751.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5612038 c.454A>C p.M152L Chr.8: 6732609-6732609 on Build GRCh38
5925408 c.494C>T p.P165L Chr.8: 6732649-6732649 on Build GRCh38
5436260 c.464A>T p.K155M Chr.8: 6732619-6732619 on Build GRCh38
454743 c.418_421delTATG p.Y140fs*1 Chr.8: 6732573-6732576 on Build GRCh38
5724866 c.458G>A p.R153Q Chr.8: 6732613-6732613 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs139754032 Chr.8: 6732878-6732878 on Build GRCh38
rs201067184 Chr.8: 6732614-6732614 on Build GRCh38
rs534843026 Chr.8: 6732664-6732664 on Build GRCh38
rs761379532 Chr.8: 6732792-6732792 on Build GRCh38
rs752941173 Chr.8: 6732612-6732612 on Build GRCh38
rs374719910 Chr.8: 6732684-6732684 on Build GRCh38
rs772815234 Chr.8: 6732602-6732602 on Build GRCh38
rs558653498 Chr.8: 6732828-6732828 on Build GRCh38
rs1030753494 Chr.8: 6732822-6732822 on Build GRCh38
rs530434069 Chr.8: 6732957-6732957 on Build GRCh38
rs763607894 Chr.8: 6732658-6732658 on Build GRCh38
rs765789423 Chr.8: 6732606-6732606 on Build GRCh38
rs967155177 Chr.8: 6732847-6732847 on Build GRCh38
rs751810630 Chr.8: 6732622-6732622 on Build GRCh38
rs746699136 Chr.8: 6732699-6732699 on Build GRCh38
rs17078008 Chr.8: 6732605-6732605 on Build GRCh38
rs754283162 Chr.8: 6732548-6732548 on Build GRCh38
rs567571297 Chr.8: 6732777-6732777 on Build GRCh38
rs149707176 Chr.8: 6732621-6732621 on Build GRCh38
rs887919045 Chr.8: 6732767-6732767 on Build GRCh38
rs781093363 Chr.8: 6732630-6732630 on Build GRCh38
rs181667589 Chr.8: 6732591-6732591 on Build GRCh38
rs530859671 Chr.8: 6732629-6732629 on Build GRCh38
rs373334231 Chr.8: 6732665-6732665 on Build GRCh38
rs1020422082 Chr.8: 6732842-6732842 on Build GRCh38
rs758217036 Chr.8: 6732670-6732670 on Build GRCh38
rs759575712 Chr.8: 6732518-6732518 on Build GRCh38
rs752006586 Chr.8: 6732662-6732662 on Build GRCh38
rs775096325 Chr.8: 6732583-6732583 on Build GRCh38
rs776124883 Chr.8: 6732750-6732750 on Build GRCh38
rs149350461 Chr.8: 6732632-6732632 on Build GRCh38
rs765873406 Chr.8: 6732647-6732647 on Build GRCh38
rs777878360 Chr.8: 6732788-6732788 on Build GRCh38
rs1049277839 Chr.8: 6732749-6732749 on Build GRCh38
rs1047520122 Chr.8: 6732768-6732768 on Build GRCh38
rs571345904 Chr.8: 6732738-6732738 on Build GRCh38
rs919230266 Chr.8: 6732962-6732962 on Build GRCh38
rs373446573 Chr.8: 6732572-6732572 on Build GRCh38
rs777479219 Chr.8: 6732695-6732695 on Build GRCh38
rs780050923 Chr.8: 6732675-6732675 on Build GRCh38
rs745984507 Chr.8: 6732631-6732631 on Build GRCh38
rs998537352 Chr.8: 6732865-6732865 on Build GRCh38
rs758575939 Chr.8: 6732664-6732665 on Build GRCh38
rs191022960 Chr.8: 6732798-6732798 on Build GRCh38
rs182443550 Chr.8: 6732845-6732845 on Build GRCh38
rs1055451339 Chr.8: 6732764-6732764 on Build GRCh38
rs375650774 Chr.8: 6732638-6732638 on Build GRCh38
rs763756656 Chr.8: 6732540-6732540 on Build GRCh38
rs201289468 Chr.8: 6732544-6732544 on Build GRCh38
rs1032819609 Chr.8: 6732868-6732868 on Build GRCh38
rs572993833 Chr.8: 6732824-6732824 on Build GRCh38
rs745763532 Chr.8: 6732577-6732577 on Build GRCh38
rs553140956 Chr.8: 6732814-6732814 on Build GRCh38
rs751012520 Chr.8: 6732543-6732543 on Build GRCh38
rs981471771 Chr.8: 6732941-6732941 on Build GRCh38
rs779056908 Chr.8: 6732613-6732613 on Build GRCh38
rs764526076 Chr.8: 6732659-6732659 on Build GRCh38
rs753753446 Chr.8: 6732681-6732681 on Build GRCh38
rs913683150 Chr.8: 6732568-6732568 on Build GRCh38
rs551550888 Chr.8: 6732737-6732737 on Build GRCh38
rs200273834 Chr.8: 6732633-6732633 on Build GRCh38
rs917875658 Chr.8: 6732765-6732765 on Build GRCh38
rs746670742 Chr.8: 6732648-6732648 on Build GRCh38
rs768422781 Chr.8: 6732635-6732635 on Build GRCh38
rs937685540 Chr.8: 6732761-6732761 on Build GRCh38
rs753129849 Chr.8: 6732553-6732553 on Build GRCh38
rs532731784 Chr.8: 6732691-6732691 on Build GRCh38
rs561081835 Chr.8: 6732886-6732886 on Build GRCh38
rs372712764 Chr.8: 6732532-6732532 on Build GRCh38
rs781383212 Chr.8: 6732575-6732575 on Build GRCh38
rs547176488 Chr.8: 6732640-6732640 on Build GRCh38
rs769454660 Chr.8: 6732522-6732522 on Build GRCh38
rs544800403 Chr.8: 6732827-6732827 on Build GRCh38
rs566913444 Chr.8: 6732649-6732649 on Build GRCh38
rs971836154 Chr.8: 6732927-6732927 on Build GRCh38
rs552862246 Chr.8: 6732554-6732554 on Build GRCh38
rs917577251 Chr.8: 6732942-6732942 on Build GRCh38
rs945125513 Chr.8: 6732574-6732574 on Build GRCh38
rs752926708 Chr.8: 6732662-6732663 on Build GRCh38
rs756210228 Chr.8: 6732668-6732668 on Build GRCh38
rs755555291 Chr.8: 6732551-6732551 on Build GRCh38
rs900506644 Chr.8: 6732804-6732804 on Build GRCh38
rs745730097 Chr.8: 6732616-6732616 on Build GRCh38
rs767077316 Chr.8: 6732545-6732545 on Build GRCh38
rs754947813 Chr.8: 6732683-6732683 on Build GRCh38
rs762471694 Chr.8: 6732536-6732536 on Build GRCh38
rs147468529 Chr.8: 6732740-6732740 on Build GRCh38
rs996372891 Chr.8: 6732697-6732697 on Build GRCh38
rs186524928 Chr.8: 6732770-6732770 on Build GRCh38
rs769802831 Chr.8: 6732582-6732582 on Build GRCh38
rs950059659 Chr.8: 6732736-6732736 on Build GRCh38
rs139524305 Chr.8: 6732639-6732639 on Build GRCh38
rs369742435 Chr.8: 6732557-6732557 on Build GRCh38
rs761507909 Chr.8: 6732596-6732596 on Build GRCh38
rs1030619980 Chr.8: 6732720-6732720 on Build GRCh38
rs966425837 Chr.8: 6732739-6732739 on Build GRCh38
rs1010923172 Chr.8: 6732835-6732835 on Build GRCh38
rs768230665 Chr.8: 6732592-6732592 on Build GRCh38
rs1022876117 Chr.8: 6732905-6732905 on Build GRCh38
rs756114238 Chr.8: 6732755-6732761 on Build GRCh38
rs375690911 Chr.8: 6732552-6732552 on Build GRCh38
rs766987735 Chr.8: 6732600-6732600 on Build GRCh38

View Full Product Details