Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 6754865-6755381 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGPAT5
Entrez Gene ID
55326
Gene Name
1-acylglycerol-3-phosphate O-acyltransferase 5
Gene Aliases
1AGPAT5, LPAATE
UniGene
Hs.624002
Gene Chromosome Location
Chr.8: 6708357-6761503 on Build GRCh38
Transcripts
NM_018361.3 XM_011534751.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6596400 c.796G>A p.D266N Chr.8: 6755101-6755101 on Build GRCh38
5409763 c.848A>G p.E283G Chr.8: 6755153-6755153 on Build GRCh38
6596401 c.856G>A p.E286K Chr.8: 6755161-6755161 on Build GRCh38
1101020 c.750T>C p.F250F Chr.8: 6755055-6755055 on Build GRCh38
1101021 c.791G>A p.R264H Chr.8: 6755096-6755096 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748447210 Chr.8: 6755031-6755031 on Build GRCh38
rs769547566 Chr.8: 6755066-6755066 on Build GRCh38
rs77592079 Chr.8: 6755318-6755318 on Build GRCh38
rs550269011 Chr.8: 6755132-6755132 on Build GRCh38
rs763560825 Chr.8: 6755082-6755082 on Build GRCh38
rs745684355 Chr.8: 6755063-6755063 on Build GRCh38
rs755097515 Chr.8: 6755029-6755029 on Build GRCh38
rs763211515 Chr.8: 6755155-6755155 on Build GRCh38
rs781181470 Chr.8: 6755177-6755177 on Build GRCh38
rs141288167 Chr.8: 6755138-6755138 on Build GRCh38
rs923233499 Chr.8: 6755278-6755278 on Build GRCh38
rs909123914 Chr.8: 6755251-6755251 on Build GRCh38
rs1032577053 Chr.8: 6755195-6755195 on Build GRCh38
rs771588932 Chr.8: 6755194-6755194 on Build GRCh38
rs776231721 Chr.8: 6755049-6755049 on Build GRCh38
rs751787560 Chr.8: 6755215-6755215 on Build GRCh38
rs535022183 Chr.8: 6755229-6755229 on Build GRCh38
rs995977055 Chr.8: 6754896-6754896 on Build GRCh38
rs890312394 Chr.8: 6755299-6755299 on Build GRCh38
rs11997032 Chr.8: 6754913-6754913 on Build GRCh38
rs750910515 Chr.8: 6755292-6755292 on Build GRCh38
rs772629927 Chr.8: 6755196-6755196 on Build GRCh38
rs562418107 Chr.8: 6754945-6754945 on Build GRCh38
rs778041069 Chr.8: 6755036-6755036 on Build GRCh38
rs756074105 Chr.8: 6755184-6755184 on Build GRCh38
rs574641058 Chr.8: 6755115-6755115 on Build GRCh38
rs10692820 Chr.8: 6754896-6754897 on Build GRCh38
rs1037600241 Chr.8: 6754895-6754895 on Build GRCh38
rs753136839 Chr.8: 6755150-6755150 on Build GRCh38
rs772884046 Chr.8: 6755028-6755028 on Build GRCh38
rs771547871 Chr.8: 6755141-6755141 on Build GRCh38
rs1055756865 Chr.8: 6755343-6755343 on Build GRCh38
rs957003794 Chr.8: 6754930-6754930 on Build GRCh38
rs1012580759 Chr.8: 6755203-6755203 on Build GRCh38
rs900381528 Chr.8: 6755124-6755124 on Build GRCh38
rs201277690 Chr.8: 6755030-6755030 on Build GRCh38
rs539276684 Chr.8: 6755018-6755019 on Build GRCh38
rs770668527 Chr.8: 6754966-6754966 on Build GRCh38
rs907559666 Chr.8: 6755042-6755042 on Build GRCh38
rs971675691 Chr.8: 6754996-6754996 on Build GRCh38
rs749091250 Chr.8: 6755186-6755186 on Build GRCh38
rs765753866 Chr.8: 6755096-6755096 on Build GRCh38
rs765658153 Chr.8: 6755149-6755149 on Build GRCh38
rs372017823 Chr.8: 6755201-6755201 on Build GRCh38
rs149508418 Chr.8: 6755334-6755334 on Build GRCh38
rs746562673 Chr.8: 6755043-6755045 on Build GRCh38
rs768314818 Chr.8: 6755133-6755133 on Build GRCh38
rs60834832 Chr.8: 6755302-6755302 on Build GRCh38
rs543620323 Chr.8: 6755262-6755262 on Build GRCh38
rs777951901 Chr.8: 6755113-6755113 on Build GRCh38
rs779773321 Chr.8: 6755185-6755185 on Build GRCh38
rs373235746 Chr.8: 6755023-6755023 on Build GRCh38
rs764417548 Chr.8: 6755160-6755160 on Build GRCh38
rs539211862 Chr.8: 6755100-6755100 on Build GRCh38
rs759279355 Chr.8: 6755146-6755146 on Build GRCh38
rs545890196 Chr.8: 6754914-6754914 on Build GRCh38
rs941649567 Chr.8: 6755101-6755101 on Build GRCh38
rs773982882 Chr.8: 6755089-6755089 on Build GRCh38
rs200414711 Chr.8: 6755027-6755027 on Build GRCh38
rs372314411 Chr.8: 6754904-6754904 on Build GRCh38
rs183421077 Chr.8: 6755247-6755247 on Build GRCh38
rs750904814 Chr.8: 6755000-6755000 on Build GRCh38
rs1020314126 Chr.8: 6754958-6754958 on Build GRCh38
rs773879896 Chr.8: 6755088-6755088 on Build GRCh38
rs770924591 Chr.8: 6755041-6755041 on Build GRCh38
rs57125367 Chr.8: 6755191-6755191 on Build GRCh38
rs527632127 Chr.8: 6755033-6755033 on Build GRCh38
rs775301782 Chr.8: 6755130-6755130 on Build GRCh38
rs773071881 Chr.8: 6754890-6754890 on Build GRCh38
rs931948574 Chr.8: 6755271-6755271 on Build GRCh38
rs376372622 Chr.8: 6755210-6755210 on Build GRCh38
rs751350821 Chr.8: 6754921-6754921 on Build GRCh38
rs564382822 Chr.8: 6754903-6754903 on Build GRCh38
rs377449323 Chr.8: 6755148-6755148 on Build GRCh38
rs1051640200 Chr.8: 6755142-6755142 on Build GRCh38
rs773789806 Chr.8: 6755134-6755134 on Build GRCh38
rs974775923 Chr.8: 6755268-6755268 on Build GRCh38
rs973107166 Chr.8: 6755248-6755248 on Build GRCh38
rs761985316 Chr.8: 6755218-6755218 on Build GRCh38
rs570636724 Chr.8: 6755095-6755095 on Build GRCh38
rs770440658 Chr.8: 6755202-6755202 on Build GRCh38
rs1010731693 Chr.8: 6754957-6754957 on Build GRCh38
rs781190048 Chr.8: 6755119-6755119 on Build GRCh38
rs1026163857 Chr.8: 6755227-6755227 on Build GRCh38
rs762646139 Chr.8: 6755077-6755077 on Build GRCh38
rs375711445 Chr.8: 6755230-6755230 on Build GRCh38
rs936810750 Chr.8: 6755327-6755327 on Build GRCh38
rs1028808189 Chr.8: 6754912-6754912 on Build GRCh38
rs771707447 Chr.8: 6755019-6755019 on Build GRCh38
rs757078913 Chr.8: 6755118-6755118 on Build GRCh38
rs2980700 Chr.8: 6755315-6755315 on Build GRCh38
rs547705041 Chr.8: 6755093-6755093 on Build GRCh38
rs775113398 Chr.8: 6755069-6755069 on Build GRCh38
rs745853589 Chr.8: 6755120-6755120 on Build GRCh38
rs777180093 Chr.8: 6755144-6755144 on Build GRCh38
rs201303494 Chr.8: 6755106-6755106 on Build GRCh38
rs202152371 Chr.8: 6755161-6755161 on Build GRCh38
rs768226942 Chr.8: 6755188-6755188 on Build GRCh38
rs765852279 Chr.8: 6755209-6755209 on Build GRCh38
rs756682658 Chr.8: 6755004-6755004 on Build GRCh38
rs371758285 Chr.8: 6755206-6755206 on Build GRCh38
rs966991241 Chr.8: 6754904-6754904 on Build GRCh38
rs958869960 Chr.8: 6755011-6755011 on Build GRCh38
rs58366914 Chr.8: 6755190-6755190 on Build GRCh38
rs191993349 Chr.8: 6754983-6754983 on Build GRCh38
rs570017791 Chr.8: 6755152-6755152 on Build GRCh38
rs921592226 Chr.8: 6755269-6755269 on Build GRCh38
rs1054075282 Chr.8: 6755332-6755332 on Build GRCh38
rs117694987 Chr.8: 6755351-6755351 on Build GRCh38
rs746906053 Chr.8: 6755037-6755037 on Build GRCh38
rs1052099099 Chr.8: 6755294-6755294 on Build GRCh38
rs570063451 Chr.8: 6754895-6754896 on Build GRCh38
rs769558727 Chr.8: 6755125-6755125 on Build GRCh38
rs770545239 Chr.8: 6755181-6755183 on Build GRCh38
rs750346829 Chr.8: 6755182-6755182 on Build GRCh38

View Full Product Details