Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 11424431-11424925 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM167A
Entrez Gene ID
83648
Gene Name
family with sequence similarity 167 member A
Gene Aliases
C8orf13, D8S265
UniGene
Hs.124299
Gene Chromosome Location
Chr.8: 11421464-11475904 on Build GRCh38
Transcripts
NM_053279.2 XM_005272398.4 XM_011543837.1 XM_011543838.2 XM_011543840.2 NR_026814.1

Gene Symbol
FAM167A-AS1
Entrez Gene ID
83656
Gene Name
FAM167A antisense RNA 1
Gene Aliases
C8orf12
UniGene
Hs.583896
Gene Chromosome Location
Chr.8: 11368402-11438657 on Build GRCh38
Transcripts
NM_053279.2 XM_005272398.4 XM_011543837.1 XM_011543838.2 XM_011543840.2 NR_026814.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1643471 c.563C>A p.S188Y Chr.8: 11424455-11424455 on Build GRCh38
4749961 c.520C>T p.R174W Chr.8: 11424498-11424498 on Build GRCh38
7367129 c.460G>C p.E154Q Chr.8: 11424558-11424558 on Build GRCh38
266501 c.437G>T p.G146V Chr.8: 11424581-11424581 on Build GRCh38
1623518 c.528G>A p.E176E Chr.8: 11424490-11424490 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs922544676 Chr.8: 11424777-11424777 on Build GRCh38
rs768510757 Chr.8: 11424454-11424454 on Build GRCh38
rs760183930 Chr.8: 11424626-11424626 on Build GRCh38
rs780501946 Chr.8: 11424558-11424558 on Build GRCh38
rs111729419 Chr.8: 11424684-11424684 on Build GRCh38
rs752012096 Chr.8: 11424488-11424488 on Build GRCh38
rs567362067 Chr.8: 11424642-11424642 on Build GRCh38
rs1010572287 Chr.8: 11424896-11424896 on Build GRCh38
rs538682492 Chr.8: 11424719-11424719 on Build GRCh38
rs140423769 Chr.8: 11424545-11424545 on Build GRCh38
rs538000985 Chr.8: 11424698-11424698 on Build GRCh38
rs750839979 Chr.8: 11424494-11424494 on Build GRCh38
rs148850114 Chr.8: 11424712-11424712 on Build GRCh38
rs756046453 Chr.8: 11424654-11424655 on Build GRCh38
rs1011016398 Chr.8: 11424672-11424672 on Build GRCh38
rs940693820 Chr.8: 11424607-11424607 on Build GRCh38
rs778894458 Chr.8: 11424568-11424568 on Build GRCh38
rs761836545 Chr.8: 11424793-11424796 on Build GRCh38
rs980256350 Chr.8: 11424746-11424746 on Build GRCh38
rs762583224 Chr.8: 11424593-11424593 on Build GRCh38
rs1022062195 Chr.8: 11424559-11424559 on Build GRCh38
rs564594491 Chr.8: 11424646-11424647 on Build GRCh38
rs1017007026 Chr.8: 11424763-11424763 on Build GRCh38
rs577482093 Chr.8: 11424713-11424713 on Build GRCh38
rs369090343 Chr.8: 11424497-11424497 on Build GRCh38
rs761747530 Chr.8: 11424464-11424464 on Build GRCh38
rs533627555 Chr.8: 11424634-11424634 on Build GRCh38
rs139424835 Chr.8: 11424455-11424455 on Build GRCh38
rs762338989 Chr.8: 11424533-11424533 on Build GRCh38
rs1041736999 Chr.8: 11424680-11424680 on Build GRCh38
rs774348671 Chr.8: 11424571-11424571 on Build GRCh38
rs755367917 Chr.8: 11424606-11424606 on Build GRCh38
rs370927037 Chr.8: 11424596-11424596 on Build GRCh38
rs561322004 Chr.8: 11424483-11424483 on Build GRCh38
rs757296081 Chr.8: 11424668-11424668 on Build GRCh38
rs1006115476 Chr.8: 11424885-11424885 on Build GRCh38
rs146816358 Chr.8: 11424490-11424490 on Build GRCh38
rs763106621 Chr.8: 11424552-11424552 on Build GRCh38
rs565530389 Chr.8: 11424548-11424548 on Build GRCh38
rs150406646 Chr.8: 11424550-11424550 on Build GRCh38
rs17153216 Chr.8: 11424757-11424757 on Build GRCh38
rs752201866 Chr.8: 11424532-11424532 on Build GRCh38
rs748294847 Chr.8: 11424575-11424575 on Build GRCh38
rs572423147 Chr.8: 11424845-11424845 on Build GRCh38
rs1055995813 Chr.8: 11424705-11424705 on Build GRCh38
rs73540421 Chr.8: 11424876-11424876 on Build GRCh38
rs892100302 Chr.8: 11424670-11424670 on Build GRCh38
rs1029670849 Chr.8: 11424900-11424900 on Build GRCh38
rs745542182 Chr.8: 11424590-11424590 on Build GRCh38
rs755206316 Chr.8: 11424567-11424567 on Build GRCh38
rs765541335 Chr.8: 11424609-11424609 on Build GRCh38
rs780996003 Chr.8: 11424628-11424628 on Build GRCh38
rs771780754 Chr.8: 11424465-11424465 on Build GRCh38
rs901019403 Chr.8: 11424514-11424514 on Build GRCh38
rs550216545 Chr.8: 11424531-11424531 on Build GRCh38
rs960504292 Chr.8: 11424768-11424768 on Build GRCh38
rs372968845 Chr.8: 11424505-11424505 on Build GRCh38
rs1025810371 Chr.8: 11424739-11424739 on Build GRCh38
rs184887418 Chr.8: 11424878-11424878 on Build GRCh38
rs566603440 Chr.8: 11424625-11424625 on Build GRCh38
rs372350472 Chr.8: 11424473-11424473 on Build GRCh38
rs758382682 Chr.8: 11424579-11424579 on Build GRCh38
rs553728707 Chr.8: 11424724-11424724 on Build GRCh38
rs754305260 Chr.8: 11424734-11424734 on Build GRCh38
rs950336785 Chr.8: 11424852-11424852 on Build GRCh38
rs765620953 Chr.8: 11424515-11424522 on Build GRCh38
rs772010209 Chr.8: 11424508-11424508 on Build GRCh38
rs1043834323 Chr.8: 11424676-11424676 on Build GRCh38
rs913743855 Chr.8: 11424802-11424802 on Build GRCh38
rs766283803 Chr.8: 11424668-11424669 on Build GRCh38
rs779654158 Chr.8: 11424687-11424687 on Build GRCh38
rs749519377 Chr.8: 11424498-11424498 on Build GRCh38
rs750355812 Chr.8: 11424686-11424686 on Build GRCh38
rs758597360 Chr.8: 11424824-11424824 on Build GRCh38
rs373127114 Chr.8: 11424660-11424660 on Build GRCh38
rs903470906 Chr.8: 11424683-11424683 on Build GRCh38
rs558608053 Chr.8: 11424766-11424766 on Build GRCh38
rs777251081 Chr.8: 11424652-11424652 on Build GRCh38
rs776331059 Chr.8: 11424523-11424523 on Build GRCh38
rs146979421 Chr.8: 11424591-11424591 on Build GRCh38
rs757255814 Chr.8: 11424507-11424507 on Build GRCh38
rs940685207 Chr.8: 11424817-11424817 on Build GRCh38
rs561393537 Chr.8: 11424779-11424779 on Build GRCh38
rs1001750743 Chr.8: 11424558-11424559 on Build GRCh38
rs770696934 Chr.8: 11424522-11424522 on Build GRCh38
rs761299096 Chr.8: 11424649-11424649 on Build GRCh38
rs911705504 Chr.8: 11424860-11424860 on Build GRCh38
rs1043475779 Chr.8: 11424819-11424819 on Build GRCh38
rs201058431 Chr.8: 11424528-11424528 on Build GRCh38
rs754766828 Chr.8: 11424671-11424679 on Build GRCh38
rs1008791752 Chr.8: 11424650-11424650 on Build GRCh38
rs149662282 Chr.8: 11424511-11424511 on Build GRCh38
rs942595478 Chr.8: 11424496-11424496 on Build GRCh38
rs1043444067 Chr.8: 11424899-11424899 on Build GRCh38
rs145481836 Chr.8: 11424584-11424584 on Build GRCh38
rs138151717 Chr.8: 11424600-11424600 on Build GRCh38
rs576562407 Chr.8: 11424823-11424823 on Build GRCh38
rs79924610 Chr.8: 11424888-11424888 on Build GRCh38
rs779243865 Chr.8: 11424820-11424820 on Build GRCh38
rs746960113 Chr.8: 11424627-11424627 on Build GRCh38
rs760272883 Chr.8: 11424479-11424479 on Build GRCh38
rs931797359 Chr.8: 11424633-11424633 on Build GRCh38
rs143521673 Chr.8: 11424635-11424635 on Build GRCh38
rs754218407 Chr.8: 11424602-11424602 on Build GRCh38
rs946515609 Chr.8: 11424816-11424816 on Build GRCh38
rs775785936 Chr.8: 11424516-11424517 on Build GRCh38
rs752970462 Chr.8: 11424657-11424657 on Build GRCh38
rs138628802 Chr.8: 11424565-11424565 on Build GRCh38
rs200337863 Chr.8: 11424595-11424595 on Build GRCh38
rs752846903 Chr.8: 11424612-11424612 on Build GRCh38
rs757724438 Chr.8: 11424489-11424489 on Build GRCh38
rs542744918 Chr.8: 11424776-11424776 on Build GRCh38
rs200487180 Chr.8: 11424546-11424546 on Build GRCh38
rs753694360 Chr.8: 11424670-11424670 on Build GRCh38
rs781164142 Chr.8: 11424673-11424673 on Build GRCh38
rs759102263 Chr.8: 11424484-11424484 on Build GRCh38
rs767037170 Chr.8: 11424651-11424651 on Build GRCh38
rs536449463 Chr.8: 11424807-11424807 on Build GRCh38
rs760000055 Chr.8: 11424654-11424654 on Build GRCh38
rs368405922 Chr.8: 11424492-11424492 on Build GRCh38
rs773953753 Chr.8: 11424648-11424648 on Build GRCh38
rs775280270 Chr.8: 11424647-11424647 on Build GRCh38
rs749935355 Chr.8: 11424585-11424585 on Build GRCh38
rs760508594 Chr.8: 11424516-11424516 on Build GRCh38
rs892200041 Chr.8: 11424542-11424542 on Build GRCh38
rs766056567 Chr.8: 11424481-11424481 on Build GRCh38
rs751599012 Chr.8: 11424661-11424661 on Build GRCh38
rs951596015 Chr.8: 11424749-11424749 on Build GRCh38
rs988423024 Chr.8: 11424791-11424794 on Build GRCh38
rs1022034019 Chr.8: 11424690-11424690 on Build GRCh38
rs761556428 Chr.8: 11424580-11424580 on Build GRCh38
rs896059633 Chr.8: 11424711-11424711 on Build GRCh38

View Full Product Details