Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 56958002-56958533 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IMPAD1
Entrez Gene ID
54928
Gene Name
inositol monophosphatase domain containing 1
Gene Aliases
GPAPP, IMP 3, IMP-3, IMPA3
UniGene
Hs.438689
Gene Chromosome Location
Chr.8: 56957929-56993874 on Build GRCh38
Transcripts
NM_017813.4

SNP Detail

SNP ID Chromosome Location
rs1041649196 Chr.8: 56958042-56958042 on Build GRCh38
rs539157521 Chr.8: 56958351-56958351 on Build GRCh38
rs758718311 Chr.8: 56958082-56958082 on Build GRCh38
rs778368311 Chr.8: 56958336-56958336 on Build GRCh38
rs976885405 Chr.8: 56958458-56958458 on Build GRCh38
rs566730602 Chr.8: 56958342-56958342 on Build GRCh38
rs111509692 Chr.8: 56958159-56958159 on Build GRCh38
rs558774184 Chr.8: 56958415-56958415 on Build GRCh38
rs894589663 Chr.8: 56958147-56958147 on Build GRCh38
rs979504258 Chr.8: 56958141-56958144 on Build GRCh38
rs886063000 Chr.8: 56958041-56958041 on Build GRCh38
rs946256666 Chr.8: 56958297-56958297 on Build GRCh38
rs748963295 Chr.8: 56958312-56958312 on Build GRCh38
rs147613200 Chr.8: 56958179-56958179 on Build GRCh38
rs752863184 Chr.8: 56958151-56958151 on Build GRCh38
rs1017375276 Chr.8: 56958251-56958251 on Build GRCh38
rs901418794 Chr.8: 56958058-56958058 on Build GRCh38
rs973036238 Chr.8: 56958166-56958166 on Build GRCh38
rs9315 Chr.8: 56958281-56958281 on Build GRCh38
rs757255652 Chr.8: 56958152-56958152 on Build GRCh38
rs755055338 Chr.8: 56958101-56958101 on Build GRCh38
rs533853243 Chr.8: 56958184-56958184 on Build GRCh38
rs946773320 Chr.8: 56958118-56958118 on Build GRCh38
rs926223097 Chr.8: 56958309-56958309 on Build GRCh38
rs80163062 Chr.8: 56958063-56958063 on Build GRCh38
rs113019850 Chr.8: 56958301-56958301 on Build GRCh38
rs770098671 Chr.8: 56958461-56958461 on Build GRCh38
rs765382421 Chr.8: 56958103-56958103 on Build GRCh38
rs952369156 Chr.8: 56958196-56958196 on Build GRCh38
rs956064298 Chr.8: 56958084-56958084 on Build GRCh38
rs761057108 Chr.8: 56958509-56958509 on Build GRCh38
rs34733086 Chr.8: 56958071-56958071 on Build GRCh38
rs575841443 Chr.8: 56958439-56958439 on Build GRCh38
rs745619288 Chr.8: 56958225-56958225 on Build GRCh38
rs1034423120 Chr.8: 56958137-56958137 on Build GRCh38
rs766553227 Chr.8: 56958053-56958053 on Build GRCh38
rs10763 Chr.8: 56958491-56958491 on Build GRCh38
rs531529689 Chr.8: 56958104-56958104 on Build GRCh38
rs200958042 Chr.8: 56958466-56958466 on Build GRCh38
rs1030645756 Chr.8: 56958291-56958291 on Build GRCh38
rs926760812 Chr.8: 56958350-56958350 on Build GRCh38
rs894486700 Chr.8: 56958328-56958328 on Build GRCh38
rs193125151 Chr.8: 56958446-56958446 on Build GRCh38
rs747090240 Chr.8: 56958249-56958249 on Build GRCh38
rs781048135 Chr.8: 56958186-56958186 on Build GRCh38
rs979450429 Chr.8: 56958338-56958338 on Build GRCh38
rs904501474 Chr.8: 56958100-56958100 on Build GRCh38
rs8718 Chr.8: 56958288-56958288 on Build GRCh38
rs1045830825 Chr.8: 56958333-56958333 on Build GRCh38
rs561796338 Chr.8: 56958398-56958398 on Build GRCh38
rs994043942 Chr.8: 56958374-56958374 on Build GRCh38
rs112348747 Chr.8: 56958244-56958244 on Build GRCh38
rs879207350 Chr.8: 56958460-56958460 on Build GRCh38
rs540499421 Chr.8: 56958493-56958493 on Build GRCh38
rs542901433 Chr.8: 56958330-56958330 on Build GRCh38
rs200630025 Chr.8: 56958103-56958104 on Build GRCh38
rs913975105 Chr.8: 56958112-56958112 on Build GRCh38
rs760451290 Chr.8: 56958112-56958112 on Build GRCh38
rs532473229 Chr.8: 56958081-56958081 on Build GRCh38
rs747726327 Chr.8: 56958414-56958414 on Build GRCh38
rs1017862490 Chr.8: 56958432-56958432 on Build GRCh38
rs926192112 Chr.8: 56958479-56958479 on Build GRCh38
rs1047899779 Chr.8: 56958210-56958210 on Build GRCh38
rs781611206 Chr.8: 56958305-56958305 on Build GRCh38
rs935970734 Chr.8: 56958311-56958311 on Build GRCh38
rs572170196 Chr.8: 56958325-56958326 on Build GRCh38
rs983773195 Chr.8: 56958200-56958200 on Build GRCh38

View Full Product Details