Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 56960740-56961250 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IMPAD1
Entrez Gene ID
54928
Gene Name
inositol monophosphatase domain containing 1
Gene Aliases
GPAPP, IMP 3, IMP-3, IMPA3
UniGene
Hs.438689
Gene Chromosome Location
Chr.8: 56957929-56993874 on Build GRCh38
Transcripts
NM_017813.4

SNP Detail

SNP ID Chromosome Location
rs142089059 Chr.8: 56961018-56961018 on Build GRCh38
rs547787251 Chr.8: 56961003-56961003 on Build GRCh38
rs151168258 Chr.8: 56961162-56961162 on Build GRCh38
rs1001396551 Chr.8: 56960890-56960890 on Build GRCh38
rs17188824 Chr.8: 56961185-56961185 on Build GRCh38
rs893821599 Chr.8: 56960838-56960838 on Build GRCh38
rs549076597 Chr.8: 56961212-56961212 on Build GRCh38
rs935317890 Chr.8: 56960806-56960806 on Build GRCh38
rs563142061 Chr.8: 56960858-56960858 on Build GRCh38
rs990195578 Chr.8: 56960894-56960894 on Build GRCh38
rs1029157112 Chr.8: 56961213-56961213 on Build GRCh38
rs1045194205 Chr.8: 56960847-56960847 on Build GRCh38
rs772899410 Chr.8: 56960968-56960968 on Build GRCh38
rs1052692288 Chr.8: 56961067-56961067 on Build GRCh38
rs953961094 Chr.8: 56960785-56960785 on Build GRCh38
rs535290326 Chr.8: 56961186-56961186 on Build GRCh38
rs529475020 Chr.8: 56961063-56961065 on Build GRCh38
rs550541748 Chr.8: 56961163-56961163 on Build GRCh38
rs935537110 Chr.8: 56961062-56961062 on Build GRCh38
rs886063006 Chr.8: 56961067-56961067 on Build GRCh38
rs1019368210 Chr.8: 56960822-56960822 on Build GRCh38
rs987697635 Chr.8: 56961033-56961033 on Build GRCh38
rs905666321 Chr.8: 56960848-56960848 on Build GRCh38
rs1045535979 Chr.8: 56961121-56961121 on Build GRCh38
rs529096539 Chr.8: 56960891-56960891 on Build GRCh38
rs915223888 Chr.8: 56961081-56961081 on Build GRCh38
rs1036011855 Chr.8: 56960897-56960897 on Build GRCh38
rs937161433 Chr.8: 56961146-56961146 on Build GRCh38
rs963032684 Chr.8: 56960972-56960972 on Build GRCh38
rs746490472 Chr.8: 56961117-56961117 on Build GRCh38
rs1010958390 Chr.8: 56960840-56960840 on Build GRCh38
rs1036765683 Chr.8: 56961063-56961063 on Build GRCh38
rs953654185 Chr.8: 56961031-56961031 on Build GRCh38
rs1052470122 Chr.8: 56960836-56960836 on Build GRCh38
rs1008582814 Chr.8: 56960814-56960814 on Build GRCh38
rs973097974 Chr.8: 56960988-56960988 on Build GRCh38
rs73681999 Chr.8: 56960930-56960930 on Build GRCh38
rs527659094 Chr.8: 56960987-56960987 on Build GRCh38
rs957581556 Chr.8: 56960862-56960862 on Build GRCh38
rs912170999 Chr.8: 56960804-56960804 on Build GRCh38
rs921731467 Chr.8: 56961029-56961029 on Build GRCh38
rs775806342 Chr.8: 56961119-56961119 on Build GRCh38
rs749451389 Chr.8: 56961141-56961141 on Build GRCh38
rs1052433688 Chr.8: 56960800-56960800 on Build GRCh38
rs1049383938 Chr.8: 56961172-56961172 on Build GRCh38
rs886063005 Chr.8: 56960800-56960800 on Build GRCh38
rs768963800 Chr.8: 56961144-56961144 on Build GRCh38
rs924737211 Chr.8: 56960856-56960856 on Build GRCh38
rs1014806444 Chr.8: 56961178-56961178 on Build GRCh38
rs1056931310 Chr.8: 56961157-56961157 on Build GRCh38
rs1004763985 Chr.8: 56960937-56960937 on Build GRCh38
rs571017371 Chr.8: 56961051-56961051 on Build GRCh38
rs771696234 Chr.8: 56960885-56960885 on Build GRCh38
rs561877081 Chr.8: 56960978-56960978 on Build GRCh38

View Full Product Details