Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 56962340-56962856 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IMPAD1
Entrez Gene ID
54928
Gene Name
inositol monophosphatase domain containing 1
Gene Aliases
GPAPP, IMP 3, IMP-3, IMPA3
UniGene
Hs.438689
Gene Chromosome Location
Chr.8: 56957929-56993874 on Build GRCh38
Transcripts
NM_017813.4

SNP Detail

SNP ID Chromosome Location
rs969441828 Chr.8: 56962651-56962651 on Build GRCh38
rs559826487 Chr.8: 56962775-56962775 on Build GRCh38
rs75984016 Chr.8: 56962517-56962517 on Build GRCh38
rs919110006 Chr.8: 56962475-56962475 on Build GRCh38
rs879024718 Chr.8: 56962779-56962779 on Build GRCh38
rs558202697 Chr.8: 56962619-56962619 on Build GRCh38
rs879131458 Chr.8: 56962450-56962450 on Build GRCh38
rs543293679 Chr.8: 56962760-56962760 on Build GRCh38
rs965887969 Chr.8: 56962426-56962426 on Build GRCh38
rs748318896 Chr.8: 56962655-56962664 on Build GRCh38
rs1054892998 Chr.8: 56962609-56962609 on Build GRCh38
rs1011067509 Chr.8: 56962632-56962632 on Build GRCh38
rs975979329 Chr.8: 56962445-56962445 on Build GRCh38
rs919057911 Chr.8: 56962716-56962716 on Build GRCh38
rs960591558 Chr.8: 56962672-56962672 on Build GRCh38
rs901460502 Chr.8: 56962545-56962545 on Build GRCh38
rs886063014 Chr.8: 56962660-56962666 on Build GRCh38
rs965856654 Chr.8: 56962794-56962794 on Build GRCh38
rs969117748 Chr.8: 56962391-56962391 on Build GRCh38
rs942680159 Chr.8: 56962780-56962780 on Build GRCh38
rs896622253 Chr.8: 56962637-56962637 on Build GRCh38
rs1039912776 Chr.8: 56962544-56962547 on Build GRCh38
rs780664648 Chr.8: 56962669-56962670 on Build GRCh38
rs113846940 Chr.8: 56962813-56962813 on Build GRCh38
rs756685856 Chr.8: 56962531-56962531 on Build GRCh38
rs879483892 Chr.8: 56962827-56962830 on Build GRCh38
rs892644976 Chr.8: 56962627-56962627 on Build GRCh38
rs1027368551 Chr.8: 56962694-56962694 on Build GRCh38
rs1031090846 Chr.8: 56962599-56962599 on Build GRCh38
rs567177259 Chr.8: 56962427-56962427 on Build GRCh38
rs996193609 Chr.8: 56962424-56962424 on Build GRCh38
rs886063015 Chr.8: 56962709-56962709 on Build GRCh38
rs980492428 Chr.8: 56962666-56962666 on Build GRCh38
rs780767619 Chr.8: 56962667-56962667 on Build GRCh38
rs1027060835 Chr.8: 56962387-56962387 on Build GRCh38
rs528660052 Chr.8: 56962797-56962797 on Build GRCh38
rs981093063 Chr.8: 56962398-56962398 on Build GRCh38
rs749367444 Chr.8: 56962721-56962722 on Build GRCh38
rs578154957 Chr.8: 56962644-56962644 on Build GRCh38
rs1030398180 Chr.8: 56962488-56962488 on Build GRCh38
rs945016996 Chr.8: 56962496-56962496 on Build GRCh38
rs998404986 Chr.8: 56962574-56962574 on Build GRCh38
rs985572290 Chr.8: 56962399-56962399 on Build GRCh38
rs566643756 Chr.8: 56962553-56962553 on Build GRCh38
rs1048480322 Chr.8: 56962514-56962514 on Build GRCh38
rs767114591 Chr.8: 56962395-56962395 on Build GRCh38
rs149350368 Chr.8: 56962417-56962417 on Build GRCh38
rs749994859 Chr.8: 56962479-56962479 on Build GRCh38
rs971976992 Chr.8: 56962675-56962675 on Build GRCh38
rs913850270 Chr.8: 56962465-56962465 on Build GRCh38
rs995462662 Chr.8: 56962681-56962681 on Build GRCh38
rs199907097 Chr.8: 56962480-56962480 on Build GRCh38
rs557393997 Chr.8: 56962727-56962727 on Build GRCh38
rs369301033 Chr.8: 56962670-56962670 on Build GRCh38
rs143679493 Chr.8: 56962573-56962573 on Build GRCh38
rs543588217 Chr.8: 56962455-56962455 on Build GRCh38
rs574127712 Chr.8: 56962741-56962741 on Build GRCh38
rs897443481 Chr.8: 56962680-56962680 on Build GRCh38
rs553033659 Chr.8: 56962540-56962540 on Build GRCh38

View Full Product Details