Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 61287821-61288325 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLVS1
Entrez Gene ID
157807
Gene Name
clavesin 1
Gene Aliases
CRALBPL, RLBP1L1
UniGene
Hs.591874
Gene Chromosome Location
Chr.8: 61057158-61501645 on Build GRCh38
Transcripts
NM_173519.2 XM_017013141.1 XM_017013142.1

SNP Detail

SNP ID Chromosome Location
rs575449347 Chr.8: 61288196-61288196 on Build GRCh38
rs894069873 Chr.8: 61288013-61288013 on Build GRCh38
rs769975384 Chr.8: 61288234-61288234 on Build GRCh38
rs552207841 Chr.8: 61287906-61287906 on Build GRCh38
rs946806014 Chr.8: 61288282-61288282 on Build GRCh38
rs953817863 Chr.8: 61288237-61288237 on Build GRCh38
rs562162782 Chr.8: 61288293-61288293 on Build GRCh38
rs765136102 Chr.8: 61288258-61288259 on Build GRCh38
rs765708736 Chr.8: 61288154-61288154 on Build GRCh38
rs529440088 Chr.8: 61288304-61288304 on Build GRCh38
rs117448847 Chr.8: 61287984-61287984 on Build GRCh38
rs982540903 Chr.8: 61287940-61287940 on Build GRCh38
rs746312902 Chr.8: 61288269-61288269 on Build GRCh38
rs962484784 Chr.8: 61288217-61288217 on Build GRCh38
rs914029186 Chr.8: 61288143-61288143 on Build GRCh38
rs1016073862 Chr.8: 61288167-61288167 on Build GRCh38
rs754196408 Chr.8: 61288106-61288106 on Build GRCh38
rs563836023 Chr.8: 61287960-61287960 on Build GRCh38
rs113830027 Chr.8: 61287882-61287882 on Build GRCh38
rs961135461 Chr.8: 61287938-61287938 on Build GRCh38
rs1057089500 Chr.8: 61288184-61288184 on Build GRCh38
rs771644464 Chr.8: 61288277-61288277 on Build GRCh38
rs920918886 Chr.8: 61288046-61288046 on Build GRCh38
rs779487790 Chr.8: 61288253-61288253 on Build GRCh38
rs888832316 Chr.8: 61288121-61288121 on Build GRCh38
rs995220155 Chr.8: 61288231-61288231 on Build GRCh38
rs573152589 Chr.8: 61288225-61288225 on Build GRCh38
rs576098462 Chr.8: 61287980-61287980 on Build GRCh38
rs763684409 Chr.8: 61287933-61287933 on Build GRCh38
rs776349144 Chr.8: 61288162-61288163 on Build GRCh38
rs12677628 Chr.8: 61287916-61287916 on Build GRCh38
rs1007404308 Chr.8: 61288123-61288123 on Build GRCh38
rs768287914 Chr.8: 61288283-61288283 on Build GRCh38
rs985978775 Chr.8: 61287881-61287881 on Build GRCh38
rs151282857 Chr.8: 61288066-61288066 on Build GRCh38
rs938219572 Chr.8: 61288008-61288008 on Build GRCh38
rs953367487 Chr.8: 61287856-61287856 on Build GRCh38
rs926657738 Chr.8: 61288286-61288286 on Build GRCh38
rs915387449 Chr.8: 61288236-61288236 on Build GRCh38
rs761531336 Chr.8: 61288004-61288004 on Build GRCh38
rs1022252113 Chr.8: 61287903-61287903 on Build GRCh38
rs546448967 Chr.8: 61288210-61288210 on Build GRCh38
rs905153060 Chr.8: 61288164-61288164 on Build GRCh38
rs184823895 Chr.8: 61288190-61288190 on Build GRCh38
rs945603606 Chr.8: 61288024-61288024 on Build GRCh38
rs1044176688 Chr.8: 61288285-61288285 on Build GRCh38
rs866727154 Chr.8: 61288288-61288288 on Build GRCh38
rs1033657508 Chr.8: 61288303-61288308 on Build GRCh38
rs970634280 Chr.8: 61288298-61288298 on Build GRCh38
rs1052444838 Chr.8: 61287853-61287853 on Build GRCh38
rs4033256 Chr.8: 61287893-61287893 on Build GRCh38
rs946796038 Chr.8: 61288150-61288150 on Build GRCh38
rs181326827 Chr.8: 61287904-61287904 on Build GRCh38
rs574238705 Chr.8: 61288069-61288069 on Build GRCh38
rs540311089 Chr.8: 61288280-61288280 on Build GRCh38
rs540184034 Chr.8: 61288193-61288193 on Build GRCh38
rs971172122 Chr.8: 61288206-61288206 on Build GRCh38
rs949393113 Chr.8: 61287967-61287967 on Build GRCh38
rs758430749 Chr.8: 61288255-61288255 on Build GRCh38
rs972070297 Chr.8: 61287908-61287908 on Build GRCh38
rs973772561 Chr.8: 61288045-61288045 on Build GRCh38
rs755373230 Chr.8: 61288126-61288126 on Build GRCh38
rs1026691017 Chr.8: 61288291-61288291 on Build GRCh38
rs1051402501 Chr.8: 61288280-61288280 on Build GRCh38
rs993435795 Chr.8: 61288187-61288187 on Build GRCh38
rs1053845703 Chr.8: 61288012-61288012 on Build GRCh38
rs905479120 Chr.8: 61287926-61287926 on Build GRCh38
rs4738880 Chr.8: 61288263-61288263 on Build GRCh38
rs372097615 Chr.8: 61288195-61288195 on Build GRCh38
rs1002478310 Chr.8: 61288166-61288166 on Build GRCh38
rs4033255 Chr.8: 61287876-61287876 on Build GRCh38
rs1002836128 Chr.8: 61287930-61287930 on Build GRCh38
rs963307160 Chr.8: 61288174-61288174 on Build GRCh38
rs188139656 Chr.8: 61287892-61287892 on Build GRCh38
rs1015380132 Chr.8: 61287969-61287969 on Build GRCh38
rs1041384298 Chr.8: 61288062-61288062 on Build GRCh38
rs912496472 Chr.8: 61288259-61288259 on Build GRCh38
rs80218182 Chr.8: 61288068-61288068 on Build GRCh38
rs1052868095 Chr.8: 61288131-61288131 on Build GRCh38
rs1003963818 Chr.8: 61288301-61288301 on Build GRCh38
rs559697414 Chr.8: 61288211-61288211 on Build GRCh38
rs759513976 Chr.8: 61288181-61288181 on Build GRCh38

View Full Product Details