Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 66080240-66080747 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC5B
Entrez Gene ID
85479
Gene Name
DnaJ heat shock protein family (Hsp40) member C5 beta
Gene Aliases
CSP-beta
UniGene
Hs.491885
Gene Chromosome Location
Chr.8: 66014983-66100520 on Build GRCh38
Transcripts
NM_033105.4 XM_011517618.2 XM_011517619.2 XM_011517620.2 XM_011517621.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4564098 c.505_505+1GG>AA p.? Chr.8: 66080548-66080549 on Build GRCh38
6647378 c.456T>C p.Y152Y Chr.8: 66080499-66080499 on Build GRCh38
4445816 c.505+3G>A p.? Chr.8: 66080551-66080551 on Build GRCh38
6647376 c.334-2A>G p.? Chr.8: 66080375-66080375 on Build GRCh38
1624133 c.411T>G p.C137W Chr.8: 66080454-66080454 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368047108 Chr.8: 66080490-66080490 on Build GRCh38
rs754838580 Chr.8: 66080335-66080335 on Build GRCh38
rs955388563 Chr.8: 66080637-66080637 on Build GRCh38
rs765490496 Chr.8: 66080564-66080564 on Build GRCh38
rs766338536 Chr.8: 66080421-66080421 on Build GRCh38
rs932374784 Chr.8: 66080319-66080319 on Build GRCh38
rs139910886 Chr.8: 66080465-66080465 on Build GRCh38
rs756394113 Chr.8: 66080615-66080615 on Build GRCh38
rs960741861 Chr.8: 66080262-66080262 on Build GRCh38
rs751559667 Chr.8: 66080445-66080445 on Build GRCh38
rs780597116 Chr.8: 66080480-66080480 on Build GRCh38
rs202118812 Chr.8: 66080463-66080463 on Build GRCh38
rs999839694 Chr.8: 66080283-66080283 on Build GRCh38
rs866458732 Chr.8: 66080591-66080591 on Build GRCh38
rs28469631 Chr.8: 66080707-66080707 on Build GRCh38
rs748117601 Chr.8: 66080329-66080329 on Build GRCh38
rs374416310 Chr.8: 66080408-66080408 on Build GRCh38
rs761558528 Chr.8: 66080396-66080396 on Build GRCh38
rs777650802 Chr.8: 66080654-66080654 on Build GRCh38
rs780044313 Chr.8: 66080328-66080335 on Build GRCh38
rs762513760 Chr.8: 66080526-66080526 on Build GRCh38
rs189225900 Chr.8: 66080579-66080579 on Build GRCh38
rs370404881 Chr.8: 66080392-66080392 on Build GRCh38
rs750666882 Chr.8: 66080567-66080567 on Build GRCh38
rs536367236 Chr.8: 66080616-66080616 on Build GRCh38
rs1001368619 Chr.8: 66080672-66080672 on Build GRCh38
rs559194838 Chr.8: 66080676-66080676 on Build GRCh38
rs956178174 Chr.8: 66080309-66080309 on Build GRCh38
rs373118799 Chr.8: 66080330-66080330 on Build GRCh38
rs538898617 Chr.8: 66080642-66080642 on Build GRCh38
rs776361838 Chr.8: 66080393-66080393 on Build GRCh38
rs76648418 Chr.8: 66080306-66080306 on Build GRCh38
rs779083037 Chr.8: 66080365-66080365 on Build GRCh38
rs755903106 Chr.8: 66080563-66080563 on Build GRCh38
rs367555846 Chr.8: 66080455-66080455 on Build GRCh38
rs974018711 Chr.8: 66080428-66080428 on Build GRCh38
rs267601967 Chr.8: 66080542-66080542 on Build GRCh38
rs749952458 Chr.8: 66080265-66080265 on Build GRCh38
rs7005882 Chr.8: 66080275-66080275 on Build GRCh38
rs777783426 Chr.8: 66080442-66080444 on Build GRCh38
rs769072687 Chr.8: 66080496-66080496 on Build GRCh38
rs7006394 Chr.8: 66080722-66080722 on Build GRCh38
rs571495249 Chr.8: 66080498-66080498 on Build GRCh38
rs11324426 Chr.8: 66080636-66080636 on Build GRCh38
rs184274551 Chr.8: 66080632-66080632 on Build GRCh38
rs769470113 Chr.8: 66080635-66080636 on Build GRCh38
rs752468232 Chr.8: 66080561-66080561 on Build GRCh38
rs754020297 Chr.8: 66080464-66080464 on Build GRCh38
rs962705246 Chr.8: 66080427-66080427 on Build GRCh38
rs200440195 Chr.8: 66080405-66080405 on Build GRCh38
rs192091139 Chr.8: 66080612-66080612 on Build GRCh38
rs1029548771 Chr.8: 66080633-66080633 on Build GRCh38
rs531830481 Chr.8: 66080682-66080682 on Build GRCh38
rs770932147 Chr.8: 66080355-66080355 on Build GRCh38
rs1020934221 Chr.8: 66080549-66080549 on Build GRCh38
rs918574363 Chr.8: 66080434-66080434 on Build GRCh38
rs370767323 Chr.8: 66080584-66080584 on Build GRCh38
rs1036678444 Chr.8: 66080631-66080631 on Build GRCh38
rs774210595 Chr.8: 66080556-66080556 on Build GRCh38
rs7006209 Chr.8: 66080502-66080502 on Build GRCh38
rs1013575812 Chr.8: 66080518-66080518 on Build GRCh38
rs905817854 Chr.8: 66080670-66080670 on Build GRCh38
rs746313110 Chr.8: 66080382-66080382 on Build GRCh38
rs751328445 Chr.8: 66080334-66080334 on Build GRCh38
rs556303518 Chr.8: 66080619-66080619 on Build GRCh38
rs925473202 Chr.8: 66080316-66080316 on Build GRCh38
rs897058428 Chr.8: 66080439-66080439 on Build GRCh38
rs772177656 Chr.8: 66080382-66080383 on Build GRCh38
rs762414366 Chr.8: 66080410-66080410 on Build GRCh38
rs78575551 Chr.8: 66080469-66080469 on Build GRCh38
rs551707091 Chr.8: 66080488-66080488 on Build GRCh38
rs769462328 Chr.8: 66080501-66080501 on Build GRCh38
rs779053977 Chr.8: 66080466-66080466 on Build GRCh38
rs777961233 Chr.8: 66080342-66080342 on Build GRCh38
rs962797834 Chr.8: 66080698-66080698 on Build GRCh38
rs1014104884 Chr.8: 66080687-66080687 on Build GRCh38
rs999931350 Chr.8: 66080595-66080595 on Build GRCh38
rs143168011 Chr.8: 66080470-66080470 on Build GRCh38
rs747204756 Chr.8: 66080543-66080543 on Build GRCh38
rs747474547 Chr.8: 66080588-66080588 on Build GRCh38
rs749528340 Chr.8: 66080351-66080351 on Build GRCh38
rs943812440 Chr.8: 66080629-66080629 on Build GRCh38
rs764610686 Chr.8: 66080406-66080406 on Build GRCh38
rs1044132936 Chr.8: 66080263-66080263 on Build GRCh38
rs754713862 Chr.8: 66080447-66080447 on Build GRCh38
rs945659256 Chr.8: 66080302-66080302 on Build GRCh38
rs765824449 Chr.8: 66080533-66080533 on Build GRCh38
rs912172304 Chr.8: 66080371-66080371 on Build GRCh38
rs755561301 Chr.8: 66080598-66080598 on Build GRCh38
rs184897451 Chr.8: 66080385-66080385 on Build GRCh38
rs377233329 Chr.8: 66080391-66080391 on Build GRCh38
rs767251279 Chr.8: 66080558-66080558 on Build GRCh38
rs925421030 Chr.8: 66080573-66080573 on Build GRCh38
rs761724249 Chr.8: 66080603-66080603 on Build GRCh38

View Full Product Details